Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 92384584-92385123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPP
Entrez Gene ID
401541
Gene Name
centromere protein P
Gene Aliases
CENP-P
UniGene
Hs.713775
Gene Chromosome Location
Chr.9: 92325457-92615164 on Build GRCh38
Transcripts
NM_001012267.2 NM_001286969.1 XM_011518689.1 NM_014057.4 NM_024416.4 NM_033014.3

Gene Symbol
OGN
Entrez Gene ID
4969
Gene Name
osteoglycin
Gene Aliases
OG, OIF, SLRR3A
UniGene
Hs.109439
Gene Chromosome Location
Chr.9: 92383271-92404699 on Build GRCh38
Transcripts
NM_001012267.2 NM_001286969.1 XM_011518689.1 NM_014057.4 NM_024416.4 NM_033014.3

SNP Detail

SNP ID Chromosome Location
rs894423451 Chr.9: 92384868-92384868 on Build GRCh38
rs913609619 Chr.9: 92384851-92384851 on Build GRCh38
rs906941883 Chr.9: 92384888-92384888 on Build GRCh38
rs767186135 Chr.9: 92384937-92384937 on Build GRCh38
rs780004291 Chr.9: 92384887-92384887 on Build GRCh38
rs897271515 Chr.9: 92384648-92384648 on Build GRCh38
rs934109813 Chr.9: 92384761-92384761 on Build GRCh38
rs543086251 Chr.9: 92384797-92384797 on Build GRCh38
rs578048491 Chr.9: 92384878-92384878 on Build GRCh38
rs757683225 Chr.9: 92384809-92384809 on Build GRCh38
rs543014116 Chr.9: 92384681-92384681 on Build GRCh38
rs1041685279 Chr.9: 92384763-92384763 on Build GRCh38
rs927633849 Chr.9: 92384909-92384909 on Build GRCh38
rs897208917 Chr.9: 92385054-92385054 on Build GRCh38
rs527636146 Chr.9: 92385060-92385060 on Build GRCh38
rs1012683097 Chr.9: 92384871-92384871 on Build GRCh38
rs989756829 Chr.9: 92384843-92384843 on Build GRCh38
rs1039924647 Chr.9: 92384706-92384706 on Build GRCh38
rs993663753 Chr.9: 92384913-92384913 on Build GRCh38
rs545447053 Chr.9: 92384967-92384967 on Build GRCh38
rs544036922 Chr.9: 92384803-92384803 on Build GRCh38
rs562680100 Chr.9: 92384833-92384833 on Build GRCh38
rs375353345 Chr.9: 92385043-92385043 on Build GRCh38
rs749989622 Chr.9: 92384805-92384805 on Build GRCh38
rs1011454011 Chr.9: 92384931-92384931 on Build GRCh38
rs758324112 Chr.9: 92384880-92384880 on Build GRCh38
rs984376312 Chr.9: 92384666-92384666 on Build GRCh38
rs3209311 Chr.9: 92385013-92385013 on Build GRCh38
rs190336925 Chr.9: 92384767-92384767 on Build GRCh38
rs373598362 Chr.9: 92384808-92384808 on Build GRCh38
rs45553339 Chr.9: 92384739-92384739 on Build GRCh38
rs756864145 Chr.9: 92384635-92384635 on Build GRCh38
rs910029975 Chr.9: 92384675-92384675 on Build GRCh38
rs954604122 Chr.9: 92384781-92384781 on Build GRCh38
rs957887417 Chr.9: 92384840-92384840 on Build GRCh38
rs781743412 Chr.9: 92384918-92384918 on Build GRCh38
rs941535623 Chr.9: 92385017-92385017 on Build GRCh38
rs1015672247 Chr.9: 92384626-92384626 on Build GRCh38
rs937685764 Chr.9: 92384909-92384909 on Build GRCh38

View Full Product Details