Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 104569082-104569581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR13C8
Entrez Gene ID
138802
Gene Name
olfactory receptor family 13 subfamily C member 8
Gene Aliases
OR37H, OR9-10
UniGene
Hs.553599
Gene Chromosome Location
Chr.9: 104569168-104570130 on Build GRCh38
Transcripts
NM_001004483.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5875271 c.280T>G p.F94V Chr.9: 104569447-104569447 on Build GRCh38
1103485 c.336C>T p.C112C Chr.9: 104569503-104569503 on Build GRCh38
6841502 c.346G>T p.G116C Chr.9: 104569513-104569513 on Build GRCh38
4599498 c.274G>C p.V92L Chr.9: 104569441-104569441 on Build GRCh38
5912939 c.16G>A p.D6N Chr.9: 104569183-104569183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770089876 Chr.9: 104569341-104569341 on Build GRCh38
rs369663795 Chr.9: 104569396-104569396 on Build GRCh38
rs915586331 Chr.9: 104569510-104569510 on Build GRCh38
rs770550248 Chr.9: 104569234-104569234 on Build GRCh38
rs771734811 Chr.9: 104569378-104569378 on Build GRCh38
rs767798778 Chr.9: 104569488-104569488 on Build GRCh38
rs868444855 Chr.9: 104569194-104569194 on Build GRCh38
rs762779018 Chr.9: 104569529-104569529 on Build GRCh38
rs771649107 Chr.9: 104569460-104569460 on Build GRCh38
rs772677139 Chr.9: 104569227-104569227 on Build GRCh38
rs752507265 Chr.9: 104569190-104569190 on Build GRCh38
rs779890472 Chr.9: 104569517-104569517 on Build GRCh38
rs142090161 Chr.9: 104569441-104569441 on Build GRCh38
rs766879087 Chr.9: 104569522-104569522 on Build GRCh38
rs893230520 Chr.9: 104569344-104569344 on Build GRCh38
rs753495338 Chr.9: 104569135-104569136 on Build GRCh38
rs758631231 Chr.9: 104569299-104569299 on Build GRCh38
rs965899891 Chr.9: 104569455-104569455 on Build GRCh38
rs760211614 Chr.9: 104569159-104569159 on Build GRCh38
rs201377165 Chr.9: 104569189-104569189 on Build GRCh38
rs543122936 Chr.9: 104569464-104569464 on Build GRCh38
rs776567596 Chr.9: 104569521-104569521 on Build GRCh38
rs763995213 Chr.9: 104569395-104569395 on Build GRCh38
rs75087819 Chr.9: 104569453-104569453 on Build GRCh38
rs141158932 Chr.9: 104569364-104569364 on Build GRCh38
rs201255016 Chr.9: 104569324-104569324 on Build GRCh38
rs761551947 Chr.9: 104569398-104569398 on Build GRCh38
rs760503352 Chr.9: 104569278-104569278 on Build GRCh38
rs561391168 Chr.9: 104569508-104569508 on Build GRCh38
rs867857374 Chr.9: 104569480-104569480 on Build GRCh38
rs772992048 Chr.9: 104569527-104569527 on Build GRCh38
rs756200270 Chr.9: 104569500-104569500 on Build GRCh38
rs764610178 Chr.9: 104569502-104569502 on Build GRCh38
rs778607270 Chr.9: 104569434-104569435 on Build GRCh38
rs748464867 Chr.9: 104569224-104569224 on Build GRCh38
rs755074602 Chr.9: 104569335-104569335 on Build GRCh38
rs984544900 Chr.9: 104569497-104569497 on Build GRCh38
rs978013778 Chr.9: 104569368-104569368 on Build GRCh38
rs74498060 Chr.9: 104569193-104569193 on Build GRCh38
rs752699615 Chr.9: 104569264-104569264 on Build GRCh38
rs758343445 Chr.9: 104569206-104569206 on Build GRCh38
rs752745726 Chr.9: 104569496-104569496 on Build GRCh38
rs570976035 Chr.9: 104569399-104569399 on Build GRCh38
rs1002915437 Chr.9: 104569284-104569284 on Build GRCh38
rs370298142 Chr.9: 104569176-104569176 on Build GRCh38
rs750553242 Chr.9: 104569288-104569288 on Build GRCh38
rs796473637 Chr.9: 104569223-104569225 on Build GRCh38
rs867329527 Chr.9: 104569171-104569171 on Build GRCh38
rs772874765 Chr.9: 104569156-104569156 on Build GRCh38
rs944419640 Chr.9: 104569185-104569185 on Build GRCh38
rs139563439 Chr.9: 104569435-104569435 on Build GRCh38
rs763984512 Chr.9: 104569279-104569279 on Build GRCh38
rs768605904 Chr.9: 104569469-104569469 on Build GRCh38
rs140558238 Chr.9: 104569518-104569518 on Build GRCh38
rs750684846 Chr.9: 104569199-104569199 on Build GRCh38
rs930401392 Chr.9: 104569468-104569468 on Build GRCh38
rs373254255 Chr.9: 104569515-104569515 on Build GRCh38
rs183100737 Chr.9: 104569457-104569457 on Build GRCh38
rs756611719 Chr.9: 104569436-104569436 on Build GRCh38
rs199619511 Chr.9: 104569277-104569277 on Build GRCh38
rs751638088 Chr.9: 104569327-104569327 on Build GRCh38
rs776457369 Chr.9: 104569473-104569473 on Build GRCh38
rs944601525 Chr.9: 104569388-104569388 on Build GRCh38
rs868190039 Chr.9: 104569456-104569456 on Build GRCh38
rs1049231133 Chr.9: 104569198-104569198 on Build GRCh38
rs754516707 Chr.9: 104569316-104569316 on Build GRCh38
rs780206122 Chr.9: 104569313-104569313 on Build GRCh38
rs75834331 Chr.9: 104569540-104569540 on Build GRCh38
rs7026705 Chr.9: 104569223-104569223 on Build GRCh38
rs79042036 Chr.9: 104569531-104569531 on Build GRCh38
rs754290078 Chr.9: 104569555-104569555 on Build GRCh38
rs766575705 Chr.9: 104569416-104569416 on Build GRCh38
rs759744210 Chr.9: 104569482-104569482 on Build GRCh38
rs778191495 Chr.9: 104569343-104569343 on Build GRCh38
rs759154891 Chr.9: 104569251-104569251 on Build GRCh38
rs751813254 Chr.9: 104569418-104569418 on Build GRCh38
rs575094153 Chr.9: 104569461-104569461 on Build GRCh38
rs757571331 Chr.9: 104569511-104569511 on Build GRCh38
rs369313427 Chr.9: 104569514-104569514 on Build GRCh38
rs78381147 Chr.9: 104569191-104569191 on Build GRCh38
rs770589581 Chr.9: 104569363-104569363 on Build GRCh38
rs150711746 Chr.9: 104569419-104569419 on Build GRCh38
rs767536072 Chr.9: 104569252-104569252 on Build GRCh38
rs1005042695 Chr.9: 104569486-104569486 on Build GRCh38
rs769829143 Chr.9: 104569525-104569525 on Build GRCh38
rs78431564 Chr.9: 104569446-104569446 on Build GRCh38
rs528726403 Chr.9: 104569124-104569124 on Build GRCh38
rs760700371 Chr.9: 104569382-104569382 on Build GRCh38
rs376299203 Chr.9: 104569513-104569513 on Build GRCh38
rs774500176 Chr.9: 104569479-104569479 on Build GRCh38
rs79248671 Chr.9: 104569183-104569183 on Build GRCh38
rs761631239 Chr.9: 104569475-104569475 on Build GRCh38
rs769145127 Chr.9: 104569213-104569213 on Build GRCh38
rs200376236 Chr.9: 104569157-104569157 on Build GRCh38
rs371137098 Chr.9: 104569303-104569303 on Build GRCh38
rs145860428 Chr.9: 104569173-104569173 on Build GRCh38
rs761240395 Chr.9: 104569478-104569478 on Build GRCh38
rs747114447 Chr.9: 104569212-104569212 on Build GRCh38
rs762207559 Chr.9: 104569147-104569147 on Build GRCh38
rs555075613 Chr.9: 104569233-104569233 on Build GRCh38
rs775497787 Chr.9: 104569379-104569379 on Build GRCh38
rs202237752 Chr.9: 104569125-104569125 on Build GRCh38
rs773921698 Chr.9: 104569375-104569375 on Build GRCh38
rs74582350 Chr.9: 104569225-104569225 on Build GRCh38
rs764848633 Chr.9: 104569150-104569150 on Build GRCh38
rs79044641 Chr.9: 104569337-104569337 on Build GRCh38
rs780194572 Chr.9: 104569211-104569211 on Build GRCh38
rs749684568 Chr.9: 104569358-104569358 on Build GRCh38
rs112888510 Chr.9: 104569243-104569243 on Build GRCh38

View Full Product Details