Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 68356918-68357452 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGM5
Entrez Gene ID
5239
Gene Name
phosphoglucomutase 5
Gene Aliases
PGMRP
UniGene
Hs.307835
Gene Chromosome Location
Chr.9: 68356438-68531061 on Build GRCh38
Transcripts
NM_021965.3 XM_011518782.2 XM_011518783.2 XM_017014808.1 NR_015423.2 NR_121191.1 NR_121192.1

Gene Symbol
PGM5-AS1
Entrez Gene ID
572558
Gene Name
PGM5 antisense RNA 1
Gene Aliases
FAM233A
UniGene
Hs.552819
Gene Chromosome Location
Chr.9: 68355164-68357866 on Build GRCh38
Transcripts
NM_021965.3 XM_011518782.2 XM_011518783.2 XM_017014808.1 NR_015423.2 NR_121191.1 NR_121192.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6306984 c.70G>T p.G24C Chr.9: 68357197-68357197 on Build GRCh38
5159479 c.66C>T p.A22A Chr.9: 68357193-68357193 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs782465065 Chr.9: 68357384-68357384 on Build GRCh38
rs79088300 Chr.9: 68357193-68357193 on Build GRCh38
rs189205417 Chr.9: 68356969-68356969 on Build GRCh38
rs781896763 Chr.9: 68357329-68357329 on Build GRCh38
rs563239193 Chr.9: 68357277-68357277 on Build GRCh38
rs192866274 Chr.9: 68357027-68357027 on Build GRCh38
rs570769500 Chr.9: 68356939-68356939 on Build GRCh38
rs782726866 Chr.9: 68357088-68357088 on Build GRCh38
rs565099195 Chr.9: 68357133-68357133 on Build GRCh38
rs577139170 Chr.9: 68357149-68357149 on Build GRCh38
rs782038388 Chr.9: 68357313-68357313 on Build GRCh38
rs548165385 Chr.9: 68357233-68357233 on Build GRCh38
rs10867824 Chr.9: 68357096-68357096 on Build GRCh38
rs183111052 Chr.9: 68357039-68357039 on Build GRCh38
rs782406945 Chr.9: 68357288-68357289 on Build GRCh38
rs2482232 Chr.9: 68357130-68357130 on Build GRCh38
rs782096409 Chr.9: 68357255-68357255 on Build GRCh38
rs373859763 Chr.9: 68357221-68357221 on Build GRCh38
rs2257408 Chr.9: 68357061-68357061 on Build GRCh38
rs782453490 Chr.9: 68357409-68357409 on Build GRCh38
rs572502570 Chr.9: 68357014-68357014 on Build GRCh38
rs781796598 Chr.9: 68357404-68357404 on Build GRCh38
rs529695457 Chr.9: 68357224-68357224 on Build GRCh38
rs541351359 Chr.9: 68357172-68357172 on Build GRCh38
rs782629119 Chr.9: 68357398-68357398 on Build GRCh38
rs552417887 Chr.9: 68357367-68357367 on Build GRCh38
rs782458432 Chr.9: 68357112-68357112 on Build GRCh38
rs782642165 Chr.9: 68357148-68357148 on Build GRCh38
rs782376464 Chr.9: 68357424-68357424 on Build GRCh38
rs782363651 Chr.9: 68357163-68357163 on Build GRCh38
rs782678833 Chr.9: 68357124-68357124 on Build GRCh38
rs782197468 Chr.9: 68357203-68357203 on Build GRCh38
rs782405703 Chr.9: 68357205-68357205 on Build GRCh38
rs782519297 Chr.9: 68357136-68357136 on Build GRCh38
rs376077882 Chr.9: 68356979-68356979 on Build GRCh38
rs782032898 Chr.9: 68357206-68357206 on Build GRCh38
rs2811064 Chr.9: 68356982-68356982 on Build GRCh38
rs782761842 Chr.9: 68357362-68357362 on Build GRCh38
rs782662050 Chr.9: 68357416-68357416 on Build GRCh38
rs55710121 Chr.9: 68357025-68357025 on Build GRCh38
rs782785842 Chr.9: 68357226-68357226 on Build GRCh38
rs554808515 Chr.9: 68357040-68357040 on Build GRCh38
rs2482231 Chr.9: 68357191-68357191 on Build GRCh38
rs534883359 Chr.9: 68356943-68356943 on Build GRCh38
rs782720701 Chr.9: 68357313-68357313 on Build GRCh38
rs530587671 Chr.9: 68357353-68357353 on Build GRCh38
rs782634508 Chr.9: 68357425-68357425 on Build GRCh38
rs765233766 Chr.9: 68356944-68356944 on Build GRCh38

View Full Product Details