Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 124810086-124810578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OLFML2A
Entrez Gene ID
169611
Gene Name
olfactomedin like 2A
Gene Aliases
PRO34319
UniGene
Hs.357004
Gene Chromosome Location
Chr.9: 124777158-124814891 on Build GRCh38
Transcripts
NM_001282715.1 NM_182487.3 XM_005251760.4 XM_006716989.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5951200 c.1024G>A p.G342S Chr.9: 124810119-124810119 on Build GRCh38
3929796 c.1798T>C p.Y600H Chr.9: 124810251-124810251 on Build GRCh38
3654295 c.1086G>A p.G362G Chr.9: 124810181-124810181 on Build GRCh38
4799985 c.1096C>T p.L366L Chr.9: 124810191-124810191 on Build GRCh38
5875531 c.1056G>A p.W352* Chr.9: 124810151-124810151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779101234 Chr.9: 124810269-124810269 on Build GRCh38
rs756480639 Chr.9: 124810262-124810262 on Build GRCh38
rs762355556 Chr.9: 124810284-124810284 on Build GRCh38
rs375314642 Chr.9: 124810119-124810119 on Build GRCh38
rs373271578 Chr.9: 124810314-124810314 on Build GRCh38
rs868364160 Chr.9: 124810201-124810201 on Build GRCh38
rs528691764 Chr.9: 124810118-124810118 on Build GRCh38
rs73669209 Chr.9: 124810421-124810421 on Build GRCh38
rs779002596 Chr.9: 124810308-124810308 on Build GRCh38
rs753469559 Chr.9: 124810167-124810167 on Build GRCh38
rs1004314020 Chr.9: 124810175-124810175 on Build GRCh38
rs761581392 Chr.9: 124810279-124810279 on Build GRCh38
rs758421591 Chr.9: 124810310-124810310 on Build GRCh38
rs1004147465 Chr.9: 124810266-124810266 on Build GRCh38
rs372868161 Chr.9: 124810449-124810449 on Build GRCh38
rs201325396 Chr.9: 124810369-124810369 on Build GRCh38
rs372603527 Chr.9: 124810168-124810168 on Build GRCh38
rs749877608 Chr.9: 124810243-124810243 on Build GRCh38
rs149771487 Chr.9: 124810332-124810332 on Build GRCh38
rs375300859 Chr.9: 124810133-124810133 on Build GRCh38
rs778041459 Chr.9: 124810264-124810264 on Build GRCh38
rs565686022 Chr.9: 124810430-124810430 on Build GRCh38
rs753559501 Chr.9: 124810208-124810208 on Build GRCh38
rs768576456 Chr.9: 124810276-124810276 on Build GRCh38
rs773319843 Chr.9: 124810146-124810146 on Build GRCh38
rs755030817 Chr.9: 124810388-124810388 on Build GRCh38
rs747950766 Chr.9: 124810331-124810331 on Build GRCh38
rs763839265 Chr.9: 124810165-124810165 on Build GRCh38
rs763016440 Chr.9: 124810147-124810147 on Build GRCh38
rs774282004 Chr.9: 124810151-124810151 on Build GRCh38
rs752387481 Chr.9: 124810159-124810159 on Build GRCh38
rs905972598 Chr.9: 124810546-124810546 on Build GRCh38
rs141355595 Chr.9: 124810270-124810270 on Build GRCh38
rs35114578 Chr.9: 124810379-124810379 on Build GRCh38
rs201864898 Chr.9: 124810443-124810443 on Build GRCh38
rs757608633 Chr.9: 124810361-124810361 on Build GRCh38
rs754271636 Chr.9: 124810360-124810360 on Build GRCh38
rs764873477 Chr.9: 124810223-124810223 on Build GRCh38
rs374898874 Chr.9: 124810288-124810288 on Build GRCh38
rs759045202 Chr.9: 124810412-124810412 on Build GRCh38
rs770069153 Chr.9: 124810179-124810179 on Build GRCh38
rs781260837 Chr.9: 124810389-124810389 on Build GRCh38
rs199680116 Chr.9: 124810215-124810215 on Build GRCh38
rs769080092 Chr.9: 124810138-124810138 on Build GRCh38
rs760387398 Chr.9: 124810204-124810204 on Build GRCh38
rs770999605 Chr.9: 124810185-124810185 on Build GRCh38
rs755848099 Chr.9: 124810165-124810165 on Build GRCh38
rs780114840 Chr.9: 124810313-124810313 on Build GRCh38
rs201030938 Chr.9: 124810172-124810172 on Build GRCh38
rs145783276 Chr.9: 124810441-124810441 on Build GRCh38
rs770591877 Chr.9: 124810404-124810404 on Build GRCh38
rs759542809 Chr.9: 124810197-124810197 on Build GRCh38
rs766357503 Chr.9: 124810148-124810148 on Build GRCh38
rs765852707 Chr.9: 124810285-124810285 on Build GRCh38
rs140600560 Chr.9: 124810414-124810414 on Build GRCh38
rs765834626 Chr.9: 124810347-124810347 on Build GRCh38
rs1014869373 Chr.9: 124810526-124810526 on Build GRCh38
rs774372012 Chr.9: 124810194-124810194 on Build GRCh38
rs749077039 Chr.9: 124810455-124810455 on Build GRCh38
rs776796146 Chr.9: 124810142-124810142 on Build GRCh38
rs776606877 Chr.9: 124810325-124810325 on Build GRCh38
rs755686037 Chr.9: 124810122-124810122 on Build GRCh38
rs750015550 Chr.9: 124810281-124810281 on Build GRCh38
rs751060239 Chr.9: 124810421-124810422 on Build GRCh38
rs755547468 Chr.9: 124810247-124810247 on Build GRCh38
rs377560457 Chr.9: 124810351-124810351 on Build GRCh38
rs748040594 Chr.9: 124810392-124810392 on Build GRCh38
rs988686145 Chr.9: 124810108-124810108 on Build GRCh38
rs755118612 Chr.9: 124810438-124810438 on Build GRCh38
rs750754104 Chr.9: 124810428-124810428 on Build GRCh38
rs562272925 Chr.9: 124810206-124810206 on Build GRCh38
rs747392105 Chr.9: 124810137-124810137 on Build GRCh38
rs773016365 Chr.9: 124810395-124810395 on Build GRCh38
rs756570749 Chr.9: 124810297-124810297 on Build GRCh38
rs369037047 Chr.9: 124810265-124810265 on Build GRCh38
rs886973192 Chr.9: 124810467-124810467 on Build GRCh38
rs7020260 Chr.9: 124810431-124810431 on Build GRCh38
rs780021991 Chr.9: 124810372-124810372 on Build GRCh38
rs762207337 Chr.9: 124810458-124810458 on Build GRCh38
rs757514051 Chr.9: 124810307-124810307 on Build GRCh38
rs781655307 Chr.9: 124810178-124810178 on Build GRCh38
rs754784787 Chr.9: 124810115-124810115 on Build GRCh38
rs760856868 Chr.9: 124810191-124810191 on Build GRCh38
rs751743611 Chr.9: 124810437-124810437 on Build GRCh38
rs776529250 Chr.9: 124810277-124810277 on Build GRCh38
rs986541500 Chr.9: 124810355-124810355 on Build GRCh38
rs769883112 Chr.9: 124810145-124810145 on Build GRCh38
rs773894071 Chr.9: 124810406-124810406 on Build GRCh38
rs369965570 Chr.9: 124810164-124810164 on Build GRCh38
rs762595612 Chr.9: 124810345-124810345 on Build GRCh38
rs777449469 Chr.9: 124810130-124810130 on Build GRCh38
rs148536738 Chr.9: 124810503-124810503 on Build GRCh38
rs1037565208 Chr.9: 124810491-124810491 on Build GRCh38
rs35972823 Chr.9: 124810271-124810271 on Build GRCh38
rs769476951 Chr.9: 124810393-124810393 on Build GRCh38
rs781545777 Chr.9: 124810248-124810248 on Build GRCh38
rs748788898 Chr.9: 124810131-124810131 on Build GRCh38
rs773859088 Chr.9: 124810350-124810350 on Build GRCh38
rs765556048 Chr.9: 124810425-124810425 on Build GRCh38
rs759322338 Chr.9: 124810158-124810158 on Build GRCh38
rs753162616 Chr.9: 124810253-124810253 on Build GRCh38
rs577158818 Chr.9: 124810538-124810538 on Build GRCh38
rs771089561 Chr.9: 124810268-124810268 on Build GRCh38
rs754150730 Chr.9: 124810280-124810280 on Build GRCh38
rs762319345 Chr.9: 124810424-124810424 on Build GRCh38
rs751418527 Chr.9: 124810113-124810113 on Build GRCh38
rs369456660 Chr.9: 124810120-124810120 on Build GRCh38
rs551016152 Chr.9: 124810298-124810298 on Build GRCh38
rs748912745 Chr.9: 124810403-124810403 on Build GRCh38
rs753145599 Chr.9: 124810291-124810291 on Build GRCh38
rs200430413 Chr.9: 124810199-124810199 on Build GRCh38
rs749450633 Chr.9: 124810181-124810181 on Build GRCh38
rs758202658 Chr.9: 124810450-124810450 on Build GRCh38
rs55672025 Chr.9: 124810413-124810413 on Build GRCh38
rs371938386 Chr.9: 124810121-124810121 on Build GRCh38
rs201152586 Chr.9: 124810200-124810200 on Build GRCh38
rs748423275 Chr.9: 124810144-124810144 on Build GRCh38

View Full Product Details