Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 125236654-125237164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA5
Entrez Gene ID
3309
Gene Name
heat shock protein family A (Hsp70) member 5
Gene Aliases
BIP, GRP78, HEL-S-89n, MIF2
UniGene
Hs.743241
Gene Chromosome Location
Chr.9: 125234848-125241387 on Build GRCh38
Transcripts
NM_005347.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7157801 c.1678G>C p.D560H Chr.9: 125236879-125236879 on Build GRCh38
3432860 c.1609G>T p.E537* Chr.9: 125236948-125236948 on Build GRCh38
7162069 c.1636G>A p.E546K Chr.9: 125236921-125236921 on Build GRCh38
6281149 c.1615G>A p.E539K Chr.9: 125236942-125236942 on Build GRCh38
487040 c.1861A>G p.K621E Chr.9: 125236696-125236696 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552650805 Chr.9: 125236771-125236771 on Build GRCh38
rs761610922 Chr.9: 125236901-125236901 on Build GRCh38
rs61755725 Chr.9: 125236898-125236898 on Build GRCh38
rs772596266 Chr.9: 125236688-125236688 on Build GRCh38
rs767450286 Chr.9: 125236781-125236783 on Build GRCh38
rs775060069 Chr.9: 125236709-125236709 on Build GRCh38
rs375391643 Chr.9: 125236744-125236744 on Build GRCh38
rs768267622 Chr.9: 125236886-125236886 on Build GRCh38
rs570413832 Chr.9: 125236717-125236717 on Build GRCh38
rs779314472 Chr.9: 125236832-125236832 on Build GRCh38
rs762500054 Chr.9: 125236885-125236885 on Build GRCh38
rs747433993 Chr.9: 125236975-125236975 on Build GRCh38
rs771497933 Chr.9: 125236884-125236884 on Build GRCh38
rs767440147 Chr.9: 125236752-125236752 on Build GRCh38
rs760675080 Chr.9: 125236916-125236916 on Build GRCh38
rs755135095 Chr.9: 125236963-125236963 on Build GRCh38
rs781694226 Chr.9: 125237042-125237042 on Build GRCh38
rs759041468 Chr.9: 125236703-125236703 on Build GRCh38
rs374062060 Chr.9: 125236705-125236705 on Build GRCh38
rs77527276 Chr.9: 125236756-125236756 on Build GRCh38
rs151232464 Chr.9: 125237000-125237000 on Build GRCh38
rs774217470 Chr.9: 125236979-125236979 on Build GRCh38
rs753935146 Chr.9: 125236953-125236953 on Build GRCh38
rs768167536 Chr.9: 125237075-125237075 on Build GRCh38
rs748757250 Chr.9: 125237057-125237057 on Build GRCh38
rs56136100 Chr.9: 125236887-125236887 on Build GRCh38
rs773987528 Chr.9: 125237104-125237104 on Build GRCh38
rs1032511460 Chr.9: 125237028-125237028 on Build GRCh38
rs755226649 Chr.9: 125236812-125236812 on Build GRCh38
rs925239059 Chr.9: 125237076-125237076 on Build GRCh38
rs779151361 Chr.9: 125236965-125236965 on Build GRCh38
rs889983653 Chr.9: 125237029-125237029 on Build GRCh38
rs749540875 Chr.9: 125236895-125236895 on Build GRCh38
rs978428907 Chr.9: 125237018-125237018 on Build GRCh38
rs909088557 Chr.9: 125236783-125236783 on Build GRCh38
rs572560070 Chr.9: 125236792-125236792 on Build GRCh38
rs143920039 Chr.9: 125236755-125236755 on Build GRCh38
rs144050975 Chr.9: 125236875-125236875 on Build GRCh38
rs748501134 Chr.9: 125236841-125236841 on Build GRCh38
rs898175617 Chr.9: 125236745-125236745 on Build GRCh38
rs368193721 Chr.9: 125236713-125236713 on Build GRCh38
rs191957514 Chr.9: 125236937-125236937 on Build GRCh38
rs147263674 Chr.9: 125236970-125236970 on Build GRCh38
rs79731156 Chr.9: 125236757-125236757 on Build GRCh38
rs949430875 Chr.9: 125236940-125236940 on Build GRCh38
rs541632037 Chr.9: 125236865-125236865 on Build GRCh38
rs368840257 Chr.9: 125236962-125236962 on Build GRCh38
rs1007136118 Chr.9: 125237085-125237085 on Build GRCh38
rs767220151 Chr.9: 125236904-125236904 on Build GRCh38
rs1005567551 Chr.9: 125236727-125236727 on Build GRCh38
rs750297913 Chr.9: 125236906-125236906 on Build GRCh38
rs139483439 Chr.9: 125237032-125237032 on Build GRCh38
rs35356639 Chr.9: 125236930-125236930 on Build GRCh38
rs533357661 Chr.9: 125236989-125236989 on Build GRCh38
rs371951832 Chr.9: 125236721-125236721 on Build GRCh38
rs777998455 Chr.9: 125236972-125236972 on Build GRCh38
rs148665244 Chr.9: 125236943-125236943 on Build GRCh38
rs756195128 Chr.9: 125236758-125236758 on Build GRCh38
rs139238747 Chr.9: 125236702-125236702 on Build GRCh38
rs936657160 Chr.9: 125237128-125237128 on Build GRCh38
rs758725284 Chr.9: 125236971-125236971 on Build GRCh38
rs773627521 Chr.9: 125236696-125236696 on Build GRCh38
rs61740953 Chr.9: 125237005-125237005 on Build GRCh38

View Full Product Details