Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128165961-128166468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIZ1
Entrez Gene ID
25792
Gene Name
CDKN1A interacting zinc finger protein 1
Gene Aliases
LSFR1, NP94, ZNF356
UniGene
Hs.212395
Gene Chromosome Location
Chr.9: 128166065-128204383 on Build GRCh38
Transcripts
NM_001131015.1 NM_001131016.1 NM_001131017.1 NM_001131018.1 NM_001257975.1 NM_001257976.1 NM_012127.2 XM_005251888.3 XM_005251891.3 XM_005251892.3 XM_005251893.4 XM_006717039.3 XM_011518481.1 XM_011518482.2 XM_011518483.1 XM_017014594.1 XM_017014595.1 XM_017014596.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4814913 c.2731T>C p.S911P Chr.9: 128166241-128166241 on Build GRCh38
6415251 c.2696A>C p.Q899P Chr.9: 128166276-128166276 on Build GRCh38
4163363 c.2513G>T p.S838I Chr.9: 128166381-128166381 on Build GRCh38
6417005 c.2684A>C p.Q895P Chr.9: 128166288-128166288 on Build GRCh38
4163364 c.2510C>T p.P837L Chr.9: 128166384-128166384 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs956240699 Chr.9: 128166424-128166424 on Build GRCh38
rs774961220 Chr.9: 128166199-128166199 on Build GRCh38
rs1000193021 Chr.9: 128166021-128166022 on Build GRCh38
rs192626276 Chr.9: 128166289-128166289 on Build GRCh38
rs959076798 Chr.9: 128166089-128166089 on Build GRCh38
rs555112968 Chr.9: 128166304-128166304 on Build GRCh38
rs984625469 Chr.9: 128166078-128166078 on Build GRCh38
rs1012778680 Chr.9: 128166047-128166047 on Build GRCh38
rs990984939 Chr.9: 128166117-128166117 on Build GRCh38
rs184638839 Chr.9: 128166374-128166374 on Build GRCh38
rs749514915 Chr.9: 128166231-128166231 on Build GRCh38
rs11549260 Chr.9: 128166354-128166354 on Build GRCh38
rs760629916 Chr.9: 128166305-128166305 on Build GRCh38
rs890219762 Chr.9: 128166241-128166241 on Build GRCh38
rs973730685 Chr.9: 128166380-128166380 on Build GRCh38
rs762632388 Chr.9: 128166201-128166201 on Build GRCh38
rs745411073 Chr.9: 128166252-128166252 on Build GRCh38
rs569581982 Chr.9: 128166422-128166422 on Build GRCh38
rs1025173521 Chr.9: 128166360-128166360 on Build GRCh38
rs1051504532 Chr.9: 128166227-128166227 on Build GRCh38
rs917563369 Chr.9: 128166137-128166137 on Build GRCh38
rs761732326 Chr.9: 128166417-128166417 on Build GRCh38
rs767499188 Chr.9: 128166209-128166209 on Build GRCh38
rs533363221 Chr.9: 128166235-128166235 on Build GRCh38
rs986747946 Chr.9: 128166418-128166418 on Build GRCh38
rs745732312 Chr.9: 128166034-128166034 on Build GRCh38
rs12376026 Chr.9: 128166079-128166079 on Build GRCh38
rs756099972 Chr.9: 128166211-128166211 on Build GRCh38
rs775040434 Chr.9: 128166281-128166281 on Build GRCh38
rs1042230806 Chr.9: 128166268-128166268 on Build GRCh38
rs530868239 Chr.9: 128166204-128166204 on Build GRCh38
rs764962208 Chr.9: 128166194-128166194 on Build GRCh38
rs932214542 Chr.9: 128166190-128166190 on Build GRCh38
rs934813268 Chr.9: 128166233-128166233 on Build GRCh38
rs186647759 Chr.9: 128166148-128166148 on Build GRCh38
rs766419405 Chr.9: 128166321-128166321 on Build GRCh38
rs540054846 Chr.9: 128166005-128166005 on Build GRCh38
rs768439923 Chr.9: 128166401-128166401 on Build GRCh38
rs997417730 Chr.9: 128166019-128166019 on Build GRCh38
rs188910316 Chr.9: 128166443-128166443 on Build GRCh38
rs990411829 Chr.9: 128166439-128166439 on Build GRCh38
rs45509191 Chr.9: 128166035-128166035 on Build GRCh38
rs780591748 Chr.9: 128166135-128166135 on Build GRCh38
rs376443811 Chr.9: 128166341-128166341 on Build GRCh38
rs546872134 Chr.9: 128166243-128166243 on Build GRCh38
rs566854296 Chr.9: 128166265-128166265 on Build GRCh38
rs771454057 Chr.9: 128166366-128166366 on Build GRCh38
rs920871023 Chr.9: 128166167-128166167 on Build GRCh38
rs753941795 Chr.9: 128166324-128166324 on Build GRCh38
rs978200196 Chr.9: 128166386-128166386 on Build GRCh38
rs757332837 Chr.9: 128166207-128166207 on Build GRCh38
rs570681166 Chr.9: 128166229-128166229 on Build GRCh38
rs772580580 Chr.9: 128166246-128166246 on Build GRCh38
rs11549264 Chr.9: 128166332-128166332 on Build GRCh38
rs760296322 Chr.9: 128166335-128166335 on Build GRCh38
rs981703874 Chr.9: 128166164-128166164 on Build GRCh38
rs923603851 Chr.9: 128166415-128166415 on Build GRCh38
rs759206711 Chr.9: 128166396-128166396 on Build GRCh38
rs369660377 Chr.9: 128166300-128166300 on Build GRCh38
rs982901354 Chr.9: 128166111-128166111 on Build GRCh38
rs950541895 Chr.9: 128166054-128166054 on Build GRCh38
rs769253600 Chr.9: 128166286-128166286 on Build GRCh38
rs1027919169 Chr.9: 128166395-128166395 on Build GRCh38
rs774295374 Chr.9: 128166297-128166297 on Build GRCh38
rs958597934 Chr.9: 128166024-128166024 on Build GRCh38
rs748442534 Chr.9: 128166245-128166245 on Build GRCh38
rs370138543 Chr.9: 128166441-128166441 on Build GRCh38
rs773659289 Chr.9: 128166251-128166251 on Build GRCh38
rs1004042992 Chr.9: 128166350-128166350 on Build GRCh38
rs746175610 Chr.9: 128166133-128166133 on Build GRCh38
rs750548461 Chr.9: 128166210-128166210 on Build GRCh38
rs1032959588 Chr.9: 128166022-128166022 on Build GRCh38
rs778076408 Chr.9: 128166355-128166355 on Build GRCh38
rs770351790 Chr.9: 128166338-128166338 on Build GRCh38
rs755157078 Chr.9: 128166331-128166331 on Build GRCh38
rs373808543 Chr.9: 128166301-128166301 on Build GRCh38
rs1041726161 Chr.9: 128165986-128165986 on Build GRCh38
rs551056588 Chr.9: 128166205-128166205 on Build GRCh38
rs747516769 Chr.9: 128166362-128166362 on Build GRCh38
rs1017568353 Chr.9: 128165982-128165982 on Build GRCh38
rs1050569115 Chr.9: 128166197-128166197 on Build GRCh38
rs774102626 Chr.9: 128166414-128166414 on Build GRCh38
rs943403155 Chr.9: 128166258-128166258 on Build GRCh38
rs779500279 Chr.9: 128166367-128166367 on Build GRCh38

View Full Product Details