Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 229271257-229271741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB4A
Entrez Gene ID
5867
Gene Name
RAB4A, member RAS oncogene family
Gene Aliases
HRES-1, HRES-1/RAB4, HRES1, RAB4
UniGene
Hs.296169
Gene Chromosome Location
Chr.1: 229271062-229305894 on Build GRCh38
Transcripts
NM_001271998.1 NM_004578.3 NR_073545.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4431438 c.31+4C>A p.? Chr.1: 229271374-229271374 on Build GRCh38
3385878 c.1A>G p.M1V Chr.1: 229271340-229271340 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779584961 Chr.1: 229271672-229271672 on Build GRCh38
rs944521515 Chr.1: 229271704-229271704 on Build GRCh38
rs868156076 Chr.1: 229271374-229271374 on Build GRCh38
rs1042613147 Chr.1: 229271360-229271360 on Build GRCh38
rs7528776 Chr.1: 229271626-229271626 on Build GRCh38
rs999903377 Chr.1: 229271555-229271555 on Build GRCh38
rs1025542368 Chr.1: 229271497-229271497 on Build GRCh38
rs551641598 Chr.1: 229271380-229271380 on Build GRCh38
rs970520335 Chr.1: 229271480-229271480 on Build GRCh38
rs901980629 Chr.1: 229271329-229271329 on Build GRCh38
rs565498653 Chr.1: 229271452-229271452 on Build GRCh38
rs902791555 Chr.1: 229271361-229271361 on Build GRCh38
rs895158544 Chr.1: 229271428-229271428 on Build GRCh38
rs909539497 Chr.1: 229271690-229271690 on Build GRCh38
rs750538629 Chr.1: 229271354-229271354 on Build GRCh38
rs779099867 Chr.1: 229271326-229271326 on Build GRCh38
rs375093875 Chr.1: 229271330-229271330 on Build GRCh38
rs937034984 Chr.1: 229271705-229271705 on Build GRCh38
rs983795307 Chr.1: 229271566-229271566 on Build GRCh38
rs12028696 Chr.1: 229271475-229271475 on Build GRCh38
rs7519709 Chr.1: 229271486-229271486 on Build GRCh38
rs942372840 Chr.1: 229271582-229271582 on Build GRCh38
rs531922721 Chr.1: 229271293-229271293 on Build GRCh38
rs1018353822 Chr.1: 229271620-229271620 on Build GRCh38
rs895306126 Chr.1: 229271310-229271310 on Build GRCh38
rs951350263 Chr.1: 229271679-229271679 on Build GRCh38
rs984470261 Chr.1: 229271680-229271680 on Build GRCh38
rs35236671 Chr.1: 229271459-229271459 on Build GRCh38
rs1042971089 Chr.1: 229271666-229271666 on Build GRCh38
rs1032361530 Chr.1: 229271573-229271573 on Build GRCh38
rs755730200 Chr.1: 229271644-229271644 on Build GRCh38
rs910852875 Chr.1: 229271575-229271575 on Build GRCh38
rs965145211 Chr.1: 229271625-229271625 on Build GRCh38
rs757241854 Chr.1: 229271320-229271320 on Build GRCh38
rs1046643417 Chr.1: 229271427-229271427 on Build GRCh38
rs893822401 Chr.1: 229271578-229271578 on Build GRCh38
rs369909116 Chr.1: 229271347-229271347 on Build GRCh38
rs566112772 Chr.1: 229271534-229271534 on Build GRCh38
rs546326727 Chr.1: 229271523-229271523 on Build GRCh38
rs1006963218 Chr.1: 229271595-229271595 on Build GRCh38
rs986374055 Chr.1: 229271434-229271434 on Build GRCh38
rs949568547 Chr.1: 229271331-229271331 on Build GRCh38
rs952778268 Chr.1: 229271520-229271520 on Build GRCh38
rs936886794 Chr.1: 229271685-229271685 on Build GRCh38

View Full Product Details