Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65882524-65883028 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSW
Entrez Gene ID
1521
Gene Name
cathepsin W
Gene Aliases
LYPN
UniGene
Hs.416848
Gene Chromosome Location
Chr.11: 65879813-65883741 on Build GRCh38
Transcripts
NM_001335.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7299795 c.577G>T p.G193C Chr.11: 65882647-65882647 on Build GRCh38
5448239 c.704C>T p.A235V Chr.11: 65882863-65882863 on Build GRCh38
5529392 c.656G>A p.G219D Chr.11: 65882815-65882815 on Build GRCh38
3752646 c.577G>A p.G193S Chr.11: 65882647-65882647 on Build GRCh38
4646372 c.732G>A p.Q244Q Chr.11: 65882891-65882891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142718745 Chr.11: 65882897-65882897 on Build GRCh38
rs753478003 Chr.11: 65882728-65882728 on Build GRCh38
rs143198836 Chr.11: 65882807-65882807 on Build GRCh38
rs762294939 Chr.11: 65882892-65882894 on Build GRCh38
rs761451395 Chr.11: 65882632-65882632 on Build GRCh38
rs773310207 Chr.11: 65882593-65882593 on Build GRCh38
rs780992002 Chr.11: 65882956-65882956 on Build GRCh38
rs371248183 Chr.11: 65882892-65882892 on Build GRCh38
rs767717590 Chr.11: 65882832-65882832 on Build GRCh38
rs756696488 Chr.11: 65882570-65882570 on Build GRCh38
rs879150826 Chr.11: 65882701-65882701 on Build GRCh38
rs994212384 Chr.11: 65882979-65882979 on Build GRCh38
rs746089901 Chr.11: 65882754-65882754 on Build GRCh38
rs540500544 Chr.11: 65882781-65882781 on Build GRCh38
rs753141739 Chr.11: 65882667-65882667 on Build GRCh38
rs1046218558 Chr.11: 65882629-65882629 on Build GRCh38
rs764661300 Chr.11: 65882921-65882921 on Build GRCh38
rs768564165 Chr.11: 65882684-65882684 on Build GRCh38
rs375768698 Chr.11: 65882578-65882578 on Build GRCh38
rs763001396 Chr.11: 65882557-65882557 on Build GRCh38
rs771944144 Chr.11: 65882780-65882780 on Build GRCh38
rs747066710 Chr.11: 65882677-65882677 on Build GRCh38
rs377646629 Chr.11: 65882647-65882647 on Build GRCh38
rs774713967 Chr.11: 65882889-65882896 on Build GRCh38
rs757011700 Chr.11: 65882953-65882953 on Build GRCh38
rs748310981 Chr.11: 65882595-65882595 on Build GRCh38
rs748138206 Chr.11: 65882546-65882546 on Build GRCh38
rs779362414 Chr.11: 65882591-65882591 on Build GRCh38
rs756344680 Chr.11: 65882756-65882756 on Build GRCh38
rs751543309 Chr.11: 65882951-65882951 on Build GRCh38
rs200545067 Chr.11: 65882930-65882930 on Build GRCh38
rs752104783 Chr.11: 65882815-65882815 on Build GRCh38
rs759436373 Chr.11: 65882548-65882548 on Build GRCh38
rs764625596 Chr.11: 65882856-65882856 on Build GRCh38
rs749149766 Chr.11: 65882683-65882683 on Build GRCh38
rs780292297 Chr.11: 65882884-65882884 on Build GRCh38
rs753051901 Chr.11: 65882736-65882736 on Build GRCh38
rs200723179 Chr.11: 65882678-65882678 on Build GRCh38
rs939206582 Chr.11: 65882911-65882911 on Build GRCh38
rs770425141 Chr.11: 65882798-65882798 on Build GRCh38
rs201310333 Chr.11: 65882903-65882903 on Build GRCh38
rs140611694 Chr.11: 65882627-65882627 on Build GRCh38
rs909801720 Chr.11: 65882603-65882603 on Build GRCh38
rs34335714 Chr.11: 65882779-65882779 on Build GRCh38
rs767784015 Chr.11: 65882908-65882909 on Build GRCh38
rs374328043 Chr.11: 65882908-65882908 on Build GRCh38
rs955261783 Chr.11: 65883000-65883000 on Build GRCh38
rs200255094 Chr.11: 65882909-65882909 on Build GRCh38
rs959247627 Chr.11: 65882692-65882692 on Build GRCh38
rs775452812 Chr.11: 65882615-65882615 on Build GRCh38
rs778593957 Chr.11: 65882762-65882762 on Build GRCh38
rs749890812 Chr.11: 65882655-65882655 on Build GRCh38
rs772977511 Chr.11: 65882724-65882725 on Build GRCh38
rs759086234 Chr.11: 65882859-65882859 on Build GRCh38
rs372724815 Chr.11: 65882929-65882929 on Build GRCh38
rs754243026 Chr.11: 65882662-65882662 on Build GRCh38
rs150421438 Chr.11: 65882656-65882656 on Build GRCh38
rs763304872 Chr.11: 65882915-65882915 on Build GRCh38
rs764078565 Chr.11: 65882567-65882567 on Build GRCh38
rs549142832 Chr.11: 65882605-65882605 on Build GRCh38
rs762247589 Chr.11: 65882925-65882925 on Build GRCh38
rs1039211827 Chr.11: 65882994-65882994 on Build GRCh38
rs766070620 Chr.11: 65882720-65882720 on Build GRCh38
rs773286144 Chr.11: 65882547-65882547 on Build GRCh38
rs757770389 Chr.11: 65882817-65882817 on Build GRCh38
rs750729700 Chr.11: 65882942-65882959 on Build GRCh38
rs372329192 Chr.11: 65882602-65882602 on Build GRCh38
rs747942291 Chr.11: 65882890-65882890 on Build GRCh38
rs768035105 Chr.11: 65882926-65882926 on Build GRCh38
rs764857527 Chr.11: 65882914-65882914 on Build GRCh38
rs759024429 Chr.11: 65882817-65882818 on Build GRCh38
rs750037258 Chr.11: 65882571-65882571 on Build GRCh38
rs746580479 Chr.11: 65882784-65882784 on Build GRCh38
rs778126530 Chr.11: 65882601-65882601 on Build GRCh38
rs114732493 Chr.11: 65882619-65882619 on Build GRCh38
rs776054466 Chr.11: 65882802-65882802 on Build GRCh38
rs606830 Chr.11: 65882812-65882812 on Build GRCh38
rs41302419 Chr.11: 65882676-65882676 on Build GRCh38
rs771762277 Chr.11: 65882652-65882654 on Build GRCh38
rs776144235 Chr.11: 65882907-65882907 on Build GRCh38
rs770799333 Chr.11: 65882905-65882905 on Build GRCh38
rs200117928 Chr.11: 65882729-65882729 on Build GRCh38
rs775362604 Chr.11: 65882551-65882551 on Build GRCh38
rs567023032 Chr.11: 65882599-65882599 on Build GRCh38
rs34035680 Chr.11: 65882706-65882706 on Build GRCh38
rs201964451 Chr.11: 65882746-65882746 on Build GRCh38
rs775883600 Chr.11: 65882880-65882880 on Build GRCh38
rs781627115 Chr.11: 65882751-65882751 on Build GRCh38
rs533711333 Chr.11: 65882665-65882665 on Build GRCh38
rs369469906 Chr.11: 65882950-65882950 on Build GRCh38
rs758200815 Chr.11: 65882894-65882894 on Build GRCh38
rs755511300 Chr.11: 65882580-65882580 on Build GRCh38
rs776150365 Chr.11: 65882809-65882810 on Build GRCh38
rs79769976 Chr.11: 65882747-65882747 on Build GRCh38
rs576058318 Chr.11: 65882735-65882735 on Build GRCh38
rs746656491 Chr.11: 65882758-65882761 on Build GRCh38
rs765122797 Chr.11: 65882549-65882549 on Build GRCh38
rs755315668 Chr.11: 65882664-65882664 on Build GRCh38
rs748018695 Chr.11: 65882611-65882613 on Build GRCh38
rs772628082 Chr.11: 65882705-65882705 on Build GRCh38
rs755077391 Chr.11: 65882949-65882949 on Build GRCh38
rs201632151 Chr.11: 65882626-65882626 on Build GRCh38
rs896840391 Chr.11: 65882966-65882966 on Build GRCh38
rs770504692 Chr.11: 65882808-65882809 on Build GRCh38
rs764898943 Chr.11: 65882806-65882806 on Build GRCh38
rs751116620 Chr.11: 65882568-65882568 on Build GRCh38
rs1000433510 Chr.11: 65883003-65883003 on Build GRCh38
rs913261216 Chr.11: 65882809-65882809 on Build GRCh38
rs763349460 Chr.11: 65882811-65882811 on Build GRCh38
rs774127579 Chr.11: 65882702-65882702 on Build GRCh38
rs753955168 Chr.11: 65882943-65882943 on Build GRCh38
rs201461378 Chr.11: 65882704-65882704 on Build GRCh38
rs765976779 Chr.11: 65882658-65882658 on Build GRCh38
rs1029424359 Chr.11: 65882997-65882997 on Build GRCh38
rs759202628 Chr.11: 65882804-65882804 on Build GRCh38
rs755724397 Chr.11: 65882577-65882577 on Build GRCh38
rs972899188 Chr.11: 65882559-65882559 on Build GRCh38
rs750480993 Chr.11: 65882852-65882852 on Build GRCh38
rs766310987 Chr.11: 65882940-65882940 on Build GRCh38

View Full Product Details