Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67428904-67429406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS6KB2
Entrez Gene ID
6199
Gene Name
ribosomal protein S6 kinase B2
Gene Aliases
KLS, P70-beta, P70-beta-1, P70-beta-2, S6K-beta2, S6K2, SRK, STK14B, p70(S6K)-beta, p70S6Kb
UniGene
Hs.534345
Gene Chromosome Location
Chr.11: 67428460-67435408 on Build GRCh38
Transcripts
NM_003952.2 XM_005274164.1 XM_005274165.4 XM_006718655.3 XM_006718656.3 XM_006718657.1 XM_017018108.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3739251 c.142G>T p.V48L Chr.11: 67429142-67429142 on Build GRCh38
6950902 c.129_130delAC p.H44fs*2 Chr.11: 67429129-67429130 on Build GRCh38
95660 c.198C>A p.C66* Chr.11: 67429198-67429198 on Build GRCh38
6769592 c.236G>A p.G79D Chr.11: 67429236-67429236 on Build GRCh38
4722757 c.135T>C p.Y45Y Chr.11: 67429135-67429135 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375911451 Chr.11: 67429052-67429052 on Build GRCh38
rs745469039 Chr.11: 67428923-67428923 on Build GRCh38
rs778267371 Chr.11: 67429012-67429012 on Build GRCh38
rs925916708 Chr.11: 67429343-67429343 on Build GRCh38
rs552449411 Chr.11: 67429370-67429370 on Build GRCh38
rs529680841 Chr.11: 67429193-67429193 on Build GRCh38
rs370223731 Chr.11: 67429197-67429197 on Build GRCh38
rs778528616 Chr.11: 67429192-67429192 on Build GRCh38
rs778841712 Chr.11: 67429061-67429061 on Build GRCh38
rs375384876 Chr.11: 67429045-67429045 on Build GRCh38
rs775290362 Chr.11: 67428929-67428929 on Build GRCh38
rs759623354 Chr.11: 67429116-67429116 on Build GRCh38
rs765446490 Chr.11: 67428994-67428994 on Build GRCh38
rs765358313 Chr.11: 67429118-67429118 on Build GRCh38
rs762671473 Chr.11: 67428961-67428961 on Build GRCh38
rs764010513 Chr.11: 67428964-67428964 on Build GRCh38
rs779657734 Chr.11: 67429158-67429158 on Build GRCh38
rs527462988 Chr.11: 67429313-67429313 on Build GRCh38
rs564437531 Chr.11: 67429033-67429033 on Build GRCh38
rs571558437 Chr.11: 67429281-67429281 on Build GRCh38
rs752857156 Chr.11: 67429120-67429120 on Build GRCh38
rs772023583 Chr.11: 67429097-67429097 on Build GRCh38
rs982777833 Chr.11: 67429217-67429217 on Build GRCh38
rs976837922 Chr.11: 67429295-67429295 on Build GRCh38
rs974997211 Chr.11: 67429365-67429365 on Build GRCh38
rs769602600 Chr.11: 67428925-67428925 on Build GRCh38
rs764381535 Chr.11: 67429237-67429237 on Build GRCh38
rs773118951 Chr.11: 67429212-67429212 on Build GRCh38
rs751985817 Chr.11: 67429129-67429129 on Build GRCh38
rs762255901 Chr.11: 67429246-67429246 on Build GRCh38
rs757772646 Chr.11: 67429135-67429135 on Build GRCh38
rs749038713 Chr.11: 67429070-67429070 on Build GRCh38
rs566606831 Chr.11: 67429264-67429264 on Build GRCh38
rs915034326 Chr.11: 67429013-67429013 on Build GRCh38
rs932509384 Chr.11: 67429334-67429334 on Build GRCh38
rs1028152373 Chr.11: 67429219-67429219 on Build GRCh38
rs201078105 Chr.11: 67429241-67429241 on Build GRCh38
rs201720757 Chr.11: 67429044-67429044 on Build GRCh38
rs764343123 Chr.11: 67429124-67429124 on Build GRCh38
rs369666279 Chr.11: 67428984-67428984 on Build GRCh38
rs779561932 Chr.11: 67429054-67429054 on Build GRCh38
rs761854122 Chr.11: 67428981-67428981 on Build GRCh38
rs768381085 Chr.11: 67429181-67429181 on Build GRCh38
rs775686892 Chr.11: 67429226-67429226 on Build GRCh38
rs760177099 Chr.11: 67429063-67429063 on Build GRCh38
rs758160055 Chr.11: 67429211-67429212 on Build GRCh38
rs200927741 Chr.11: 67429072-67429072 on Build GRCh38
rs760670106 Chr.11: 67429220-67429220 on Build GRCh38
rs1043354724 Chr.11: 67429336-67429336 on Build GRCh38
rs776623821 Chr.11: 67429115-67429115 on Build GRCh38
rs375607820 Chr.11: 67429136-67429136 on Build GRCh38
rs373246105 Chr.11: 67428991-67428991 on Build GRCh38
rs755684612 Chr.11: 67429141-67429141 on Build GRCh38
rs778639193 Chr.11: 67429276-67429276 on Build GRCh38
rs368777814 Chr.11: 67429251-67429251 on Build GRCh38
rs777506399 Chr.11: 67429148-67429151 on Build GRCh38
rs763046615 Chr.11: 67429231-67429231 on Build GRCh38
rs546014798 Chr.11: 67429000-67429000 on Build GRCh38
rs550099148 Chr.11: 67429194-67429194 on Build GRCh38
rs747853092 Chr.11: 67429279-67429279 on Build GRCh38
rs748874235 Chr.11: 67429171-67429171 on Build GRCh38
rs769729806 Chr.11: 67429223-67429223 on Build GRCh38
rs758674760 Chr.11: 67429002-67429002 on Build GRCh38
rs922212226 Chr.11: 67429376-67429376 on Build GRCh38
rs1027266907 Chr.11: 67429152-67429152 on Build GRCh38
rs754516115 Chr.11: 67429274-67429274 on Build GRCh38
rs774154588 Chr.11: 67428970-67428970 on Build GRCh38
rs200751735 Chr.11: 67429102-67429102 on Build GRCh38
rs917570 Chr.11: 67429073-67429073 on Build GRCh38
rs763276533 Chr.11: 67429123-67429123 on Build GRCh38
rs756738750 Chr.11: 67429260-67429260 on Build GRCh38
rs753415953 Chr.11: 67429272-67429272 on Build GRCh38
rs780810372 Chr.11: 67429042-67429042 on Build GRCh38
rs751978487 Chr.11: 67429022-67429022 on Build GRCh38
rs750931103 Chr.11: 67429258-67429258 on Build GRCh38
rs906432148 Chr.11: 67429082-67429082 on Build GRCh38

View Full Product Details