Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7504945-7505531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf45
Entrez Gene ID
374877
Gene Name
chromosome 19 open reading frame 45
Gene Aliases
-
UniGene
Hs.631862
Gene Chromosome Location
Chr.19: 7497559-7508457 on Build GRCh38
Transcripts
NM_198534.2 XM_005272473.3 XM_011527993.1 XM_011527995.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6446444 c.799G>A p.A267T Chr.19: 7505340-7505340 on Build GRCh38
5527601 c.835G>A p.G279S Chr.19: 7505376-7505376 on Build GRCh38
1003238 c.800G>A p.G267D Chr.19: 7505383-7505383 on Build GRCh38
4891593 c.829G>A p.E277K Chr.19: 7505412-7505412 on Build GRCh38
4667173 c.744T>C p.Y248Y Chr.19: 7505327-7505327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773620907 Chr.19: 7505284-7505284 on Build GRCh38
rs760002051 Chr.19: 7505422-7505422 on Build GRCh38
rs537584211 Chr.19: 7505069-7505069 on Build GRCh38
rs770631218 Chr.19: 7505301-7505301 on Build GRCh38
rs758266201 Chr.19: 7505445-7505445 on Build GRCh38
rs768308014 Chr.19: 7504988-7504989 on Build GRCh38
rs370927165 Chr.19: 7505423-7505423 on Build GRCh38
rs764777860 Chr.19: 7505504-7505504 on Build GRCh38
rs770130261 Chr.19: 7505274-7505274 on Build GRCh38
rs565322369 Chr.19: 7505303-7505303 on Build GRCh38
rs774682444 Chr.19: 7505459-7505459 on Build GRCh38
rs756215406 Chr.19: 7505450-7505450 on Build GRCh38
rs1014216710 Chr.19: 7504982-7504982 on Build GRCh38
rs756973864 Chr.19: 7505326-7505326 on Build GRCh38
rs996983064 Chr.19: 7505202-7505202 on Build GRCh38
rs371986711 Chr.19: 7505451-7505451 on Build GRCh38
rs776103934 Chr.19: 7505427-7505427 on Build GRCh38
rs368431274 Chr.19: 7505449-7505449 on Build GRCh38
rs199595215 Chr.19: 7504976-7504976 on Build GRCh38
rs755697927 Chr.19: 7505366-7505366 on Build GRCh38
rs557111614 Chr.19: 7505108-7505108 on Build GRCh38
rs965598777 Chr.19: 7505070-7505070 on Build GRCh38
rs940029503 Chr.19: 7504973-7504974 on Build GRCh38
rs752056718 Chr.19: 7505304-7505304 on Build GRCh38
rs1044241356 Chr.19: 7504987-7504987 on Build GRCh38
rs749544184 Chr.19: 7505413-7505413 on Build GRCh38
rs1046967731 Chr.19: 7505426-7505426 on Build GRCh38
rs926046642 Chr.19: 7505329-7505329 on Build GRCh38
rs793873 Chr.19: 7505246-7505246 on Build GRCh38
rs776266850 Chr.19: 7505334-7505334 on Build GRCh38
rs764934152 Chr.19: 7505441-7505441 on Build GRCh38
rs1029363174 Chr.19: 7505239-7505239 on Build GRCh38
rs377746817 Chr.19: 7505292-7505292 on Build GRCh38
rs899557176 Chr.19: 7505146-7505146 on Build GRCh38
rs373129992 Chr.19: 7504989-7504989 on Build GRCh38
rs908438903 Chr.19: 7505432-7505432 on Build GRCh38
rs770039022 Chr.19: 7505410-7505410 on Build GRCh38
rs977007090 Chr.19: 7505079-7505079 on Build GRCh38
rs653751 Chr.19: 7505145-7505145 on Build GRCh38
rs756905262 Chr.19: 7505437-7505437 on Build GRCh38
rs775811909 Chr.19: 7505474-7505474 on Build GRCh38
rs1009866092 Chr.19: 7505031-7505031 on Build GRCh38
rs771366571 Chr.19: 7505415-7505415 on Build GRCh38
rs148157401 Chr.19: 7505377-7505377 on Build GRCh38
rs773528740 Chr.19: 7505412-7505412 on Build GRCh38
rs766988329 Chr.19: 7505322-7505322 on Build GRCh38
rs751690598 Chr.19: 7505195-7505195 on Build GRCh38
rs771276741 Chr.19: 7505458-7505458 on Build GRCh38
rs143810135 Chr.19: 7505370-7505370 on Build GRCh38
rs371253795 Chr.19: 7505464-7505464 on Build GRCh38
rs761179182 Chr.19: 7505478-7505478 on Build GRCh38
rs758261623 Chr.19: 7505391-7505391 on Build GRCh38
rs978921573 Chr.19: 7505318-7505318 on Build GRCh38
rs984192788 Chr.19: 7505132-7505132 on Build GRCh38
rs1036916312 Chr.19: 7504974-7504985 on Build GRCh38
rs760138380 Chr.19: 7505429-7505429 on Build GRCh38
rs763611605 Chr.19: 7505430-7505430 on Build GRCh38
rs763230417 Chr.19: 7505144-7505144 on Build GRCh38
rs774585242 Chr.19: 7505420-7505420 on Build GRCh38
rs745748012 Chr.19: 7505385-7505385 on Build GRCh38
rs374380068 Chr.19: 7505347-7505347 on Build GRCh38
rs949999351 Chr.19: 7505086-7505086 on Build GRCh38
rs371062992 Chr.19: 7504976-7504976 on Build GRCh38
rs761368982 Chr.19: 7505494-7505494 on Build GRCh38
rs1029005235 Chr.19: 7505447-7505447 on Build GRCh38
rs375967023 Chr.19: 7505481-7505481 on Build GRCh38
rs1020560006 Chr.19: 7505477-7505477 on Build GRCh38
rs149405481 Chr.19: 7504976-7504978 on Build GRCh38
rs967960299 Chr.19: 7505306-7505306 on Build GRCh38
rs895736282 Chr.19: 7504977-7504977 on Build GRCh38
rs749456191 Chr.19: 7505456-7505456 on Build GRCh38
rs201352391 Chr.19: 7505392-7505392 on Build GRCh38
rs370107008 Chr.19: 7505317-7505317 on Build GRCh38
rs756993335 Chr.19: 7505374-7505374 on Build GRCh38
rs768459904 Chr.19: 7505318-7505319 on Build GRCh38
rs753520918 Chr.19: 7505373-7505373 on Build GRCh38
rs764943136 Chr.19: 7505394-7505394 on Build GRCh38
rs189398427 Chr.19: 7505142-7505142 on Build GRCh38
rs950051935 Chr.19: 7505411-7505411 on Build GRCh38
rs568734 Chr.19: 7504989-7504989 on Build GRCh38
rs573918943 Chr.19: 7505117-7505117 on Build GRCh38
rs150707140 Chr.19: 7505043-7505043 on Build GRCh38
rs557607367 Chr.19: 7504990-7504990 on Build GRCh38
rs750173695 Chr.19: 7505444-7505444 on Build GRCh38
rs181052021 Chr.19: 7505436-7505436 on Build GRCh38
rs917287384 Chr.19: 7505213-7505213 on Build GRCh38
rs991518668 Chr.19: 7505371-7505371 on Build GRCh38
rs113513342 Chr.19: 7504988-7504988 on Build GRCh38
rs185298941 Chr.19: 7505463-7505463 on Build GRCh38
rs551090645 Chr.19: 7504976-7504977 on Build GRCh38

View Full Product Details