Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41755695-41756196 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JUP
Entrez Gene ID
3728
Gene Name
junction plakoglobin
Gene Aliases
ARVD12, CTNNG, DP3, DPIII, PDGB, PKGB
UniGene
Hs.514174
Gene Chromosome Location
Chr.17: 41754607-41786853 on Build GRCh38
Transcripts
NM_002230.2 NM_021991.2 XM_006721873.2 XM_006721874.2 XM_006721875.1 XM_006721878.1 XM_011524753.2 XM_011524755.1 XM_011524756.1 XM_011524757.2 XM_011524758.1 XM_017024588.1 XM_017024589.1 XM_017024590.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4805465 c.2158A>G p.M720V Chr.17: 41755824-41755824 on Build GRCh38
5386361 c.2083G>A p.D695N Chr.17: 41756178-41756178 on Build GRCh38
3517548 c.2076C>T p.P692P Chr.17: 41756185-41756185 on Build GRCh38
4693934 c.2104C>T p.R702C Chr.17: 41755878-41755878 on Build GRCh38
3517547 c.2130C>T p.P710P Chr.17: 41755852-41755852 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761413148 Chr.17: 41755967-41755967 on Build GRCh38
rs727504940 Chr.17: 41756168-41756168 on Build GRCh38
rs1126821 Chr.17: 41755893-41755893 on Build GRCh38
rs782012233 Chr.17: 41755725-41755725 on Build GRCh38
rs782165431 Chr.17: 41755843-41755843 on Build GRCh38
rs727504485 Chr.17: 41756161-41756161 on Build GRCh38
rs200690479 Chr.17: 41755877-41755877 on Build GRCh38
rs568454747 Chr.17: 41755992-41755992 on Build GRCh38
rs782240573 Chr.17: 41756113-41756113 on Build GRCh38
rs202006846 Chr.17: 41755918-41755918 on Build GRCh38
rs782772448 Chr.17: 41756142-41756142 on Build GRCh38
rs142102308 Chr.17: 41755762-41755762 on Build GRCh38
rs368443574 Chr.17: 41755943-41755943 on Build GRCh38
rs41275669 Chr.17: 41755723-41755723 on Build GRCh38
rs983436296 Chr.17: 41756030-41756030 on Build GRCh38
rs529520010 Chr.17: 41756131-41756131 on Build GRCh38
rs971866982 Chr.17: 41756042-41756042 on Build GRCh38
rs530653041 Chr.17: 41755844-41755844 on Build GRCh38
rs781848758 Chr.17: 41755768-41755768 on Build GRCh38
rs936260883 Chr.17: 41756110-41756110 on Build GRCh38
rs917361425 Chr.17: 41756035-41756035 on Build GRCh38
rs782592198 Chr.17: 41755917-41755917 on Build GRCh38
rs781885294 Chr.17: 41755788-41755788 on Build GRCh38
rs980310597 Chr.17: 41756075-41756075 on Build GRCh38
rs61064630 Chr.17: 41756011-41756012 on Build GRCh38
rs886052917 Chr.17: 41755900-41755900 on Build GRCh38
rs927421117 Chr.17: 41756079-41756079 on Build GRCh38
rs890224256 Chr.17: 41756148-41756148 on Build GRCh38
rs112879398 Chr.17: 41755742-41755742 on Build GRCh38
rs782336064 Chr.17: 41755829-41755829 on Build GRCh38
rs782532135 Chr.17: 41755887-41755887 on Build GRCh38
rs545547083 Chr.17: 41755878-41755878 on Build GRCh38
rs112298260 Chr.17: 41756173-41756173 on Build GRCh38
rs781826116 Chr.17: 41756156-41756156 on Build GRCh38
rs782095299 Chr.17: 41756157-41756157 on Build GRCh38
rs368465276 Chr.17: 41756138-41756138 on Build GRCh38
rs794729027 Chr.17: 41755860-41755860 on Build GRCh38
rs781857012 Chr.17: 41755880-41755880 on Build GRCh38
rs1054675510 Chr.17: 41756121-41756121 on Build GRCh38
rs141295561 Chr.17: 41755804-41755804 on Build GRCh38
rs781804177 Chr.17: 41755860-41755860 on Build GRCh38
rs782693565 Chr.17: 41755854-41755854 on Build GRCh38
rs782345850 Chr.17: 41755795-41755795 on Build GRCh38
rs1009184280 Chr.17: 41755794-41755794 on Build GRCh38
rs782096012 Chr.17: 41755850-41755850 on Build GRCh38
rs782084645 Chr.17: 41755733-41755733 on Build GRCh38
rs950727434 Chr.17: 41756000-41756000 on Build GRCh38
rs199683273 Chr.17: 41755789-41755789 on Build GRCh38
rs781839198 Chr.17: 41755909-41755909 on Build GRCh38
rs782191490 Chr.17: 41755824-41755824 on Build GRCh38
rs546184858 Chr.17: 41755966-41755966 on Build GRCh38
rs782005417 Chr.17: 41755732-41755732 on Build GRCh38
rs782653458 Chr.17: 41755936-41755936 on Build GRCh38
rs772656361 Chr.17: 41756060-41756060 on Build GRCh38
rs782067092 Chr.17: 41756134-41756134 on Build GRCh38
rs782574785 Chr.17: 41755778-41755778 on Build GRCh38
rs371395790 Chr.17: 41755837-41755837 on Build GRCh38
rs200802496 Chr.17: 41756132-41756132 on Build GRCh38
rs151178348 Chr.17: 41755775-41755775 on Build GRCh38
rs371508357 Chr.17: 41755774-41755774 on Build GRCh38
rs782428567 Chr.17: 41755929-41755929 on Build GRCh38
rs200283001 Chr.17: 41755767-41755767 on Build GRCh38
rs367827360 Chr.17: 41755721-41755721 on Build GRCh38
rs782069599 Chr.17: 41755752-41755752 on Build GRCh38
rs1027571736 Chr.17: 41755940-41755940 on Build GRCh38
rs201552065 Chr.17: 41755741-41755741 on Build GRCh38
rs782031395 Chr.17: 41756139-41756139 on Build GRCh38
rs367984959 Chr.17: 41756141-41756141 on Build GRCh38
rs199935213 Chr.17: 41756162-41756162 on Build GRCh38
rs1008749253 Chr.17: 41755874-41755874 on Build GRCh38
rs782647137 Chr.17: 41755791-41755791 on Build GRCh38
rs782261124 Chr.17: 41755793-41755793 on Build GRCh38
rs886052916 Chr.17: 41755810-41755815 on Build GRCh38
rs913479376 Chr.17: 41756125-41756125 on Build GRCh38
rs781967615 Chr.17: 41755819-41755820 on Build GRCh38
rs776307936 Chr.17: 41755913-41755913 on Build GRCh38
rs141010071 Chr.17: 41756007-41756008 on Build GRCh38

View Full Product Details