Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41803206-41803713 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
P3H4
Entrez Gene ID
10609
Gene Name
prolyl 3-hydroxylase family member 4 (non-enzymatic)
Gene Aliases
LEPREL4, NO55, NOL55, SC65
UniGene
Hs.446459
Gene Chromosome Location
Chr.17: 41801952-41812617 on Build GRCh38
Transcripts
NM_006455.2 XM_006721640.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7097305 c.1153C>T p.L385L Chr.17: 41803425-41803425 on Build GRCh38
706348 c.1273G>C p.G425R Chr.17: 41803305-41803305 on Build GRCh38
5910931 c.1234G>A p.G412S Chr.17: 41803344-41803344 on Build GRCh38
3517561 c.1261C>T p.P421S Chr.17: 41803317-41803317 on Build GRCh38
349327 c.1186G>C p.D396H Chr.17: 41803392-41803392 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541534928 Chr.17: 41803299-41803299 on Build GRCh38
rs570356519 Chr.17: 41803501-41803501 on Build GRCh38
rs782470639 Chr.17: 41803324-41803324 on Build GRCh38
rs368780195 Chr.17: 41803402-41803402 on Build GRCh38
rs782421613 Chr.17: 41803463-41803463 on Build GRCh38
rs782557056 Chr.17: 41803237-41803237 on Build GRCh38
rs34539402 Chr.17: 41803375-41803375 on Build GRCh38
rs781926694 Chr.17: 41803293-41803293 on Build GRCh38
rs782664786 Chr.17: 41803329-41803329 on Build GRCh38
rs144294459 Chr.17: 41803635-41803636 on Build GRCh38
rs183381640 Chr.17: 41803300-41803300 on Build GRCh38
rs574846777 Chr.17: 41803350-41803350 on Build GRCh38
rs548624639 Chr.17: 41803493-41803493 on Build GRCh38
rs782304263 Chr.17: 41803362-41803362 on Build GRCh38
rs535125170 Chr.17: 41803651-41803651 on Build GRCh38
rs781875731 Chr.17: 41803409-41803409 on Build GRCh38
rs782363307 Chr.17: 41803469-41803469 on Build GRCh38
rs571053158 Chr.17: 41803589-41803589 on Build GRCh38
rs782543483 Chr.17: 41803261-41803262 on Build GRCh38
rs782087488 Chr.17: 41803366-41803366 on Build GRCh38
rs782668186 Chr.17: 41803242-41803242 on Build GRCh38
rs781886937 Chr.17: 41803441-41803441 on Build GRCh38
rs143451544 Chr.17: 41803357-41803357 on Build GRCh38
rs782716134 Chr.17: 41803393-41803393 on Build GRCh38
rs977945002 Chr.17: 41803549-41803549 on Build GRCh38
rs782657149 Chr.17: 41803460-41803460 on Build GRCh38
rs547044478 Chr.17: 41803446-41803446 on Build GRCh38
rs961884380 Chr.17: 41803240-41803240 on Build GRCh38
rs142371800 Chr.17: 41803378-41803378 on Build GRCh38
rs782467780 Chr.17: 41803453-41803453 on Build GRCh38
rs782217024 Chr.17: 41803265-41803265 on Build GRCh38
rs375207460 Chr.17: 41803464-41803464 on Build GRCh38
rs782519231 Chr.17: 41803432-41803432 on Build GRCh38
rs968689928 Chr.17: 41803541-41803541 on Build GRCh38
rs553461119 Chr.17: 41803662-41803662 on Build GRCh38
rs782785274 Chr.17: 41803283-41803283 on Build GRCh38
rs139476472 Chr.17: 41803294-41803294 on Build GRCh38
rs781846379 Chr.17: 41803323-41803323 on Build GRCh38
rs3809877 Chr.17: 41803259-41803259 on Build GRCh38
rs944203283 Chr.17: 41803565-41803565 on Build GRCh38
rs782138201 Chr.17: 41803476-41803476 on Build GRCh38
rs958587309 Chr.17: 41803513-41803513 on Build GRCh38
rs35908729 Chr.17: 41803636-41803637 on Build GRCh38
rs1028633659 Chr.17: 41803445-41803445 on Build GRCh38
rs921526450 Chr.17: 41803573-41803573 on Build GRCh38
rs200125154 Chr.17: 41803266-41803266 on Build GRCh38
rs116574664 Chr.17: 41803404-41803404 on Build GRCh38
rs947504570 Chr.17: 41803374-41803374 on Build GRCh38
rs200248443 Chr.17: 41803267-41803267 on Build GRCh38
rs782193092 Chr.17: 41803249-41803249 on Build GRCh38
rs781860343 Chr.17: 41803377-41803377 on Build GRCh38
rs782502830 Chr.17: 41803350-41803350 on Build GRCh38
rs559145569 Chr.17: 41803566-41803566 on Build GRCh38
rs782457623 Chr.17: 41803315-41803315 on Build GRCh38
rs371291013 Chr.17: 41803526-41803526 on Build GRCh38
rs782383241 Chr.17: 41803376-41803376 on Build GRCh38
rs373185443 Chr.17: 41803475-41803475 on Build GRCh38
rs782401195 Chr.17: 41803276-41803276 on Build GRCh38
rs924688343 Chr.17: 41803553-41803553 on Build GRCh38
rs1048657798 Chr.17: 41803603-41803603 on Build GRCh38
rs199650058 Chr.17: 41803462-41803462 on Build GRCh38
rs139076771 Chr.17: 41803506-41803506 on Build GRCh38
rs887479919 Chr.17: 41803614-41803614 on Build GRCh38
rs376475400 Chr.17: 41803405-41803405 on Build GRCh38
rs114300492 Chr.17: 41803442-41803442 on Build GRCh38
rs782244634 Chr.17: 41803344-41803344 on Build GRCh38
rs146276708 Chr.17: 41803471-41803471 on Build GRCh38
rs782185070 Chr.17: 41803465-41803465 on Build GRCh38
rs928562360 Chr.17: 41803271-41803271 on Build GRCh38
rs991353608 Chr.17: 41803524-41803524 on Build GRCh38
rs144212532 Chr.17: 41803351-41803351 on Build GRCh38
rs782162334 Chr.17: 41803270-41803270 on Build GRCh38
rs782618313 Chr.17: 41803262-41803262 on Build GRCh38
rs782800280 Chr.17: 41803582-41803582 on Build GRCh38
rs781890243 Chr.17: 41803313-41803313 on Build GRCh38
rs537616602 Chr.17: 41803234-41803234 on Build GRCh38
rs782815664 Chr.17: 41803306-41803306 on Build GRCh38
rs782808343 Chr.17: 41803434-41803434 on Build GRCh38
rs782559891 Chr.17: 41803316-41803316 on Build GRCh38
rs56097002 Chr.17: 41803663-41803663 on Build GRCh38

View Full Product Details