Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 35805202-35805690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LHFPL5
Entrez Gene ID
222662
Gene Name
lipoma HMGIC fusion partner-like 5
Gene Aliases
DFNB67, TMHS, dJ510O8.8
UniGene
Hs.367947
Gene Chromosome Location
Chr.6: 35805294-35824075 on Build GRCh38
Transcripts
NM_182548.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3626072 c.11T>A p.L4* Chr.6: 35805681-35805681 on Build GRCh38
3075685 c.19G>A p.A7T Chr.6: 35805689-35805689 on Build GRCh38
5011610 c.20C>T p.A7V Chr.6: 35805690-35805690 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs926288955 Chr.6: 35805248-35805248 on Build GRCh38
rs1041878271 Chr.6: 35805298-35805298 on Build GRCh38
rs950962130 Chr.6: 35805495-35805495 on Build GRCh38
rs1025976477 Chr.6: 35805432-35805432 on Build GRCh38
rs537524542 Chr.6: 35805229-35805229 on Build GRCh38
rs766303751 Chr.6: 35805654-35805654 on Build GRCh38
rs796589167 Chr.6: 35805528-35805528 on Build GRCh38
rs571510002 Chr.6: 35805547-35805547 on Build GRCh38
rs886061340 Chr.6: 35805366-35805366 on Build GRCh38
rs375993440 Chr.6: 35805641-35805641 on Build GRCh38
rs1033624950 Chr.6: 35805573-35805573 on Build GRCh38
rs1053530064 Chr.6: 35805292-35805292 on Build GRCh38
rs962621498 Chr.6: 35805224-35805224 on Build GRCh38
rs547739583 Chr.6: 35805259-35805259 on Build GRCh38
rs936277169 Chr.6: 35805340-35805340 on Build GRCh38
rs778669027 Chr.6: 35805665-35805665 on Build GRCh38
rs886061339 Chr.6: 35805336-35805336 on Build GRCh38
rs1051524808 Chr.6: 35805383-35805383 on Build GRCh38
rs112423472 Chr.6: 35805595-35805595 on Build GRCh38
rs768435908 Chr.6: 35805629-35805629 on Build GRCh38
rs1016793750 Chr.6: 35805501-35805501 on Build GRCh38
rs754711950 Chr.6: 35805659-35805659 on Build GRCh38
rs1039906123 Chr.6: 35805366-35805366 on Build GRCh38
rs770313507 Chr.6: 35805623-35805623 on Build GRCh38
rs183712023 Chr.6: 35805291-35805291 on Build GRCh38
rs540528658 Chr.6: 35805578-35805578 on Build GRCh38
rs906389183 Chr.6: 35805354-35805354 on Build GRCh38
rs998222359 Chr.6: 35805371-35805371 on Build GRCh38
rs753583599 Chr.6: 35805656-35805656 on Build GRCh38
rs960433307 Chr.6: 35805256-35805256 on Build GRCh38
rs536927486 Chr.6: 35805642-35805642 on Build GRCh38
rs529076791 Chr.6: 35805233-35805257 on Build GRCh38
rs377434811 Chr.6: 35805634-35805634 on Build GRCh38
rs760548807 Chr.6: 35805652-35805652 on Build GRCh38
rs41270084 Chr.6: 35805608-35805608 on Build GRCh38
rs12212321 Chr.6: 35805582-35805582 on Build GRCh38
rs945308530 Chr.6: 35805322-35805322 on Build GRCh38
rs749921858 Chr.6: 35805648-35805648 on Build GRCh38
rs146758343 Chr.6: 35805543-35805543 on Build GRCh38
rs67478220 Chr.6: 35805373-35805374 on Build GRCh38
rs887204843 Chr.6: 35805396-35805396 on Build GRCh38
rs569631248 Chr.6: 35805368-35805368 on Build GRCh38
rs143126474 Chr.6: 35805539-35805539 on Build GRCh38
rs1039280264 Chr.6: 35805251-35805251 on Build GRCh38
rs138552162 Chr.6: 35805372-35805373 on Build GRCh38
rs886061342 Chr.6: 35805510-35805510 on Build GRCh38
rs200708531 Chr.6: 35805624-35805624 on Build GRCh38
rs966436836 Chr.6: 35805627-35805627 on Build GRCh38
rs974831362 Chr.6: 35805233-35805233 on Build GRCh38
rs992215817 Chr.6: 35805283-35805283 on Build GRCh38
rs529988516 Chr.6: 35805295-35805295 on Build GRCh38
rs187215821 Chr.6: 35805334-35805334 on Build GRCh38
rs1004258758 Chr.6: 35805455-35805455 on Build GRCh38
rs886061341 Chr.6: 35805375-35805376 on Build GRCh38
rs370222283 Chr.6: 35805570-35805570 on Build GRCh38

View Full Product Details