Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 144397116-144397657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOBOX
Entrez Gene ID
135935
Gene Name
NOBOX oogenesis homeobox
Gene Aliases
OG-2, OG2, OG2X, POF5, TCAG_12042
UniGene
Hs.558628
Gene Chromosome Location
Chr.7: 144396805-144410438 on Build GRCh38
Transcripts
NM_001080413.3 XM_017011742.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5592133 c.1788G>A p.W596* Chr.7: 144397528-144397528 on Build GRCh38
5604246 c.1859G>A p.G620E Chr.7: 144397457-144397457 on Build GRCh38
7314278 c.1999C>A p.P667T Chr.7: 144397317-144397317 on Build GRCh38
3781279 c.1904A>T p.Q635L Chr.7: 144397412-144397412 on Build GRCh38
5581862 c.1953G>A p.G651G Chr.7: 144397363-144397363 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753891345 Chr.7: 144397626-144397626 on Build GRCh38
rs1004789990 Chr.7: 144397551-144397551 on Build GRCh38
rs921281200 Chr.7: 144397597-144397597 on Build GRCh38
rs187097765 Chr.7: 144397390-144397390 on Build GRCh38
rs139083352 Chr.7: 144397467-144397467 on Build GRCh38
rs13240993 Chr.7: 144397318-144397318 on Build GRCh38
rs745952977 Chr.7: 144397552-144397552 on Build GRCh38
rs928531296 Chr.7: 144397631-144397631 on Build GRCh38
rs565523105 Chr.7: 144397466-144397466 on Build GRCh38
rs57306880 Chr.7: 144397163-144397163 on Build GRCh38
rs1054093619 Chr.7: 144397397-144397397 on Build GRCh38
rs760221504 Chr.7: 144397418-144397418 on Build GRCh38
rs141612289 Chr.7: 144397200-144397200 on Build GRCh38
rs774572683 Chr.7: 144397379-144397379 on Build GRCh38
rs758227327 Chr.7: 144397507-144397507 on Build GRCh38
rs75574842 Chr.7: 144397625-144397625 on Build GRCh38
rs191595863 Chr.7: 144397462-144397462 on Build GRCh38
rs1001293169 Chr.7: 144397456-144397456 on Build GRCh38
rs768978008 Chr.7: 144397353-144397353 on Build GRCh38
rs1208216 Chr.7: 144397520-144397520 on Build GRCh38
rs560729054 Chr.7: 144397191-144397191 on Build GRCh38
rs745894153 Chr.7: 144397233-144397233 on Build GRCh38
rs115882574 Chr.7: 144397490-144397490 on Build GRCh38
rs531295691 Chr.7: 144397381-144397381 on Build GRCh38
rs913003885 Chr.7: 144397598-144397598 on Build GRCh38
rs149880524 Chr.7: 144397549-144397549 on Build GRCh38
rs906197613 Chr.7: 144397423-144397423 on Build GRCh38
rs986259540 Chr.7: 144397557-144397557 on Build GRCh38
rs144093209 Chr.7: 144397593-144397593 on Build GRCh38
rs907042627 Chr.7: 144397157-144397158 on Build GRCh38
rs758265577 Chr.7: 144397216-144397216 on Build GRCh38
rs931019585 Chr.7: 144397548-144397548 on Build GRCh38
rs958225897 Chr.7: 144397583-144397583 on Build GRCh38
rs762787878 Chr.7: 144397409-144397409 on Build GRCh38
rs886062037 Chr.7: 144397416-144397416 on Build GRCh38
rs557924552 Chr.7: 144397591-144397591 on Build GRCh38
rs950579992 Chr.7: 144397187-144397187 on Build GRCh38
rs746280975 Chr.7: 144397164-144397164 on Build GRCh38
rs908525914 Chr.7: 144397280-144397280 on Build GRCh38
rs568215435 Chr.7: 144397444-144397444 on Build GRCh38
rs1015340502 Chr.7: 144397207-144397207 on Build GRCh38
rs558592169 Chr.7: 144397485-144397485 on Build GRCh38
rs765954014 Chr.7: 144397457-144397457 on Build GRCh38
rs1018844082 Chr.7: 144397569-144397569 on Build GRCh38
rs777771566 Chr.7: 144397228-144397228 on Build GRCh38
rs377553993 Chr.7: 144397213-144397213 on Build GRCh38
rs923573920 Chr.7: 144397267-144397267 on Build GRCh38
rs954260571 Chr.7: 144397621-144397621 on Build GRCh38
rs556519281 Chr.7: 144397534-144397534 on Build GRCh38
rs866574720 Chr.7: 144397290-144397290 on Build GRCh38
rs530319568 Chr.7: 144397151-144397151 on Build GRCh38
rs762312788 Chr.7: 144397518-144397519 on Build GRCh38
rs116937127 Chr.7: 144397378-144397378 on Build GRCh38
rs776223618 Chr.7: 144397167-144397167 on Build GRCh38
rs989561641 Chr.7: 144397624-144397624 on Build GRCh38
rs976730484 Chr.7: 144397260-144397260 on Build GRCh38
rs769887721 Chr.7: 144397286-144397286 on Build GRCh38
rs891447943 Chr.7: 144397402-144397402 on Build GRCh38
rs932126753 Chr.7: 144397276-144397276 on Build GRCh38
rs146227301 Chr.7: 144397460-144397460 on Build GRCh38
rs976791776 Chr.7: 144397327-144397327 on Build GRCh38
rs560698637 Chr.7: 144397620-144397620 on Build GRCh38
rs773878182 Chr.7: 144397391-144397391 on Build GRCh38
rs77802098 Chr.7: 144397325-144397325 on Build GRCh38
rs749483359 Chr.7: 144397332-144397332 on Build GRCh38
rs1046041630 Chr.7: 144397441-144397441 on Build GRCh38

View Full Product Details