Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 37697874-37698391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF703
Entrez Gene ID
80139
Gene Name
zinc finger protein 703
Gene Aliases
NLZ1, ZEPPO1, ZNF503L, ZPO1
UniGene
Hs.288042
Gene Chromosome Location
Chr.8: 37695751-37700021 on Build GRCh38
Transcripts
NM_025069.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7324517 c.1316G>A p.G439D Chr.8: 37698217-37698217 on Build GRCh38
5738312 c.1453A>G p.T485A Chr.8: 37698354-37698354 on Build GRCh38
5409525 c.1135C>T p.P379S Chr.8: 37698036-37698036 on Build GRCh38
7326541 c.1165_1166ins42 p.A401_H402ins14 Chr.8: 37698066-37698067 on Build GRCh38
3395300 c.1279G>A p.A427T Chr.8: 37698180-37698180 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746718997 Chr.8: 37697898-37697898 on Build GRCh38
rs750405214 Chr.8: 37698005-37698005 on Build GRCh38
rs928433112 Chr.8: 37698254-37698254 on Build GRCh38
rs541419243 Chr.8: 37698066-37698067 on Build GRCh38
rs866600978 Chr.8: 37698352-37698352 on Build GRCh38
rs914750850 Chr.8: 37698308-37698308 on Build GRCh38
rs540084351 Chr.8: 37698244-37698244 on Build GRCh38
rs1006757161 Chr.8: 37698187-37698187 on Build GRCh38
rs747807487 Chr.8: 37698095-37698095 on Build GRCh38
rs533921652 Chr.8: 37698335-37698335 on Build GRCh38
rs549148011 Chr.8: 37698137-37698137 on Build GRCh38
rs567361085 Chr.8: 37698138-37698138 on Build GRCh38
rs527386789 Chr.8: 37698031-37698031 on Build GRCh38
rs918423360 Chr.8: 37698169-37698169 on Build GRCh38
rs761541735 Chr.8: 37697977-37697977 on Build GRCh38
rs759626285 Chr.8: 37697963-37697963 on Build GRCh38
rs866455450 Chr.8: 37698044-37698044 on Build GRCh38
rs929755859 Chr.8: 37698185-37698185 on Build GRCh38
rs1000393554 Chr.8: 37697947-37697947 on Build GRCh38
rs780830067 Chr.8: 37697906-37697906 on Build GRCh38
rs1043584127 Chr.8: 37698163-37698163 on Build GRCh38
rs560055898 Chr.8: 37697970-37697970 on Build GRCh38
rs762668585 Chr.8: 37698256-37698256 on Build GRCh38
rs962583895 Chr.8: 37698208-37698208 on Build GRCh38
rs766316702 Chr.8: 37698261-37698261 on Build GRCh38
rs187255379 Chr.8: 37697905-37697905 on Build GRCh38
rs775611701 Chr.8: 37698180-37698180 on Build GRCh38
rs983573851 Chr.8: 37698205-37698205 on Build GRCh38
rs751073159 Chr.8: 37698077-37698077 on Build GRCh38
rs769758323 Chr.8: 37697931-37697931 on Build GRCh38
rs749688143 Chr.8: 37698363-37698363 on Build GRCh38
rs752919031 Chr.8: 37697975-37697975 on Build GRCh38
rs773896882 Chr.8: 37697945-37697945 on Build GRCh38
rs955369997 Chr.8: 37698182-37698182 on Build GRCh38
rs945174482 Chr.8: 37698154-37698154 on Build GRCh38
rs973996589 Chr.8: 37698227-37698227 on Build GRCh38
rs867525561 Chr.8: 37698312-37698312 on Build GRCh38
rs962361157 Chr.8: 37697910-37697910 on Build GRCh38
rs557868906 Chr.8: 37698248-37698248 on Build GRCh38
rs748473336 Chr.8: 37697911-37697911 on Build GRCh38
rs756099428 Chr.8: 37698098-37698099 on Build GRCh38
rs569464701 Chr.8: 37698209-37698209 on Build GRCh38
rs200028793 Chr.8: 37698266-37698266 on Build GRCh38
rs866343222 Chr.8: 37698346-37698346 on Build GRCh38
rs878922299 Chr.8: 37698300-37698301 on Build GRCh38
rs755787894 Chr.8: 37698344-37698344 on Build GRCh38
rs929683568 Chr.8: 37697948-37697948 on Build GRCh38
rs370375841 Chr.8: 37698299-37698299 on Build GRCh38
rs868822792 Chr.8: 37698357-37698357 on Build GRCh38
rs1029331080 Chr.8: 37698016-37698016 on Build GRCh38
rs868299390 Chr.8: 37697925-37697925 on Build GRCh38
rs1018199608 Chr.8: 37698200-37698200 on Build GRCh38
rs3990472 Chr.8: 37698359-37698360 on Build GRCh38
rs62490660 Chr.8: 37698251-37698251 on Build GRCh38
rs754451955 Chr.8: 37698093-37698093 on Build GRCh38
rs551126114 Chr.8: 37698176-37698176 on Build GRCh38
rs774360079 Chr.8: 37698116-37698116 on Build GRCh38
rs765048667 Chr.8: 37697997-37697997 on Build GRCh38
rs951099652 Chr.8: 37698237-37698237 on Build GRCh38
rs867139900 Chr.8: 37698047-37698047 on Build GRCh38
rs905841745 Chr.8: 37698294-37698294 on Build GRCh38
rs759480984 Chr.8: 37698270-37698270 on Build GRCh38
rs780099353 Chr.8: 37698106-37698106 on Build GRCh38
rs780729368 Chr.8: 37698094-37698094 on Build GRCh38
rs551108251 Chr.8: 37697962-37697962 on Build GRCh38
rs760926948 Chr.8: 37698188-37698188 on Build GRCh38
rs867308634 Chr.8: 37698039-37698039 on Build GRCh38
rs754477003 Chr.8: 37698292-37698292 on Build GRCh38
rs955083332 Chr.8: 37698024-37698024 on Build GRCh38
rs1044686290 Chr.8: 37697920-37697920 on Build GRCh38
rs765070317 Chr.8: 37698221-37698221 on Build GRCh38
rs1011530876 Chr.8: 37698107-37698107 on Build GRCh38
rs956180972 Chr.8: 37697972-37697972 on Build GRCh38
rs997985619 Chr.8: 37698139-37698139 on Build GRCh38
rs1025287328 Chr.8: 37698236-37698236 on Build GRCh38
rs536090559 Chr.8: 37698165-37698165 on Build GRCh38
rs757649928 Chr.8: 37698365-37698365 on Build GRCh38
rs770106867 Chr.8: 37698111-37698111 on Build GRCh38
rs758315707 Chr.8: 37698030-37698030 on Build GRCh38
rs893424509 Chr.8: 37698074-37698074 on Build GRCh38
rs1041666351 Chr.8: 37698117-37698117 on Build GRCh38
rs767033421 Chr.8: 37698322-37698322 on Build GRCh38
rs749878063 Chr.8: 37698136-37698136 on Build GRCh38
rs913846908 Chr.8: 37698142-37698142 on Build GRCh38
rs777504459 Chr.8: 37698347-37698347 on Build GRCh38
rs763532586 Chr.8: 37697946-37697946 on Build GRCh38
rs779605254 Chr.8: 37698041-37698041 on Build GRCh38
rs985076526 Chr.8: 37698032-37698032 on Build GRCh38

View Full Product Details