Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 73447577-73448083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDX4
Entrez Gene ID
1046
Gene Name
caudal type homeobox 4
Gene Aliases
-
UniGene
Hs.553488
Gene Chromosome Location
Chr.X: 73447254-73454585 on Build GRCh38
Transcripts
NM_005193.1

SNP Detail

SNP ID Chromosome Location
rs143815058 Chr.X: 73447613-73447613 on Build GRCh38
rs770253641 Chr.X: 73447681-73447681 on Build GRCh38
rs766014861 Chr.X: 73447712-73447712 on Build GRCh38
rs991685357 Chr.X: 73447865-73447865 on Build GRCh38
rs746259806 Chr.X: 73447800-73447800 on Build GRCh38
rs778033453 Chr.X: 73447617-73447617 on Build GRCh38
rs752673817 Chr.X: 73447734-73447734 on Build GRCh38
rs746199378 Chr.X: 73447675-73447675 on Build GRCh38
rs1042207538 Chr.X: 73447991-73447991 on Build GRCh38
rs557744288 Chr.X: 73448001-73448001 on Build GRCh38
rs940875539 Chr.X: 73447879-73447879 on Build GRCh38
rs761482989 Chr.X: 73447687-73447687 on Build GRCh38
rs766678889 Chr.X: 73447920-73447920 on Build GRCh38
rs760407288 Chr.X: 73447710-73447710 on Build GRCh38
rs776034454 Chr.X: 73447684-73447684 on Build GRCh38
rs765124044 Chr.X: 73447727-73447727 on Build GRCh38
rs757497619 Chr.X: 73447795-73447795 on Build GRCh38
rs773701341 Chr.X: 73447832-73447832 on Build GRCh38
rs747248108 Chr.X: 73447627-73447627 on Build GRCh38
rs759946459 Chr.X: 73448051-73448051 on Build GRCh38
rs932214920 Chr.X: 73447913-73447913 on Build GRCh38
rs1037836619 Chr.X: 73447880-73447880 on Build GRCh38
rs148593166 Chr.X: 73447667-73447667 on Build GRCh38
rs759350426 Chr.X: 73447724-73447724 on Build GRCh38
rs146389652 Chr.X: 73447631-73447631 on Build GRCh38
rs368484635 Chr.X: 73447836-73447836 on Build GRCh38
rs943176033 Chr.X: 73448012-73448012 on Build GRCh38
rs929362411 Chr.X: 73447984-73447984 on Build GRCh38
rs756498283 Chr.X: 73447804-73447804 on Build GRCh38
rs375490577 Chr.X: 73447806-73447806 on Build GRCh38
rs1003698664 Chr.X: 73447816-73447816 on Build GRCh38
rs781445620 Chr.X: 73447666-73447666 on Build GRCh38
rs372093870 Chr.X: 73447905-73447905 on Build GRCh38
rs1038859768 Chr.X: 73448058-73448058 on Build GRCh38
rs769218001 Chr.X: 73447807-73447807 on Build GRCh38
rs1009012375 Chr.X: 73447977-73447977 on Build GRCh38
rs987939845 Chr.X: 73447985-73447985 on Build GRCh38
rs1023195609 Chr.X: 73447911-73447911 on Build GRCh38
rs973394147 Chr.X: 73447951-73447951 on Build GRCh38
rs753593735 Chr.X: 73447713-73447713 on Build GRCh38
rs772788049 Chr.X: 73447696-73447696 on Build GRCh38
rs146702029 Chr.X: 73447932-73447932 on Build GRCh38
rs919243445 Chr.X: 73447957-73447957 on Build GRCh38
rs1010547256 Chr.X: 73447755-73447755 on Build GRCh38
rs751602796 Chr.X: 73447780-73447780 on Build GRCh38
rs778831035 Chr.X: 73447679-73447681 on Build GRCh38
rs747681298 Chr.X: 73447774-73447774 on Build GRCh38
rs899424527 Chr.X: 73447882-73447885 on Build GRCh38
rs771697726 Chr.X: 73447694-73447694 on Build GRCh38
rs201345941 Chr.X: 73447776-73447776 on Build GRCh38

View Full Product Details