Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20779737-20780298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSM3
Entrez Gene ID
6296
Gene Name
acyl-CoA synthetase medium-chain family member 3
Gene Aliases
SA, SAH
UniGene
Hs.706754
Gene Chromosome Location
Chr.16: 20763990-20797157 on Build GRCh38
Transcripts
NM_005622.3 NM_202000.2 XM_011545911.1 XM_017023523.1 XM_017023524.1 NM_080663.2

Gene Symbol
ERI2
Entrez Gene ID
112479
Gene Name
ERI1 exoribonuclease family member 2
Gene Aliases
EXOD1, ZGRF5
UniGene
Hs.248437
Gene Chromosome Location
Chr.16: 20780193-20806473 on Build GRCh38
Transcripts
NM_005622.3 NM_202000.2 XM_011545911.1 XM_017023523.1 XM_017023524.1 NM_080663.2

SNP Detail

SNP ID Chromosome Location
rs1002428369 Chr.16: 20780188-20780188 on Build GRCh38
rs530734131 Chr.16: 20780135-20780135 on Build GRCh38
rs11648370 Chr.16: 20780184-20780184 on Build GRCh38
rs902859086 Chr.16: 20779899-20779899 on Build GRCh38
rs760464303 Chr.16: 20779772-20779773 on Build GRCh38
rs929360782 Chr.16: 20780114-20780114 on Build GRCh38
rs969588356 Chr.16: 20779818-20779818 on Build GRCh38
rs764088294 Chr.16: 20779885-20779885 on Build GRCh38
rs760391796 Chr.16: 20779847-20779847 on Build GRCh38
rs78246163 Chr.16: 20779761-20779761 on Build GRCh38
rs997049363 Chr.16: 20779966-20779966 on Build GRCh38
rs747484753 Chr.16: 20779772-20779774 on Build GRCh38
rs1033786778 Chr.16: 20780056-20780056 on Build GRCh38
rs974124384 Chr.16: 20780055-20780055 on Build GRCh38
rs922945180 Chr.16: 20779790-20779790 on Build GRCh38
rs371082860 Chr.16: 20779874-20779874 on Build GRCh38
rs899225661 Chr.16: 20780044-20780044 on Build GRCh38
rs374448089 Chr.16: 20780267-20780267 on Build GRCh38
rs761351064 Chr.16: 20779937-20779937 on Build GRCh38
rs58469103 Chr.16: 20779908-20779908 on Build GRCh38
rs1024771142 Chr.16: 20779807-20779807 on Build GRCh38
rs1032131949 Chr.16: 20780204-20780204 on Build GRCh38
rs767101261 Chr.16: 20779826-20779826 on Build GRCh38
rs777295993 Chr.16: 20779771-20779772 on Build GRCh38
rs568173724 Chr.16: 20779760-20779760 on Build GRCh38
rs540062664 Chr.16: 20779830-20779830 on Build GRCh38
rs140659228 Chr.16: 20780164-20780164 on Build GRCh38
rs558834892 Chr.16: 20779834-20779834 on Build GRCh38
rs755203216 Chr.16: 20780192-20780192 on Build GRCh38
rs58558409 Chr.16: 20779772-20779772 on Build GRCh38
rs563725365 Chr.16: 20780121-20780121 on Build GRCh38
rs1015561959 Chr.16: 20780045-20780045 on Build GRCh38
rs559466390 Chr.16: 20780012-20780012 on Build GRCh38
rs766154793 Chr.16: 20780154-20780154 on Build GRCh38
rs1054146551 Chr.16: 20779917-20779917 on Build GRCh38
rs376689153 Chr.16: 20780043-20780043 on Build GRCh38
rs1013405922 Chr.16: 20779796-20779796 on Build GRCh38
rs887022767 Chr.16: 20779782-20779782 on Build GRCh38
rs974156107 Chr.16: 20779821-20779821 on Build GRCh38
rs186781242 Chr.16: 20779835-20779835 on Build GRCh38
rs774106565 Chr.16: 20780177-20780177 on Build GRCh38
rs899977885 Chr.16: 20779962-20779962 on Build GRCh38
rs145438997 Chr.16: 20779827-20779827 on Build GRCh38
rs962710708 Chr.16: 20780053-20780053 on Build GRCh38
rs1013458182 Chr.16: 20780159-20780159 on Build GRCh38
rs1030316918 Chr.16: 20779974-20779974 on Build GRCh38
rs575652341 Chr.16: 20780157-20780157 on Build GRCh38
rs1015677750 Chr.16: 20779774-20779774 on Build GRCh38
rs375798927 Chr.16: 20779822-20779822 on Build GRCh38
rs878895277 Chr.16: 20780229-20780229 on Build GRCh38
rs767397994 Chr.16: 20779984-20779984 on Build GRCh38
rs962824796 Chr.16: 20779820-20779820 on Build GRCh38
rs914191705 Chr.16: 20779907-20779907 on Build GRCh38
rs949932303 Chr.16: 20780126-20780126 on Build GRCh38
rs991366775 Chr.16: 20780120-20780120 on Build GRCh38
rs576254166 Chr.16: 20779823-20779823 on Build GRCh38
rs977317760 Chr.16: 20780128-20780128 on Build GRCh38
rs542364646 Chr.16: 20780070-20780070 on Build GRCh38
rs529078039 Chr.16: 20779787-20779789 on Build GRCh38
rs528350618 Chr.16: 20780244-20780244 on Build GRCh38
rs760446951 Chr.16: 20780074-20780074 on Build GRCh38
rs899612676 Chr.16: 20780168-20780168 on Build GRCh38
rs924252783 Chr.16: 20780134-20780134 on Build GRCh38

View Full Product Details