Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32187155-32187663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PBX2
Entrez Gene ID
5089
Gene Name
PBX homeobox 2
Gene Aliases
G17, HOX12, PBX2MHC
UniGene
Hs.509545
Gene Chromosome Location
Chr.6: 32184733-32190186 on Build GRCh38
Transcripts
NM_002586.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4884448 c.1024+1G>T p.? Chr.6: 32187241-32187241 on Build GRCh38
3366460 c.989G>A p.S330N Chr.6: 32187277-32187277 on Build GRCh38
6738460 c.1004delC p.P335fs*65 Chr.6: 32187262-32187262 on Build GRCh38
6471116 c.908A>T p.K303M Chr.6: 32187358-32187358 on Build GRCh38
6389872 c.1022C>T p.A341V Chr.6: 32187244-32187244 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760411922 Chr.6: 32187637-32187637 on Build GRCh38
rs746219137 Chr.6: 32187438-32187438 on Build GRCh38
rs762660661 Chr.6: 32187266-32187266 on Build GRCh38
rs892628344 Chr.6: 32187574-32187574 on Build GRCh38
rs376462410 Chr.6: 32187280-32187281 on Build GRCh38
rs760075442 Chr.6: 32187261-32187261 on Build GRCh38
rs772984194 Chr.6: 32187426-32187427 on Build GRCh38
rs751229633 Chr.6: 32187323-32187323 on Build GRCh38
rs772251003 Chr.6: 32187210-32187210 on Build GRCh38
rs761609684 Chr.6: 32187332-32187332 on Build GRCh38
rs763787702 Chr.6: 32187318-32187318 on Build GRCh38
rs781177001 Chr.6: 32187280-32187280 on Build GRCh38
rs756013391 Chr.6: 32187390-32187390 on Build GRCh38
rs773002174 Chr.6: 32187634-32187634 on Build GRCh38
rs760516888 Chr.6: 32187429-32187430 on Build GRCh38
rs146965329 Chr.6: 32187348-32187348 on Build GRCh38
rs767284310 Chr.6: 32187633-32187633 on Build GRCh38
rs748392602 Chr.6: 32187205-32187205 on Build GRCh38
rs139454692 Chr.6: 32187213-32187216 on Build GRCh38
rs1052580871 Chr.6: 32187561-32187561 on Build GRCh38
rs757178759 Chr.6: 32187631-32187631 on Build GRCh38
rs143029272 Chr.6: 32187278-32187279 on Build GRCh38
rs772033850 Chr.6: 32187300-32187301 on Build GRCh38
rs548074947 Chr.6: 32187284-32187284 on Build GRCh38
rs747332178 Chr.6: 32187300-32187300 on Build GRCh38
rs183881283 Chr.6: 32187305-32187305 on Build GRCh38
rs372029104 Chr.6: 32187397-32187397 on Build GRCh38
rs750313675 Chr.6: 32187350-32187350 on Build GRCh38
rs371557584 Chr.6: 32187473-32187473 on Build GRCh38
rs750366628 Chr.6: 32187283-32187283 on Build GRCh38
rs777448292 Chr.6: 32187316-32187316 on Build GRCh38
rs535104160 Chr.6: 32187435-32187435 on Build GRCh38
rs770275017 Chr.6: 32187442-32187442 on Build GRCh38
rs751338061 Chr.6: 32187274-32187274 on Build GRCh38
rs116295847 Chr.6: 32187406-32187406 on Build GRCh38
rs144536029 Chr.6: 32187302-32187302 on Build GRCh38
rs771379661 Chr.6: 32187239-32187239 on Build GRCh38
rs368138195 Chr.6: 32187403-32187403 on Build GRCh38
rs777868924 Chr.6: 32187416-32187416 on Build GRCh38
rs1003229811 Chr.6: 32187462-32187462 on Build GRCh38
rs761042026 Chr.6: 32187222-32187222 on Build GRCh38
rs189647609 Chr.6: 32187606-32187606 on Build GRCh38
rs749343499 Chr.6: 32187192-32187192 on Build GRCh38
rs778910897 Chr.6: 32187202-32187202 on Build GRCh38
rs368574948 Chr.6: 32187630-32187630 on Build GRCh38
rs777993999 Chr.6: 32187286-32187286 on Build GRCh38
rs485704 Chr.6: 32187493-32187493 on Build GRCh38
rs374809621 Chr.6: 32187605-32187605 on Build GRCh38
rs757153246 Chr.6: 32187275-32187275 on Build GRCh38
rs763653442 Chr.6: 32187273-32187273 on Build GRCh38
rs375618773 Chr.6: 32187585-32187585 on Build GRCh38
rs204994 Chr.6: 32187221-32187221 on Build GRCh38
rs747207411 Chr.6: 32187420-32187420 on Build GRCh38
rs947679838 Chr.6: 32187419-32187419 on Build GRCh38
rs80169725 Chr.6: 32187308-32187308 on Build GRCh38
rs772279858 Chr.6: 32187285-32187285 on Build GRCh38
rs3216796 Chr.6: 32187216-32187219 on Build GRCh38
rs374560583 Chr.6: 32187401-32187401 on Build GRCh38
rs774993601 Chr.6: 32187314-32187314 on Build GRCh38
rs547043112 Chr.6: 32187196-32187196 on Build GRCh38
rs764780868 Chr.6: 32187262-32187262 on Build GRCh38
rs757532970 Chr.6: 32187427-32187427 on Build GRCh38
rs986235471 Chr.6: 32187303-32187303 on Build GRCh38
rs752205434 Chr.6: 32187265-32187265 on Build GRCh38
rs529964370 Chr.6: 32187249-32187249 on Build GRCh38
rs976556563 Chr.6: 32187361-32187361 on Build GRCh38
rs775881359 Chr.6: 32187445-32187445 on Build GRCh38
rs777029225 Chr.6: 32187304-32187304 on Build GRCh38

View Full Product Details