Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43171194-43171783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EBNA1BP2
Entrez Gene ID
10969
Gene Name
EBNA1 binding protein 2
Gene Aliases
EBP2, NOBP, P40
UniGene
Hs.346868
Gene Chromosome Location
Chr.1: 43164174-43172570 on Build GRCh38
Transcripts
NM_001159936.1 NM_006824.2 NR_106791.1

Gene Symbol
MIR6733
Entrez Gene ID
102465439
Gene Name
microRNA 6733
Gene Aliases
hsa-mir-6733
UniGene
Gene Chromosome Location
Chr.1: 43171652-43171712 on Build GRCh38
Transcripts
NM_001159936.1 NM_006824.2 NR_106791.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
350057 c.295G>C p.E99Q Chr.1: 43171507-43171507 on Build GRCh38
4858246 c.402G>T p.E134D Chr.1: 43171565-43171565 on Build GRCh38
7365804 c.227C>G p.P76R Chr.1: 43171575-43171575 on Build GRCh38
6428041 c.158T>C p.L53P Chr.1: 43171644-43171644 on Build GRCh38
6651206 c.357G>A p.L119L Chr.1: 43171610-43171610 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs913339805 Chr.1: 43171403-43171403 on Build GRCh38
rs760553611 Chr.1: 43171685-43171685 on Build GRCh38
rs761396629 Chr.1: 43171566-43171566 on Build GRCh38
rs367548162 Chr.1: 43171502-43171502 on Build GRCh38
rs369261945 Chr.1: 43171651-43171651 on Build GRCh38
rs546876668 Chr.1: 43171242-43171242 on Build GRCh38
rs190321422 Chr.1: 43171508-43171508 on Build GRCh38
rs772972663 Chr.1: 43171462-43171462 on Build GRCh38
rs1053105249 Chr.1: 43171387-43171387 on Build GRCh38
rs774957422 Chr.1: 43171752-43171752 on Build GRCh38
rs114973199 Chr.1: 43171732-43171732 on Build GRCh38
rs767881842 Chr.1: 43171648-43171648 on Build GRCh38
rs149560298 Chr.1: 43171492-43171492 on Build GRCh38
rs78117278 Chr.1: 43171260-43171260 on Build GRCh38
rs764506979 Chr.1: 43171494-43171494 on Build GRCh38
rs774002054 Chr.1: 43171478-43171478 on Build GRCh38
rs1005288910 Chr.1: 43171256-43171256 on Build GRCh38
rs749122367 Chr.1: 43171569-43171569 on Build GRCh38
rs752794195 Chr.1: 43171650-43171650 on Build GRCh38
rs759223045 Chr.1: 43171578-43171578 on Build GRCh38
rs751898157 Chr.1: 43171511-43171511 on Build GRCh38
rs923081319 Chr.1: 43171349-43171349 on Build GRCh38
rs769434283 Chr.1: 43171562-43171562 on Build GRCh38
rs771688176 Chr.1: 43171579-43171579 on Build GRCh38
rs757152736 Chr.1: 43171670-43171670 on Build GRCh38
rs376746332 Chr.1: 43171560-43171560 on Build GRCh38
rs985813126 Chr.1: 43171354-43171354 on Build GRCh38
rs185905912 Chr.1: 43171368-43171368 on Build GRCh38
rs777412671 Chr.1: 43171656-43171656 on Build GRCh38
rs1010620 Chr.1: 43171303-43171303 on Build GRCh38
rs770528240 Chr.1: 43171574-43171574 on Build GRCh38
rs905893353 Chr.1: 43171463-43171463 on Build GRCh38
rs188135793 Chr.1: 43171671-43171671 on Build GRCh38
rs750219222 Chr.1: 43171728-43171728 on Build GRCh38
rs749311002 Chr.1: 43171664-43171664 on Build GRCh38
rs142191050 Chr.1: 43171249-43171249 on Build GRCh38
rs958624040 Chr.1: 43171229-43171229 on Build GRCh38
rs767016854 Chr.1: 43171742-43171742 on Build GRCh38
rs748138570 Chr.1: 43171440-43171440 on Build GRCh38
rs1029320252 Chr.1: 43171652-43171652 on Build GRCh38
rs762245946 Chr.1: 43171591-43171591 on Build GRCh38
rs544292780 Chr.1: 43171607-43171607 on Build GRCh38
rs749963250 Chr.1: 43171618-43171618 on Build GRCh38
rs771506479 Chr.1: 43171722-43171722 on Build GRCh38
rs1015380078 Chr.1: 43171257-43171257 on Build GRCh38
rs773418993 Chr.1: 43171736-43171736 on Build GRCh38
rs1029918005 Chr.1: 43171723-43171723 on Build GRCh38
rs763365878 Chr.1: 43171616-43171616 on Build GRCh38
rs767216171 Chr.1: 43171507-43171507 on Build GRCh38
rs765807678 Chr.1: 43171504-43171504 on Build GRCh38
rs781183366 Chr.1: 43171548-43171548 on Build GRCh38
rs372423504 Chr.1: 43171432-43171432 on Build GRCh38
rs763565824 Chr.1: 43171737-43171737 on Build GRCh38
rs375841312 Chr.1: 43171499-43171499 on Build GRCh38
rs1042109888 Chr.1: 43171521-43171521 on Build GRCh38
rs998224518 Chr.1: 43171606-43171606 on Build GRCh38
rs758522439 Chr.1: 43171510-43171510 on Build GRCh38
rs568444817 Chr.1: 43171331-43171331 on Build GRCh38
rs577666775 Chr.1: 43171725-43171725 on Build GRCh38
rs753789382 Chr.1: 43171749-43171749 on Build GRCh38
rs774033814 Chr.1: 43171476-43171476 on Build GRCh38
rs550680551 Chr.1: 43171379-43171379 on Build GRCh38
rs778441909 Chr.1: 43171687-43171687 on Build GRCh38
rs11559316 Chr.1: 43171575-43171575 on Build GRCh38
rs953993212 Chr.1: 43171351-43171351 on Build GRCh38
rs746633667 Chr.1: 43171728-43171728 on Build GRCh38
rs769747408 Chr.1: 43171457-43171457 on Build GRCh38
rs764950669 Chr.1: 43171503-43171505 on Build GRCh38
rs553342491 Chr.1: 43171438-43171438 on Build GRCh38
rs371499741 Chr.1: 43171699-43171699 on Build GRCh38
rs777769737 Chr.1: 43171568-43171568 on Build GRCh38
rs775086693 Chr.1: 43171583-43171583 on Build GRCh38
rs770096491 Chr.1: 43171417-43171417 on Build GRCh38
rs539456520 Chr.1: 43171398-43171398 on Build GRCh38
rs532974253 Chr.1: 43171222-43171222 on Build GRCh38
rs745416268 Chr.1: 43171714-43171714 on Build GRCh38
rs570478255 Chr.1: 43171395-43171395 on Build GRCh38
rs953882106 Chr.1: 43171705-43171705 on Build GRCh38
rs755220974 Chr.1: 43171515-43171515 on Build GRCh38
rs751669792 Chr.1: 43171633-43171633 on Build GRCh38
rs201475646 Chr.1: 43171602-43171602 on Build GRCh38
rs762554797 Chr.1: 43171468-43171468 on Build GRCh38
rs755057421 Chr.1: 43171643-43171643 on Build GRCh38

View Full Product Details