Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 30462991-30463544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMGB1
Entrez Gene ID
3146
Gene Name
high mobility group box 1
Gene Aliases
HMG-1, HMG1, HMG3, SBP-1
UniGene
Hs.434102
Gene Chromosome Location
Chr.13: 30457916-30617597 on Build GRCh38
Transcripts
NM_001313892.1 NM_001313893.1 NM_002128.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5655361 c.151-2A>T p.? Chr.13: 30463354-30463354 on Build GRCh38
6336690 c.195A>G p.K65K Chr.13: 30463308-30463308 on Build GRCh38
3793236 c.166G>C p.E56Q Chr.13: 30463337-30463337 on Build GRCh38
6388023 c.289A>G p.R97G Chr.13: 30463214-30463214 on Build GRCh38
946621 c.253A>G p.T85A Chr.13: 30463250-30463250 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769337704 Chr.13: 30463173-30463173 on Build GRCh38
rs371450041 Chr.13: 30463387-30463387 on Build GRCh38
rs143387796 Chr.13: 30463233-30463233 on Build GRCh38
rs760440175 Chr.13: 30463212-30463212 on Build GRCh38
rs912454421 Chr.13: 30463432-30463432 on Build GRCh38
rs568042384 Chr.13: 30463475-30463475 on Build GRCh38
rs566370254 Chr.13: 30463101-30463101 on Build GRCh38
rs776336301 Chr.13: 30463440-30463440 on Build GRCh38
rs1044194244 Chr.13: 30463476-30463476 on Build GRCh38
rs535561427 Chr.13: 30463490-30463490 on Build GRCh38
rs1048042168 Chr.13: 30463044-30463044 on Build GRCh38
rs374944331 Chr.13: 30463481-30463481 on Build GRCh38
rs372372227 Chr.13: 30463184-30463184 on Build GRCh38
rs142816732 Chr.13: 30463038-30463038 on Build GRCh38
rs1041418937 Chr.13: 30463135-30463135 on Build GRCh38
rs912774039 Chr.13: 30463305-30463305 on Build GRCh38
rs766586201 Chr.13: 30463506-30463506 on Build GRCh38
rs750030870 Chr.13: 30463388-30463391 on Build GRCh38
rs36106804 Chr.13: 30463036-30463037 on Build GRCh38
rs767679881 Chr.13: 30463227-30463227 on Build GRCh38
rs765418887 Chr.13: 30463079-30463079 on Build GRCh38
rs771928240 Chr.13: 30463156-30463156 on Build GRCh38
rs536197626 Chr.13: 30463504-30463504 on Build GRCh38
rs760890153 Chr.13: 30463502-30463502 on Build GRCh38
rs999806087 Chr.13: 30463158-30463158 on Build GRCh38
rs942506862 Chr.13: 30463083-30463083 on Build GRCh38
rs895734079 Chr.13: 30463462-30463463 on Build GRCh38
rs908220823 Chr.13: 30463399-30463399 on Build GRCh38
rs11546137 Chr.13: 30463283-30463283 on Build GRCh38
rs193056442 Chr.13: 30463383-30463383 on Build GRCh38
rs753926838 Chr.13: 30463509-30463509 on Build GRCh38
rs777739337 Chr.13: 30463368-30463368 on Build GRCh38
rs576510300 Chr.13: 30463059-30463059 on Build GRCh38
rs895661271 Chr.13: 30463103-30463103 on Build GRCh38
rs750614642 Chr.13: 30463499-30463499 on Build GRCh38
rs2388544 Chr.13: 30463210-30463210 on Build GRCh38
rs1048590358 Chr.13: 30463388-30463388 on Build GRCh38
rs780768302 Chr.13: 30463382-30463382 on Build GRCh38
rs763475009 Chr.13: 30463480-30463480 on Build GRCh38
rs983723 Chr.13: 30463308-30463308 on Build GRCh38
rs547646618 Chr.13: 30463105-30463105 on Build GRCh38
rs746817748 Chr.13: 30463380-30463380 on Build GRCh38
rs763365503 Chr.13: 30463511-30463512 on Build GRCh38
rs372814279 Chr.13: 30463423-30463423 on Build GRCh38
rs756214554 Chr.13: 30463262-30463262 on Build GRCh38
rs377254376 Chr.13: 30463513-30463513 on Build GRCh38
rs969425691 Chr.13: 30463191-30463191 on Build GRCh38
rs774815217 Chr.13: 30463489-30463489 on Build GRCh38
rs573372363 Chr.13: 30463092-30463092 on Build GRCh38
rs776047897 Chr.13: 30463176-30463176 on Build GRCh38
rs969248 Chr.13: 30463076-30463076 on Build GRCh38
rs761420045 Chr.13: 30463474-30463474 on Build GRCh38
rs758359066 Chr.13: 30463326-30463326 on Build GRCh38
rs770447854 Chr.13: 30463166-30463166 on Build GRCh38
rs189595132 Chr.13: 30463024-30463024 on Build GRCh38
rs780215433 Chr.13: 30463269-30463269 on Build GRCh38
rs372654406 Chr.13: 30463199-30463199 on Build GRCh38
rs367581777 Chr.13: 30463381-30463381 on Build GRCh38
rs772750574 Chr.13: 30463493-30463493 on Build GRCh38
rs368601923 Chr.13: 30463193-30463193 on Build GRCh38
rs776318545 Chr.13: 30463172-30463172 on Build GRCh38
rs549783365 Chr.13: 30463461-30463461 on Build GRCh38
rs1025665018 Chr.13: 30463412-30463412 on Build GRCh38
rs879049750 Chr.13: 30463463-30463463 on Build GRCh38
rs963850938 Chr.13: 30463447-30463447 on Build GRCh38
rs375874287 Chr.13: 30463370-30463370 on Build GRCh38
rs866251338 Chr.13: 30463421-30463421 on Build GRCh38
rs956895154 Chr.13: 30463248-30463248 on Build GRCh38
rs988415728 Chr.13: 30463290-30463290 on Build GRCh38
rs987101261 Chr.13: 30463046-30463046 on Build GRCh38
rs746679366 Chr.13: 30463162-30463162 on Build GRCh38
rs762127662 Chr.13: 30463224-30463224 on Build GRCh38
rs767919129 Chr.13: 30463496-30463496 on Build GRCh38
rs777909228 Chr.13: 30463514-30463514 on Build GRCh38
rs947064601 Chr.13: 30463125-30463125 on Build GRCh38

View Full Product Details