Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1438678-1439208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS15
Entrez Gene ID
6209
Gene Name
ribosomal protein S15
Gene Aliases
RIG, S15
UniGene
Hs.406683
Gene Chromosome Location
Chr.19: 1438364-1440497 on Build GRCh38
Transcripts
NM_001018.4 NM_001308226.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6370006 c.18G>T p.Q6H Chr.19: 1438821-1438821 on Build GRCh38
5655334 c.73C>T p.L25L Chr.19: 1438876-1438876 on Build GRCh38
5966264 c.45C>A p.F15L Chr.19: 1438848-1438848 on Build GRCh38
3756371 c.63C>T p.D21D Chr.19: 1438866-1438866 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs538748113 Chr.19: 1439116-1439116 on Build GRCh38
rs940839340 Chr.19: 1439136-1439136 on Build GRCh38
rs949885521 Chr.19: 1438751-1438751 on Build GRCh38
rs754757804 Chr.19: 1439030-1439030 on Build GRCh38
rs78277163 Chr.19: 1439075-1439075 on Build GRCh38
rs1028179492 Chr.19: 1438721-1438721 on Build GRCh38
rs949937455 Chr.19: 1439024-1439024 on Build GRCh38
rs769722014 Chr.19: 1438869-1438869 on Build GRCh38
rs988733502 Chr.19: 1438748-1438748 on Build GRCh38
rs966874827 Chr.19: 1439052-1439052 on Build GRCh38
rs889708404 Chr.19: 1439166-1439166 on Build GRCh38
rs932024608 Chr.19: 1439161-1439161 on Build GRCh38
rs553199685 Chr.19: 1438911-1438911 on Build GRCh38
rs990475708 Chr.19: 1439104-1439104 on Build GRCh38
rs780984042 Chr.19: 1438762-1438762 on Build GRCh38
rs774326723 Chr.19: 1438884-1438884 on Build GRCh38
rs978929237 Chr.19: 1438739-1438739 on Build GRCh38
rs527455840 Chr.19: 1439054-1439054 on Build GRCh38
rs920715132 Chr.19: 1439060-1439060 on Build GRCh38
rs778516776 Chr.19: 1439165-1439165 on Build GRCh38
rs916819537 Chr.19: 1438872-1438872 on Build GRCh38
rs775984405 Chr.19: 1438898-1438898 on Build GRCh38
rs765902169 Chr.19: 1438740-1438740 on Build GRCh38
rs754654464 Chr.19: 1438839-1438839 on Build GRCh38
rs1038214393 Chr.19: 1438809-1438809 on Build GRCh38
rs548174520 Chr.19: 1439016-1439016 on Build GRCh38
rs1033207117 Chr.19: 1438963-1438963 on Build GRCh38
rs955726715 Chr.19: 1438979-1438979 on Build GRCh38
rs996122002 Chr.19: 1439008-1439008 on Build GRCh38
rs934347750 Chr.19: 1438742-1438742 on Build GRCh38
rs1037780745 Chr.19: 1439143-1439143 on Build GRCh38
rs900658561 Chr.19: 1439127-1439127 on Build GRCh38
rs772956934 Chr.19: 1438820-1438822 on Build GRCh38
rs370758747 Chr.19: 1438890-1438890 on Build GRCh38
rs189727439 Chr.19: 1439110-1439110 on Build GRCh38
rs762558851 Chr.19: 1438797-1438797 on Build GRCh38
rs781295219 Chr.19: 1439064-1439064 on Build GRCh38
rs779178650 Chr.19: 1439032-1439032 on Build GRCh38
rs1039328851 Chr.19: 1439126-1439126 on Build GRCh38
rs760363066 Chr.19: 1438921-1438961 on Build GRCh38
rs761188519 Chr.19: 1438904-1438904 on Build GRCh38
rs373764728 Chr.19: 1438987-1438987 on Build GRCh38
rs764754977 Chr.19: 1438828-1438828 on Build GRCh38
rs920673569 Chr.19: 1439151-1439151 on Build GRCh38
rs769723903 Chr.19: 1438765-1438765 on Build GRCh38
rs1032761522 Chr.19: 1439165-1439165 on Build GRCh38
rs7251165 Chr.19: 1438866-1438866 on Build GRCh38
rs938080461 Chr.19: 1438801-1438801 on Build GRCh38
rs780893945 Chr.19: 1438860-1438860 on Build GRCh38
rs927125399 Chr.19: 1439036-1439036 on Build GRCh38
rs978519139 Chr.19: 1438950-1438950 on Build GRCh38
rs749309245 Chr.19: 1438878-1438878 on Build GRCh38
rs1000807244 Chr.19: 1439159-1439159 on Build GRCh38
rs148070085 Chr.19: 1438833-1438833 on Build GRCh38
rs753116376 Chr.19: 1438821-1438821 on Build GRCh38
rs571367710 Chr.19: 1439109-1439109 on Build GRCh38
rs988477047 Chr.19: 1438991-1438991 on Build GRCh38
rs890472682 Chr.19: 1439102-1439102 on Build GRCh38
rs757963954 Chr.19: 1438834-1438834 on Build GRCh38
rs779549419 Chr.19: 1438835-1438835 on Build GRCh38
rs964457594 Chr.19: 1438718-1438718 on Build GRCh38
rs532256746 Chr.19: 1438814-1438814 on Build GRCh38
rs751171632 Chr.19: 1438836-1438836 on Build GRCh38
rs970962426 Chr.19: 1439113-1439113 on Build GRCh38
rs955943967 Chr.19: 1439181-1439181 on Build GRCh38
rs1047724792 Chr.19: 1439090-1439090 on Build GRCh38
rs777810579 Chr.19: 1438876-1438876 on Build GRCh38
rs995361749 Chr.19: 1438720-1438720 on Build GRCh38
rs377627432 Chr.19: 1438786-1438786 on Build GRCh38
rs540842874 Chr.19: 1438715-1438715 on Build GRCh38
rs1044170343 Chr.19: 1438779-1438779 on Build GRCh38
rs1028231769 Chr.19: 1439010-1439010 on Build GRCh38
rs564942317 Chr.19: 1438749-1438749 on Build GRCh38
rs746037588 Chr.19: 1438885-1438885 on Build GRCh38
rs770996582 Chr.19: 1438881-1438881 on Build GRCh38
rs771030446 Chr.19: 1438787-1438787 on Build GRCh38
rs982692665 Chr.19: 1439118-1439118 on Build GRCh38
rs73920487 Chr.19: 1439050-1439050 on Build GRCh38
rs761207093 Chr.19: 1438827-1438827 on Build GRCh38
rs746748321 Chr.19: 1438697-1438697 on Build GRCh38
rs759700768 Chr.19: 1438795-1438795 on Build GRCh38
rs916871838 Chr.19: 1439105-1439106 on Build GRCh38
rs774686705 Chr.19: 1438791-1438791 on Build GRCh38
rs1019822754 Chr.19: 1438757-1438757 on Build GRCh38
rs768544407 Chr.19: 1438709-1438709 on Build GRCh38

View Full Product Details