Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 91723557-91724156 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ROR2
Entrez Gene ID
4920
Gene Name
receptor tyrosine kinase like orphan receptor 2
Gene Aliases
BDB, BDB1, NTRKR2
UniGene
Hs.644776
Gene Chromosome Location
Chr.9: 91722596-91950206 on Build GRCh38
Transcripts
NM_004560.3 XM_005252008.4 XM_005252009.3 XM_006717121.3 XM_017014762.1 XM_017014763.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4482424 c.2603C>T p.S868F Chr.9: 91723891-91723891 on Build GRCh38
6407738 c.2816T>C p.V939A Chr.9: 91723678-91723678 on Build GRCh38
48754 c.2482C>A p.P828T Chr.9: 91724012-91724012 on Build GRCh38
6013526 c.2403C>T p.F801F Chr.9: 91724091-91724091 on Build GRCh38
299772 c.2622C>T p.V874V Chr.9: 91723872-91723872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371528214 Chr.9: 91723625-91723625 on Build GRCh38
rs1031370230 Chr.9: 91724074-91724074 on Build GRCh38
rs758447257 Chr.9: 91723640-91723640 on Build GRCh38
rs56158629 Chr.9: 91723908-91723908 on Build GRCh38
rs752344305 Chr.9: 91724022-91724022 on Build GRCh38
rs778811682 Chr.9: 91723769-91723769 on Build GRCh38
rs200296562 Chr.9: 91723849-91723849 on Build GRCh38
rs200867550 Chr.9: 91723874-91723874 on Build GRCh38
rs775640546 Chr.9: 91724005-91724005 on Build GRCh38
rs761223603 Chr.9: 91723622-91723622 on Build GRCh38
rs770272877 Chr.9: 91723875-91723875 on Build GRCh38
rs200396791 Chr.9: 91724043-91724043 on Build GRCh38
rs771461100 Chr.9: 91723973-91723973 on Build GRCh38
rs145213893 Chr.9: 91724016-91724016 on Build GRCh38
rs751576095 Chr.9: 91723726-91723726 on Build GRCh38
rs779837210 Chr.9: 91723846-91723846 on Build GRCh38
rs905958690 Chr.9: 91723941-91723941 on Build GRCh38
rs749837764 Chr.9: 91723963-91723963 on Build GRCh38
rs371866105 Chr.9: 91723708-91723708 on Build GRCh38
rs373798516 Chr.9: 91723621-91723621 on Build GRCh38
rs758244691 Chr.9: 91723928-91723928 on Build GRCh38
rs148220610 Chr.9: 91724040-91724040 on Build GRCh38
rs750609013 Chr.9: 91723709-91723709 on Build GRCh38
rs773779501 Chr.9: 91723877-91723877 on Build GRCh38
rs746756515 Chr.9: 91723743-91723744 on Build GRCh38
rs774123408 Chr.9: 91723614-91723614 on Build GRCh38
rs771336632 Chr.9: 91723884-91723884 on Build GRCh38
rs781565044 Chr.9: 91724109-91724109 on Build GRCh38
rs202213533 Chr.9: 91723796-91723796 on Build GRCh38
rs754605230 Chr.9: 91723950-91723950 on Build GRCh38
rs778233521 Chr.9: 91723962-91723962 on Build GRCh38
rs757170131 Chr.9: 91723826-91723826 on Build GRCh38
rs776978802 Chr.9: 91723793-91723793 on Build GRCh38
rs899154522 Chr.9: 91723581-91723581 on Build GRCh38
rs766587026 Chr.9: 91723802-91723802 on Build GRCh38
rs772269687 Chr.9: 91723987-91723987 on Build GRCh38
rs753279278 Chr.9: 91724083-91724083 on Build GRCh38
rs768268681 Chr.9: 91723792-91723792 on Build GRCh38
rs992732083 Chr.9: 91723985-91723985 on Build GRCh38
rs775209555 Chr.9: 91724137-91724137 on Build GRCh38
rs202072718 Chr.9: 91723951-91723951 on Build GRCh38
rs560157548 Chr.9: 91724063-91724063 on Build GRCh38
rs750939153 Chr.9: 91724092-91724092 on Build GRCh38
rs369727413 Chr.9: 91723838-91723838 on Build GRCh38
rs777259749 Chr.9: 91723694-91723694 on Build GRCh38
rs1012078133 Chr.9: 91723682-91723682 on Build GRCh38
rs374824850 Chr.9: 91723770-91723770 on Build GRCh38
rs113793278 Chr.9: 91723674-91723674 on Build GRCh38
rs146987488 Chr.9: 91724108-91724108 on Build GRCh38
rs759105926 Chr.9: 91723616-91723616 on Build GRCh38
rs374141378 Chr.9: 91724113-91724113 on Build GRCh38
rs765260046 Chr.9: 91723910-91723910 on Build GRCh38
rs752753103 Chr.9: 91723817-91723817 on Build GRCh38
rs367597436 Chr.9: 91723624-91723624 on Build GRCh38
rs372530728 Chr.9: 91723651-91723651 on Build GRCh38
rs771792088 Chr.9: 91723777-91723777 on Build GRCh38
rs758362792 Chr.9: 91723699-91723700 on Build GRCh38
rs947970454 Chr.9: 91724073-91724073 on Build GRCh38
rs751339713 Chr.9: 91723946-91723946 on Build GRCh38
rs773250333 Chr.9: 91724014-91724014 on Build GRCh38
rs780709405 Chr.9: 91723867-91723867 on Build GRCh38
rs369201767 Chr.9: 91723866-91723866 on Build GRCh38
rs777560660 Chr.9: 91723738-91723738 on Build GRCh38
rs777573575 Chr.9: 91723744-91723746 on Build GRCh38
rs185808139 Chr.9: 91724052-91724052 on Build GRCh38
rs774526992 Chr.9: 91723886-91723886 on Build GRCh38
rs534997285 Chr.9: 91723976-91723976 on Build GRCh38
rs368832078 Chr.9: 91723647-91723647 on Build GRCh38
rs1035669376 Chr.9: 91723773-91723773 on Build GRCh38
rs772071230 Chr.9: 91724136-91724136 on Build GRCh38
rs376416766 Chr.9: 91724070-91724070 on Build GRCh38
rs767190671 Chr.9: 91724019-91724019 on Build GRCh38
rs751306692 Chr.9: 91724018-91724018 on Build GRCh38
rs140727324 Chr.9: 91723929-91723929 on Build GRCh38
rs759765174 Chr.9: 91723889-91723889 on Build GRCh38
rs974504456 Chr.9: 91723605-91723605 on Build GRCh38
rs777773974 Chr.9: 91723652-91723652 on Build GRCh38
rs970306168 Chr.9: 91724104-91724104 on Build GRCh38
rs2230578 Chr.9: 91723646-91723646 on Build GRCh38
rs141235720 Chr.9: 91724099-91724099 on Build GRCh38
rs917113282 Chr.9: 91723992-91723992 on Build GRCh38
rs751280391 Chr.9: 91723881-91723881 on Build GRCh38
rs939178685 Chr.9: 91724110-91724110 on Build GRCh38
rs146432734 Chr.9: 91724028-91724028 on Build GRCh38
rs141070315 Chr.9: 91723800-91723800 on Build GRCh38
rs987563826 Chr.9: 91723650-91723650 on Build GRCh38
rs770427359 Chr.9: 91723611-91723611 on Build GRCh38
rs765371195 Chr.9: 91723626-91723626 on Build GRCh38
rs755683185 Chr.9: 91723956-91723956 on Build GRCh38
rs767186362 Chr.9: 91723617-91723617 on Build GRCh38
rs770797925 Chr.9: 91723756-91723758 on Build GRCh38
rs146866887 Chr.9: 91723812-91723812 on Build GRCh38
rs199840505 Chr.9: 91723666-91723666 on Build GRCh38
rs779855786 Chr.9: 91723781-91723781 on Build GRCh38
rs1016479276 Chr.9: 91724017-91724017 on Build GRCh38
rs760794107 Chr.9: 91724009-91724009 on Build GRCh38
rs149826387 Chr.9: 91723810-91723810 on Build GRCh38
rs766605383 Chr.9: 91723724-91723724 on Build GRCh38
rs753371692 Chr.9: 91724027-91724027 on Build GRCh38
rs758757733 Chr.9: 91724095-91724095 on Build GRCh38
rs772604181 Chr.9: 91723934-91723936 on Build GRCh38
rs780298605 Chr.9: 91724034-91724034 on Build GRCh38
rs200752982 Chr.9: 91723703-91723703 on Build GRCh38
rs561004430 Chr.9: 91723600-91723600 on Build GRCh38
rs369150056 Chr.9: 91723890-91723890 on Build GRCh38
rs775582564 Chr.9: 91723900-91723900 on Build GRCh38
rs199855010 Chr.9: 91723712-91723712 on Build GRCh38
rs764317522 Chr.9: 91724010-91724010 on Build GRCh38
rs965137025 Chr.9: 91723684-91723684 on Build GRCh38
rs149056068 Chr.9: 91724033-91724033 on Build GRCh38
rs753864565 Chr.9: 91723745-91723745 on Build GRCh38
rs532042443 Chr.9: 91724112-91724112 on Build GRCh38
rs778409919 Chr.9: 91723685-91723685 on Build GRCh38
rs200970033 Chr.9: 91723861-91723861 on Build GRCh38
rs755593346 Chr.9: 91724023-91724023 on Build GRCh38
rs56091954 Chr.9: 91723767-91723767 on Build GRCh38
rs767677536 Chr.9: 91723815-91723815 on Build GRCh38
rs1057029126 Chr.9: 91723747-91723747 on Build GRCh38
rs968394913 Chr.9: 91723699-91723699 on Build GRCh38
rs920956962 Chr.9: 91723696-91723696 on Build GRCh38
rs267602312 Chr.9: 91724117-91724117 on Build GRCh38
rs1002990573 Chr.9: 91723967-91723967 on Build GRCh38
rs750442188 Chr.9: 91723925-91723925 on Build GRCh38
rs752571643 Chr.9: 91724058-91724058 on Build GRCh38
rs202010959 Chr.9: 91724049-91724049 on Build GRCh38
rs10761129 Chr.9: 91724039-91724039 on Build GRCh38
rs750698649 Chr.9: 91723639-91723639 on Build GRCh38
rs373037095 Chr.9: 91723845-91723845 on Build GRCh38
rs367647446 Chr.9: 91723783-91723783 on Build GRCh38
rs577141662 Chr.9: 91723902-91723902 on Build GRCh38
rs41277835 Chr.9: 91723689-91723689 on Build GRCh38
rs780238625 Chr.9: 91724097-91724097 on Build GRCh38
rs983485063 Chr.9: 91724129-91724129 on Build GRCh38
rs375769750 Chr.9: 91723936-91723936 on Build GRCh38
rs771882038 Chr.9: 91723688-91723688 on Build GRCh38
rs930640242 Chr.9: 91724020-91724020 on Build GRCh38
rs747693128 Chr.9: 91723954-91723954 on Build GRCh38
rs767258825 Chr.9: 91724053-91724053 on Build GRCh38
rs902441691 Chr.9: 91723632-91723632 on Build GRCh38
rs756827949 Chr.9: 91724084-91724084 on Build GRCh38
rs180825668 Chr.9: 91723797-91723797 on Build GRCh38
rs777227251 Chr.9: 91724026-91724026 on Build GRCh38
rs755155776 Chr.9: 91723737-91723737 on Build GRCh38
rs1045374295 Chr.9: 91724076-91724076 on Build GRCh38
rs942184835 Chr.9: 91723577-91723577 on Build GRCh38
rs770921333 Chr.9: 91724050-91724050 on Build GRCh38
rs779461225 Chr.9: 91724091-91724091 on Build GRCh38
rs373575567 Chr.9: 91723977-91723977 on Build GRCh38
rs774317100 Chr.9: 91724116-91724116 on Build GRCh38
rs1044883600 Chr.9: 91723931-91723931 on Build GRCh38
rs759355134 Chr.9: 91724118-91724118 on Build GRCh38
rs1037636706 Chr.9: 91724120-91724120 on Build GRCh38

View Full Product Details