Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 197511710-197512308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BDH1
Entrez Gene ID
622
Gene Name
3-hydroxybutyrate dehydrogenase, type 1
Gene Aliases
BDH, SDR9C1
UniGene
Hs.274539
Gene Chromosome Location
Chr.3: 197509783-197573323 on Build GRCh38
Transcripts
NM_004051.4 NM_203314.2 NM_203315.2 XM_005269352.3 XM_005269355.4 XM_011513067.2 XM_017007007.1 XM_017007008.1 XM_017007009.1 XM_017007010.1 XM_017007011.1 XM_017007012.1 XM_017007014.1 XM_017007015.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4152395 c.1028G>A p.R343H Chr.3: 197511899-197511899 on Build GRCh38
4149809 c.918G>A p.L306L Chr.3: 197512009-197512009 on Build GRCh38
6164166 c.641G>T p.G214V Chr.3: 197512286-197512286 on Build GRCh38
1566837 c.620C>T p.P207L Chr.3: 197512307-197512307 on Build GRCh38
209434 c.922G>A p.A308T Chr.3: 197512005-197512005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768975972 Chr.3: 197512245-197512245 on Build GRCh38
rs928975303 Chr.3: 197512209-197512209 on Build GRCh38
rs747305588 Chr.3: 197511866-197511866 on Build GRCh38
rs144840608 Chr.3: 197511986-197511986 on Build GRCh38
rs887386685 Chr.3: 197512215-197512215 on Build GRCh38
rs1013777990 Chr.3: 197511986-197511986 on Build GRCh38
rs747867608 Chr.3: 197512251-197512251 on Build GRCh38
rs748690102 Chr.3: 197511908-197511908 on Build GRCh38
rs746397441 Chr.3: 197512232-197512232 on Build GRCh38
rs374112979 Chr.3: 197511852-197511852 on Build GRCh38
rs58455529 Chr.3: 197511898-197511898 on Build GRCh38
rs987355205 Chr.3: 197512141-197512141 on Build GRCh38
rs370655515 Chr.3: 197512137-197512137 on Build GRCh38
rs767131775 Chr.3: 197512271-197512271 on Build GRCh38
rs766590693 Chr.3: 197511963-197511963 on Build GRCh38
rs759881932 Chr.3: 197511965-197511965 on Build GRCh38
rs925917663 Chr.3: 197511770-197511770 on Build GRCh38
rs780594721 Chr.3: 197511750-197511750 on Build GRCh38
rs200520797 Chr.3: 197512014-197512014 on Build GRCh38
rs969776697 Chr.3: 197511776-197511776 on Build GRCh38
rs190572089 Chr.3: 197511858-197511858 on Build GRCh38
rs762920612 Chr.3: 197512262-197512262 on Build GRCh38
rs758109338 Chr.3: 197512023-197512023 on Build GRCh38
rs749858327 Chr.3: 197512039-197512039 on Build GRCh38
rs926042642 Chr.3: 197512061-197512061 on Build GRCh38
rs757649832 Chr.3: 197512277-197512277 on Build GRCh38
rs750046707 Chr.3: 197512015-197512015 on Build GRCh38
rs934635590 Chr.3: 197511795-197511795 on Build GRCh38
rs774640647 Chr.3: 197512042-197512042 on Build GRCh38
rs140457761 Chr.3: 197512200-197512200 on Build GRCh38
rs925185989 Chr.3: 197511798-197511798 on Build GRCh38
rs745877266 Chr.3: 197512155-197512155 on Build GRCh38
rs778442247 Chr.3: 197512013-197512013 on Build GRCh38
rs1037863518 Chr.3: 197512174-197512174 on Build GRCh38
rs757702931 Chr.3: 197512224-197512224 on Build GRCh38
rs775787132 Chr.3: 197512055-197512055 on Build GRCh38
rs755632402 Chr.3: 197511849-197511849 on Build GRCh38
rs747438713 Chr.3: 197511987-197511992 on Build GRCh38
rs61745694 Chr.3: 197512183-197512183 on Build GRCh38
rs140846840 Chr.3: 197512153-197512153 on Build GRCh38
rs780257378 Chr.3: 197511984-197511985 on Build GRCh38
rs748674418 Chr.3: 197512241-197512241 on Build GRCh38
rs762275510 Chr.3: 197511880-197511880 on Build GRCh38
rs772311219 Chr.3: 197512159-197512159 on Build GRCh38
rs779524014 Chr.3: 197512225-197512225 on Build GRCh38
rs773228186 Chr.3: 197511886-197511886 on Build GRCh38
rs559084253 Chr.3: 197511964-197511964 on Build GRCh38
rs377591892 Chr.3: 197511877-197511877 on Build GRCh38
rs114600793 Chr.3: 197512284-197512284 on Build GRCh38
rs751249430 Chr.3: 197512121-197512121 on Build GRCh38
rs144830970 Chr.3: 197512286-197512286 on Build GRCh38
rs765097046 Chr.3: 197512197-197512197 on Build GRCh38
rs771624244 Chr.3: 197512136-197512136 on Build GRCh38
rs748680735 Chr.3: 197511926-197511926 on Build GRCh38
rs762905070 Chr.3: 197511955-197511955 on Build GRCh38
rs747191828 Chr.3: 197512237-197512237 on Build GRCh38
rs367827020 Chr.3: 197512122-197512122 on Build GRCh38
rs535608035 Chr.3: 197512069-197512069 on Build GRCh38
rs767417205 Chr.3: 197511970-197511970 on Build GRCh38
rs777270241 Chr.3: 197511963-197511968 on Build GRCh38
rs374291198 Chr.3: 197511916-197511916 on Build GRCh38
rs374296813 Chr.3: 197511906-197511906 on Build GRCh38
rs891773702 Chr.3: 197511734-197511734 on Build GRCh38
rs761005595 Chr.3: 197512163-197512163 on Build GRCh38
rs372458828 Chr.3: 197511732-197511732 on Build GRCh38
rs778095243 Chr.3: 197512038-197512038 on Build GRCh38
rs149945578 Chr.3: 197512216-197512216 on Build GRCh38
rs1044673041 Chr.3: 197512142-197512142 on Build GRCh38
rs752650625 Chr.3: 197511900-197511900 on Build GRCh38
rs776728556 Chr.3: 197512249-197512249 on Build GRCh38
rs1000703569 Chr.3: 197512243-197512243 on Build GRCh38
rs551796884 Chr.3: 197511901-197511901 on Build GRCh38
rs769038648 Chr.3: 197511869-197511869 on Build GRCh38
rs773113109 Chr.3: 197512260-197512260 on Build GRCh38
rs202110030 Chr.3: 197512179-197512179 on Build GRCh38
rs752919802 Chr.3: 197512230-197512230 on Build GRCh38
rs1038980493 Chr.3: 197512161-197512161 on Build GRCh38
rs753843859 Chr.3: 197511991-197511991 on Build GRCh38
rs368212467 Chr.3: 197512212-197512212 on Build GRCh38
rs773135340 Chr.3: 197512198-197512198 on Build GRCh38
rs756705857 Chr.3: 197512005-197512005 on Build GRCh38
rs559038515 Chr.3: 197512273-197512273 on Build GRCh38
rs201488968 Chr.3: 197511940-197511940 on Build GRCh38
rs774007990 Chr.3: 197511863-197511863 on Build GRCh38
rs754774070 Chr.3: 197511899-197511899 on Build GRCh38
rs772364255 Chr.3: 197512052-197512052 on Build GRCh38
rs890810529 Chr.3: 197512214-197512214 on Build GRCh38
rs145598252 Chr.3: 197512083-197512083 on Build GRCh38
rs766647802 Chr.3: 197511891-197511891 on Build GRCh38
rs371013833 Chr.3: 197511892-197511892 on Build GRCh38
rs753427741 Chr.3: 197511965-197511966 on Build GRCh38
rs183131131 Chr.3: 197512126-197512126 on Build GRCh38
rs763872527 Chr.3: 197511985-197511985 on Build GRCh38
rs201020356 Chr.3: 197511874-197511874 on Build GRCh38
rs145104101 Chr.3: 197512267-197512267 on Build GRCh38
rs779504270 Chr.3: 197512109-197512109 on Build GRCh38
rs936632262 Chr.3: 197511833-197511833 on Build GRCh38
rs140040785 Chr.3: 197512079-197512079 on Build GRCh38
rs887251105 Chr.3: 197511981-197511981 on Build GRCh38
rs752493424 Chr.3: 197511975-197511975 on Build GRCh38
rs116643903 Chr.3: 197512030-197512030 on Build GRCh38
rs71325611 Chr.3: 197512004-197512004 on Build GRCh38
rs769171161 Chr.3: 197511994-197512000 on Build GRCh38
rs993789624 Chr.3: 197511910-197511910 on Build GRCh38
rs770259888 Chr.3: 197511930-197511930 on Build GRCh38
rs371927738 Chr.3: 197512270-197512270 on Build GRCh38
rs544714134 Chr.3: 197511783-197511783 on Build GRCh38
rs562942311 Chr.3: 197511808-197511808 on Build GRCh38
rs147271608 Chr.3: 197512084-197512084 on Build GRCh38
rs771107212 Chr.3: 197512040-197512040 on Build GRCh38
rs759819465 Chr.3: 197512043-197512043 on Build GRCh38
rs186563205 Chr.3: 197511857-197511857 on Build GRCh38
rs754729827 Chr.3: 197512213-197512213 on Build GRCh38
rs760533836 Chr.3: 197511983-197511983 on Build GRCh38
rs201111186 Chr.3: 197511909-197511909 on Build GRCh38
rs913842521 Chr.3: 197512111-197512111 on Build GRCh38
rs763221427 Chr.3: 197511887-197511887 on Build GRCh38
rs745542224 Chr.3: 197511914-197511914 on Build GRCh38
rs760346357 Chr.3: 197512060-197512060 on Build GRCh38
rs765743905 Chr.3: 197512278-197512278 on Build GRCh38
rs545192310 Chr.3: 197512158-197512158 on Build GRCh38
rs758384765 Chr.3: 197512236-197512236 on Build GRCh38
rs769605817 Chr.3: 197512258-197512258 on Build GRCh38
rs1002260866 Chr.3: 197512242-197512242 on Build GRCh38
rs781519040 Chr.3: 197512029-197512029 on Build GRCh38
rs980826419 Chr.3: 197511784-197511784 on Build GRCh38
rs765324949 Chr.3: 197511885-197511885 on Build GRCh38
rs768985550 Chr.3: 197511915-197511915 on Build GRCh38
rs144603437 Chr.3: 197512171-197512171 on Build GRCh38
rs371408136 Chr.3: 197511912-197511912 on Build GRCh38
rs770197020 Chr.3: 197512035-197512035 on Build GRCh38
rs781744588 Chr.3: 197512135-197512136 on Build GRCh38
rs956149169 Chr.3: 197512066-197512066 on Build GRCh38
rs374039230 Chr.3: 197512182-197512182 on Build GRCh38
rs756991726 Chr.3: 197511975-197511982 on Build GRCh38

View Full Product Details