Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 165085439-165085787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM192
Entrez Gene ID
201931
Gene Name
transmembrane protein 192
Gene Aliases
-
UniGene
Hs.704511
Gene Chromosome Location
Chr.4: 165076078-165112913 on Build GRCh38
Transcripts
NM_001100389.1 XM_011531717.2 XM_011531718.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6803700 c.575-2A>G p.? Chr.4: 165085690-165085690 on Build GRCh38
5400763 c.671G>A p.G224E Chr.4: 165085592-165085592 on Build GRCh38
6803703 c.622A>C p.I208L Chr.4: 165085629-165085629 on Build GRCh38
4860761 c.572G>T p.R191L Chr.4: 165085679-165085679 on Build GRCh38
6803702 c.634A>C p.I212L Chr.4: 165085629-165085629 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769335031 Chr.4: 165085730-165085731 on Build GRCh38
rs757191287 Chr.4: 165085549-165085549 on Build GRCh38
rs141009367 Chr.4: 165085681-165085681 on Build GRCh38
rs755451074 Chr.4: 165085485-165085485 on Build GRCh38
rs202163114 Chr.4: 165085678-165085678 on Build GRCh38
rs946134335 Chr.4: 165085639-165085639 on Build GRCh38
rs1028992994 Chr.4: 165085515-165085515 on Build GRCh38
rs755661047 Chr.4: 165085668-165085668 on Build GRCh38
rs766384845 Chr.4: 165085716-165085716 on Build GRCh38
rs778880630 Chr.4: 165085553-165085553 on Build GRCh38
rs949722018 Chr.4: 165085699-165085700 on Build GRCh38
rs758225347 Chr.4: 165085571-165085571 on Build GRCh38
rs760806551 Chr.4: 165085620-165085620 on Build GRCh38
rs954899282 Chr.4: 165085545-165085545 on Build GRCh38
rs746689799 Chr.4: 165085575-165085575 on Build GRCh38
rs780894503 Chr.4: 165085581-165085581 on Build GRCh38
rs553668773 Chr.4: 165085679-165085679 on Build GRCh38
rs372244546 Chr.4: 165085591-165085591 on Build GRCh38
rs376642145 Chr.4: 165085642-165085642 on Build GRCh38
rs766299227 Chr.4: 165085637-165085637 on Build GRCh38
rs750377543 Chr.4: 165085626-165085626 on Build GRCh38
rs531412617 Chr.4: 165085703-165085703 on Build GRCh38
rs556161304 Chr.4: 165085574-165085574 on Build GRCh38
rs779676925 Chr.4: 165085548-165085549 on Build GRCh38
rs773752707 Chr.4: 165085592-165085592 on Build GRCh38
rs181591398 Chr.4: 165085600-165085600 on Build GRCh38
rs747668000 Chr.4: 165085663-165085663 on Build GRCh38
rs752729383 Chr.4: 165085547-165085547 on Build GRCh38
rs758226941 Chr.4: 165085628-165085628 on Build GRCh38
rs370167147 Chr.4: 165085560-165085560 on Build GRCh38
rs923692447 Chr.4: 165085734-165085734 on Build GRCh38
rs377575905 Chr.4: 165085698-165085698 on Build GRCh38
rs748524387 Chr.4: 165085644-165085644 on Build GRCh38
rs747653581 Chr.4: 165085633-165085635 on Build GRCh38
rs373469548 Chr.4: 165085715-165085715 on Build GRCh38
rs768374183 Chr.4: 165085579-165085579 on Build GRCh38
rs537452762 Chr.4: 165085526-165085526 on Build GRCh38
rs767705040 Chr.4: 165085610-165085610 on Build GRCh38
rs780984142 Chr.4: 165085662-165085662 on Build GRCh38
rs543559109 Chr.4: 165085666-165085666 on Build GRCh38
rs780881872 Chr.4: 165085628-165085629 on Build GRCh38
rs754714775 Chr.4: 165085646-165085646 on Build GRCh38
rs774661901 Chr.4: 165085608-165085608 on Build GRCh38
rs771333597 Chr.4: 165085601-165085601 on Build GRCh38
rs752815225 Chr.4: 165085617-165085617 on Build GRCh38
rs765146576 Chr.4: 165085625-165085625 on Build GRCh38
rs767619266 Chr.4: 165085535-165085535 on Build GRCh38
rs747625875 Chr.4: 165085590-165085590 on Build GRCh38
rs566875676 Chr.4: 165085521-165085521 on Build GRCh38
rs373979273 Chr.4: 165085680-165085680 on Build GRCh38
rs754230524 Chr.4: 165085752-165085752 on Build GRCh38
rs746828895 Chr.4: 165085577-165085578 on Build GRCh38
rs886819980 Chr.4: 165085714-165085714 on Build GRCh38
rs749836115 Chr.4: 165085676-165085676 on Build GRCh38
rs758181402 Chr.4: 165085760-165085760 on Build GRCh38
rs760740136 Chr.4: 165085692-165085692 on Build GRCh38
rs754532328 Chr.4: 165085587-165085587 on Build GRCh38
rs369175508 Chr.4: 165085701-165085701 on Build GRCh38
rs1005266891 Chr.4: 165085733-165085733 on Build GRCh38
rs1021949606 Chr.4: 165085699-165085699 on Build GRCh38
rs1034920218 Chr.4: 165085623-165085623 on Build GRCh38
rs751463008 Chr.4: 165085723-165085723 on Build GRCh38
rs777084713 Chr.4: 165085672-165085672 on Build GRCh38
rs775677918 Chr.4: 165085684-165085684 on Build GRCh38

View Full Product Details