Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 76054639-76055216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MDH2
Entrez Gene ID
4191
Gene Name
malate dehydrogenase 2
Gene Aliases
M-MDH, MDH, MGC:3559, MOR1
UniGene
Hs.520967
Gene Chromosome Location
Chr.7: 76048019-76067508 on Build GRCh38
Transcripts
NM_001282403.1 NM_001282404.1 NM_005918.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3028704 c.109G>A p.G37R Chr.7: 76054872-76054872 on Build GRCh38
7124628 c.204G>C p.L68L Chr.7: 76054967-76054967 on Build GRCh38
3942302 c.162C>A p.T54T Chr.7: 76054925-76054925 on Build GRCh38
6027408 c.226G>A p.A76T Chr.7: 76054989-76054989 on Build GRCh38
1452097 c.169G>C p.D57H Chr.7: 76054932-76054932 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1007364275 Chr.7: 76054947-76054947 on Build GRCh38
rs782545373 Chr.7: 76054818-76054818 on Build GRCh38
rs938398963 Chr.7: 76054671-76054671 on Build GRCh38
rs112250025 Chr.7: 76055131-76055131 on Build GRCh38
rs781789831 Chr.7: 76055010-76055010 on Build GRCh38
rs144321556 Chr.7: 76054918-76054918 on Build GRCh38
rs115117282 Chr.7: 76054901-76054901 on Build GRCh38
rs993051319 Chr.7: 76054706-76054706 on Build GRCh38
rs548813592 Chr.7: 76054943-76054943 on Build GRCh38
rs141539461 Chr.7: 76054959-76054959 on Build GRCh38
rs79663210 Chr.7: 76055008-76055008 on Build GRCh38
rs77424348 Chr.7: 76054955-76054955 on Build GRCh38
rs782717623 Chr.7: 76054937-76054937 on Build GRCh38
rs548970837 Chr.7: 76055133-76055133 on Build GRCh38
rs1016063728 Chr.7: 76055159-76055159 on Build GRCh38
rs545790335 Chr.7: 76055176-76055176 on Build GRCh38
rs1043717766 Chr.7: 76054771-76054771 on Build GRCh38
rs111633186 Chr.7: 76054809-76054809 on Build GRCh38
rs200392965 Chr.7: 76055009-76055009 on Build GRCh38
rs901290701 Chr.7: 76054917-76054917 on Build GRCh38
rs371066346 Chr.7: 76054958-76054958 on Build GRCh38
rs139725372 Chr.7: 76054878-76054878 on Build GRCh38
rs782462370 Chr.7: 76055022-76055022 on Build GRCh38
rs782308462 Chr.7: 76054872-76054872 on Build GRCh38
rs769743658 Chr.7: 76054985-76054985 on Build GRCh38
rs782031941 Chr.7: 76054704-76054704 on Build GRCh38
rs1637030 Chr.7: 76055028-76055028 on Build GRCh38
rs545666167 Chr.7: 76054732-76054732 on Build GRCh38
rs949027943 Chr.7: 76054769-76054769 on Build GRCh38
rs781799479 Chr.7: 76054848-76054848 on Build GRCh38
rs782813525 Chr.7: 76054911-76054911 on Build GRCh38
rs782532690 Chr.7: 76055084-76055084 on Build GRCh38
rs375592605 Chr.7: 76054989-76054989 on Build GRCh38
rs574576746 Chr.7: 76055054-76055054 on Build GRCh38
rs934893475 Chr.7: 76055050-76055050 on Build GRCh38
rs781880905 Chr.7: 76054942-76054942 on Build GRCh38
rs782688703 Chr.7: 76054824-76054824 on Build GRCh38
rs782756337 Chr.7: 76055004-76055004 on Build GRCh38
rs781932433 Chr.7: 76054874-76054874 on Build GRCh38
rs372524677 Chr.7: 76054791-76054791 on Build GRCh38
rs569165542 Chr.7: 76055158-76055158 on Build GRCh38
rs782057861 Chr.7: 76054931-76054931 on Build GRCh38
rs934117306 Chr.7: 76055100-76055100 on Build GRCh38
rs782339624 Chr.7: 76054954-76054954 on Build GRCh38
rs201036956 Chr.7: 76055007-76055007 on Build GRCh38
rs201367969 Chr.7: 76054851-76054851 on Build GRCh38
rs782610233 Chr.7: 76055179-76055179 on Build GRCh38
rs782728879 Chr.7: 76055025-76055025 on Build GRCh38
rs782759642 Chr.7: 76054918-76054919 on Build GRCh38
rs115021429 Chr.7: 76055059-76055059 on Build GRCh38
rs575050280 Chr.7: 76055144-76055144 on Build GRCh38
rs563278014 Chr.7: 76054705-76054705 on Build GRCh38
rs781834910 Chr.7: 76054938-76054938 on Build GRCh38
rs192888757 Chr.7: 76054799-76054799 on Build GRCh38
rs528050550 Chr.7: 76054803-76054803 on Build GRCh38
rs782724462 Chr.7: 76054784-76054784 on Build GRCh38
rs148797426 Chr.7: 76054940-76054940 on Build GRCh38
rs781942767 Chr.7: 76054988-76054988 on Build GRCh38
rs782656418 Chr.7: 76054860-76054860 on Build GRCh38
rs782167273 Chr.7: 76054977-76054977 on Build GRCh38
rs782236110 Chr.7: 76054853-76054853 on Build GRCh38
rs199565893 Chr.7: 76054859-76054859 on Build GRCh38
rs781824482 Chr.7: 76054787-76054787 on Build GRCh38
rs782507132 Chr.7: 76054939-76054939 on Build GRCh38
rs115007081 Chr.7: 76054707-76054707 on Build GRCh38
rs55790234 Chr.7: 76055043-76055043 on Build GRCh38
rs782294941 Chr.7: 76054871-76054871 on Build GRCh38
rs782356292 Chr.7: 76054890-76054890 on Build GRCh38
rs782461596 Chr.7: 76054850-76054850 on Build GRCh38
rs901154862 Chr.7: 76055092-76055092 on Build GRCh38
rs763046893 Chr.7: 76055166-76055166 on Build GRCh38
rs537742207 Chr.7: 76054974-76054974 on Build GRCh38
rs782508137 Chr.7: 76055046-76055046 on Build GRCh38
rs146206957 Chr.7: 76054834-76054834 on Build GRCh38
rs187496518 Chr.7: 76055116-76055116 on Build GRCh38
rs563930090 Chr.7: 76054740-76054740 on Build GRCh38
rs560653489 Chr.7: 76054808-76054808 on Build GRCh38

View Full Product Details