Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 19217597-19218155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMC1
Entrez Gene ID
23065
Gene Name
ER membrane protein complex subunit 1
Gene Aliases
CAVIPMR, KIAA0090
UniGene
Hs.439200
Gene Chromosome Location
Chr.1: 19215664-19251559 on Build GRCh38
Transcripts
NM_001271427.1 NM_001271428.1 NM_001271429.1 NM_015047.2 XM_005245787.1 XM_005245788.1 NR_135114.1 NR_135115.1

Gene Symbol
LOC101927895
Entrez Gene ID
101927895
Gene Name
uncharacterized LOC101927895
Gene Aliases
-
UniGene
Hs.660534
Gene Chromosome Location
Chr.1: 19210386-19240704 on Build GRCh38
Transcripts
NM_001271427.1 NM_001271428.1 NM_001271429.1 NM_015047.2 XM_005245787.1 XM_005245788.1 NR_135114.1 NR_135115.1

SNP Detail

SNP ID Chromosome Location
rs1003182304 Chr.1: 19217762-19217762 on Build GRCh38
rs551009087 Chr.1: 19217714-19217714 on Build GRCh38
rs138442691 Chr.1: 19217958-19217958 on Build GRCh38
rs893660433 Chr.1: 19217960-19217960 on Build GRCh38
rs571521086 Chr.1: 19217679-19217679 on Build GRCh38
rs996487219 Chr.1: 19217920-19217920 on Build GRCh38
rs944915767 Chr.1: 19217880-19217880 on Build GRCh38
rs950893083 Chr.1: 19217819-19217819 on Build GRCh38
rs1039023548 Chr.1: 19217628-19217628 on Build GRCh38
rs145816805 Chr.1: 19217829-19217829 on Build GRCh38
rs368929668 Chr.1: 19217810-19217810 on Build GRCh38
rs1055782531 Chr.1: 19217769-19217769 on Build GRCh38
rs915950528 Chr.1: 19218118-19218118 on Build GRCh38
rs995017298 Chr.1: 19218076-19218076 on Build GRCh38
rs558839413 Chr.1: 19217901-19217901 on Build GRCh38
rs555756545 Chr.1: 19217764-19217764 on Build GRCh38
rs946205498 Chr.1: 19217702-19217702 on Build GRCh38
rs532265090 Chr.1: 19217705-19217705 on Build GRCh38
rs748153982 Chr.1: 19218092-19218092 on Build GRCh38
rs928888474 Chr.1: 19218058-19218058 on Build GRCh38
rs534402478 Chr.1: 19217883-19217883 on Build GRCh38
rs936124034 Chr.1: 19217919-19217919 on Build GRCh38
rs984136599 Chr.1: 19217825-19217825 on Build GRCh38
rs574131226 Chr.1: 19217893-19217893 on Build GRCh38
rs569244408 Chr.1: 19217755-19217755 on Build GRCh38
rs1003984771 Chr.1: 19217855-19217855 on Build GRCh38
rs1009922456 Chr.1: 19217713-19217713 on Build GRCh38
rs957934394 Chr.1: 19217971-19217971 on Build GRCh38
rs1042181154 Chr.1: 19218007-19218007 on Build GRCh38
rs372230447 Chr.1: 19218026-19218026 on Build GRCh38
rs538123344 Chr.1: 19217979-19217979 on Build GRCh38
rs187946652 Chr.1: 19217635-19217635 on Build GRCh38
rs914780873 Chr.1: 19217684-19217684 on Build GRCh38
rs1048951616 Chr.1: 19217813-19217813 on Build GRCh38
rs764884421 Chr.1: 19217821-19217821 on Build GRCh38
rs752494685 Chr.1: 19217839-19217839 on Build GRCh38
rs938873189 Chr.1: 19218094-19218094 on Build GRCh38
rs894551759 Chr.1: 19217794-19217794 on Build GRCh38
rs760987134 Chr.1: 19217767-19217767 on Build GRCh38
rs977556222 Chr.1: 19218031-19218031 on Build GRCh38
rs751050625 Chr.1: 19218104-19218104 on Build GRCh38
rs913622180 Chr.1: 19218012-19218012 on Build GRCh38
rs936060979 Chr.1: 19217664-19217664 on Build GRCh38
rs764077248 Chr.1: 19217989-19217989 on Build GRCh38
rs1042017068 Chr.1: 19217723-19217723 on Build GRCh38
rs546624325 Chr.1: 19217675-19217675 on Build GRCh38
rs567113408 Chr.1: 19217853-19217853 on Build GRCh38
rs931305956 Chr.1: 19218119-19218119 on Build GRCh38
rs894806408 Chr.1: 19218065-19218065 on Build GRCh38
rs549423968 Chr.1: 19217663-19217663 on Build GRCh38
rs1048499233 Chr.1: 19218122-19218122 on Build GRCh38
rs1027950614 Chr.1: 19217934-19217934 on Build GRCh38
rs1033342625 Chr.1: 19218061-19218061 on Build GRCh38
rs945243959 Chr.1: 19218004-19218004 on Build GRCh38

View Full Product Details