Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 42160044-42160630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAK1
Entrez Gene ID
22906
Gene Name
trafficking kinesin protein 1
Gene Aliases
MILT1, OIP106
UniGene
Hs.535711
Gene Chromosome Location
Chr.3: 42013802-42225890 on Build GRCh38
Transcripts
NM_001042646.2 NM_001265608.1 NM_001265609.1 NM_001265610.1 NM_014965.4 XM_005264956.2 XM_005264958.2 XM_005264960.3 XM_005264962.3 XM_005264963.3 XM_006713029.2 XM_006713030.2 XM_006713031.3 XM_011533489.2 XM_017005906.1 XM_017005907.1 XM_017005908.1 XM_017005909.1 XM_017005910.1 XM_017005911.1 XM_017005912.1 XM_017005913.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6511028 c.28G>A p.E10K Chr.3: 42160389-42160389 on Build GRCh38
1186245 c.58G>C p.E20Q Chr.3: 42160419-42160419 on Build GRCh38
5486354 c.76C>A p.H26N Chr.3: 42160437-42160437 on Build GRCh38
4584344 c.21C>T p.G7G Chr.3: 42160382-42160382 on Build GRCh38
1044646 c.45C>T p.Y15Y Chr.3: 42160406-42160406 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751720606 Chr.3: 42160490-42160490 on Build GRCh38
rs770459652 Chr.3: 42160446-42160446 on Build GRCh38
rs763082922 Chr.3: 42160476-42160476 on Build GRCh38
rs557249462 Chr.3: 42160168-42160168 on Build GRCh38
rs753003476 Chr.3: 42160516-42160516 on Build GRCh38
rs953704998 Chr.3: 42160152-42160152 on Build GRCh38
rs9311304 Chr.3: 42160563-42160563 on Build GRCh38
rs565141736 Chr.3: 42160558-42160558 on Build GRCh38
rs552657030 Chr.3: 42160233-42160233 on Build GRCh38
rs1044979175 Chr.3: 42160221-42160221 on Build GRCh38
rs752622155 Chr.3: 42160208-42160208 on Build GRCh38
rs770220793 Chr.3: 42160439-42160439 on Build GRCh38
rs973475638 Chr.3: 42160135-42160135 on Build GRCh38
rs764062131 Chr.3: 42160471-42160471 on Build GRCh38
rs1048852561 Chr.3: 42160505-42160505 on Build GRCh38
rs760750844 Chr.3: 42160372-42160372 on Build GRCh38
rs935045913 Chr.3: 42160196-42160196 on Build GRCh38
rs759598405 Chr.3: 42160077-42160077 on Build GRCh38
rs552774158 Chr.3: 42160076-42160076 on Build GRCh38
rs770049079 Chr.3: 42160112-42160112 on Build GRCh38
rs776729033 Chr.3: 42160474-42160474 on Build GRCh38
rs554729216 Chr.3: 42160297-42160297 on Build GRCh38
rs748748734 Chr.3: 42160444-42160444 on Build GRCh38
rs565160282 Chr.3: 42160301-42160301 on Build GRCh38
rs780086441 Chr.3: 42160401-42160401 on Build GRCh38
rs150467214 Chr.3: 42160360-42160360 on Build GRCh38
rs537622490 Chr.3: 42160153-42160153 on Build GRCh38
rs114159123 Chr.3: 42160528-42160528 on Build GRCh38
rs749833909 Chr.3: 42160522-42160522 on Build GRCh38
rs760548430 Chr.3: 42160464-42160464 on Build GRCh38
rs763263485 Chr.3: 42160174-42160174 on Build GRCh38
rs193111040 Chr.3: 42160586-42160586 on Build GRCh38
rs150131443 Chr.3: 42160527-42160527 on Build GRCh38
rs367980895 Chr.3: 42160336-42160336 on Build GRCh38
rs9757367 Chr.3: 42160117-42160117 on Build GRCh38
rs796248612 Chr.3: 42160563-42160569 on Build GRCh38
rs765400946 Chr.3: 42160377-42160377 on Build GRCh38
rs780325053 Chr.3: 42160325-42160325 on Build GRCh38
rs745582280 Chr.3: 42160451-42160451 on Build GRCh38
rs373959536 Chr.3: 42160563-42160563 on Build GRCh38
rs1009190130 Chr.3: 42160181-42160181 on Build GRCh38
rs959581885 Chr.3: 42160565-42160569 on Build GRCh38
rs914932302 Chr.3: 42160158-42160158 on Build GRCh38
rs762456893 Chr.3: 42160349-42160349 on Build GRCh38
rs201440342 Chr.3: 42160569-42160569 on Build GRCh38
rs374936361 Chr.3: 42160354-42160354 on Build GRCh38
rs373479760 Chr.3: 42160379-42160379 on Build GRCh38
rs535940981 Chr.3: 42160229-42160229 on Build GRCh38
rs867889316 Chr.3: 42160550-42160550 on Build GRCh38
rs374035147 Chr.3: 42160268-42160268 on Build GRCh38
rs755297086 Chr.3: 42160502-42160502 on Build GRCh38
rs1027792308 Chr.3: 42160138-42160138 on Build GRCh38
rs952775848 Chr.3: 42160248-42160248 on Build GRCh38
rs945041464 Chr.3: 42160165-42160165 on Build GRCh38
rs989241935 Chr.3: 42160364-42160364 on Build GRCh38
rs778213183 Chr.3: 42160518-42160518 on Build GRCh38
rs372670418 Chr.3: 42160479-42160479 on Build GRCh38
rs747275593 Chr.3: 42160329-42160329 on Build GRCh38
rs986384327 Chr.3: 42160159-42160159 on Build GRCh38
rs538755047 Chr.3: 42160589-42160589 on Build GRCh38
rs751852677 Chr.3: 42160407-42160407 on Build GRCh38
rs752710776 Chr.3: 42160312-42160312 on Build GRCh38
rs756698635 Chr.3: 42160525-42160525 on Build GRCh38
rs966489542 Chr.3: 42160570-42160570 on Build GRCh38
rs59260540 Chr.3: 42160569-42160570 on Build GRCh38
rs971009642 Chr.3: 42160282-42160282 on Build GRCh38
rs755279519 Chr.3: 42160420-42160420 on Build GRCh38
rs372685656 Chr.3: 42160202-42160202 on Build GRCh38
rs962062149 Chr.3: 42160131-42160131 on Build GRCh38
rs923605424 Chr.3: 42160194-42160194 on Build GRCh38
rs750658959 Chr.3: 42160319-42160319 on Build GRCh38
rs1036263399 Chr.3: 42160472-42160472 on Build GRCh38
rs766534590 Chr.3: 42160485-42160485 on Build GRCh38
rs567475891 Chr.3: 42160568-42160568 on Build GRCh38
rs141039597 Chr.3: 42160247-42160247 on Build GRCh38
rs781523594 Chr.3: 42160337-42160337 on Build GRCh38
rs781665054 Chr.3: 42160421-42160421 on Build GRCh38
rs17069599 Chr.3: 42160508-42160508 on Build GRCh38
rs11370773 Chr.3: 42160558-42160559 on Build GRCh38
rs371720812 Chr.3: 42160342-42160342 on Build GRCh38
rs772778595 Chr.3: 42160348-42160348 on Build GRCh38
rs369997765 Chr.3: 42160371-42160371 on Build GRCh38
rs748652717 Chr.3: 42160430-42160430 on Build GRCh38
rs943679446 Chr.3: 42160169-42160169 on Build GRCh38
rs138273292 Chr.3: 42160448-42160448 on Build GRCh38
rs1054215719 Chr.3: 42160101-42160117 on Build GRCh38
rs771749833 Chr.3: 42160459-42160459 on Build GRCh38
rs71616033 Chr.3: 42160578-42160579 on Build GRCh38
rs947920362 Chr.3: 42160214-42160214 on Build GRCh38
rs552616319 Chr.3: 42160578-42160578 on Build GRCh38
rs775420950 Chr.3: 42160462-42160462 on Build GRCh38
rs4974003 Chr.3: 42160569-42160569 on Build GRCh38
rs764380001 Chr.3: 42160176-42160176 on Build GRCh38
rs374331015 Chr.3: 42160318-42160318 on Build GRCh38
rs959508224 Chr.3: 42160299-42160299 on Build GRCh38
rs9755034 Chr.3: 42160267-42160267 on Build GRCh38
rs1014852198 Chr.3: 42160249-42160249 on Build GRCh38
rs549537713 Chr.3: 42160073-42160073 on Build GRCh38
rs117757392 Chr.3: 42160442-42160442 on Build GRCh38
rs1019100817 Chr.3: 42160535-42160535 on Build GRCh38
rs748742164 Chr.3: 42160343-42160343 on Build GRCh38
rs774076455 Chr.3: 42160359-42160359 on Build GRCh38
rs374783274 Chr.3: 42160116-42160116 on Build GRCh38
rs575427886 Chr.3: 42160309-42160309 on Build GRCh38
rs796196526 Chr.3: 42160295-42160295 on Build GRCh38
rs930497636 Chr.3: 42160477-42160477 on Build GRCh38
rs544527801 Chr.3: 42160314-42160314 on Build GRCh38
rs201670628 Chr.3: 42160382-42160382 on Build GRCh38
rs756493357 Chr.3: 42160517-42160517 on Build GRCh38
rs906530657 Chr.3: 42160109-42160109 on Build GRCh38
rs769283794 Chr.3: 42160345-42160345 on Build GRCh38
rs758757414 Chr.3: 42160321-42160321 on Build GRCh38
rs779804036 Chr.3: 42160441-42160442 on Build GRCh38

View Full Product Details