Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 55045730-55046232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOCS4
Entrez Gene ID
122809
Gene Name
suppressor of cytokine signaling 4
Gene Aliases
SOCS7
UniGene
Hs.744302
Gene Chromosome Location
Chr.14: 55027126-55049489 on Build GRCh38
Transcripts
NM_080867.2 NM_199421.1 XM_011536425.1 XM_011536426.1

SNP Detail

SNP ID Chromosome Location
rs866066397 Chr.14: 55046014-55046014 on Build GRCh38
rs1056021837 Chr.14: 55046001-55046001 on Build GRCh38
rs904066613 Chr.14: 55045826-55045826 on Build GRCh38
rs770874488 Chr.14: 55045884-55045884 on Build GRCh38
rs776575510 Chr.14: 55045777-55045777 on Build GRCh38
rs925094809 Chr.14: 55045761-55045761 on Build GRCh38
rs984713223 Chr.14: 55046030-55046030 on Build GRCh38
rs761909072 Chr.14: 55045880-55045880 on Build GRCh38
rs901975750 Chr.14: 55045776-55045776 on Build GRCh38
rs767294024 Chr.14: 55046163-55046163 on Build GRCh38
rs945469225 Chr.14: 55046203-55046203 on Build GRCh38
rs952031667 Chr.14: 55046022-55046023 on Build GRCh38
rs1051730010 Chr.14: 55045877-55045877 on Build GRCh38
rs574702610 Chr.14: 55046020-55046020 on Build GRCh38
rs879147602 Chr.14: 55046126-55046126 on Build GRCh38
rs552663785 Chr.14: 55046017-55046017 on Build GRCh38
rs896081619 Chr.14: 55046121-55046121 on Build GRCh38
rs774249331 Chr.14: 55045893-55045893 on Build GRCh38
rs754308333 Chr.14: 55045990-55045990 on Build GRCh38
rs752451122 Chr.14: 55046205-55046205 on Build GRCh38
rs368201830 Chr.14: 55046184-55046184 on Build GRCh38
rs971689211 Chr.14: 55045942-55045942 on Build GRCh38
rs78683368 Chr.14: 55045924-55045924 on Build GRCh38
rs190454735 Chr.14: 55046125-55046125 on Build GRCh38
rs541742955 Chr.14: 55046189-55046189 on Build GRCh38
rs879167904 Chr.14: 55046008-55046008 on Build GRCh38
rs371034830 Chr.14: 55045876-55045876 on Build GRCh38
rs534624423 Chr.14: 55045945-55045945 on Build GRCh38
rs146621006 Chr.14: 55046129-55046129 on Build GRCh38
rs1034237011 Chr.14: 55045877-55045879 on Build GRCh38
rs1042380025 Chr.14: 55045799-55045799 on Build GRCh38
rs1049328935 Chr.14: 55045760-55045761 on Build GRCh38
rs186128132 Chr.14: 55045757-55045757 on Build GRCh38
rs558685372 Chr.14: 55045910-55045910 on Build GRCh38
rs999674222 Chr.14: 55046178-55046178 on Build GRCh38
rs569459881 Chr.14: 55045875-55045875 on Build GRCh38
rs975063525 Chr.14: 55045778-55045778 on Build GRCh38
rs541740394 Chr.14: 55046094-55046094 on Build GRCh38
rs1037820494 Chr.14: 55045975-55045975 on Build GRCh38
rs921855126 Chr.14: 55045874-55045874 on Build GRCh38
rs1001711090 Chr.14: 55045992-55045992 on Build GRCh38
rs918931075 Chr.14: 55045986-55045986 on Build GRCh38
rs1024055589 Chr.14: 55045904-55045904 on Build GRCh38
rs943235220 Chr.14: 55045765-55045765 on Build GRCh38

View Full Product Details