Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 1518880-1519473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PITPNA
Entrez Gene ID
5306
Gene Name
phosphatidylinositol transfer protein alpha
Gene Aliases
HEL-S-36, PI-TPalpha, PITPN, VIB1A
UniGene
Hs.429819
Gene Chromosome Location
Chr.17: 1517989-1562816 on Build GRCh38
Transcripts
NM_006224.3 XM_011523939.1 XM_011523940.1

SNP Detail

SNP ID Chromosome Location
rs114948895 Chr.17: 1519047-1519047 on Build GRCh38
rs529265002 Chr.17: 1519193-1519193 on Build GRCh38
rs576005335 Chr.17: 1519223-1519223 on Build GRCh38
rs920884059 Chr.17: 1519125-1519125 on Build GRCh38
rs957975886 Chr.17: 1519256-1519256 on Build GRCh38
rs1049891531 Chr.17: 1519179-1519179 on Build GRCh38
rs1013569497 Chr.17: 1519034-1519034 on Build GRCh38
rs1008412421 Chr.17: 1519181-1519181 on Build GRCh38
rs1026910531 Chr.17: 1519370-1519370 on Build GRCh38
rs982193078 Chr.17: 1519049-1519049 on Build GRCh38
rs940828504 Chr.17: 1519064-1519064 on Build GRCh38
rs1002606783 Chr.17: 1519338-1519338 on Build GRCh38
rs910892649 Chr.17: 1519399-1519399 on Build GRCh38
rs943278689 Chr.17: 1518942-1518942 on Build GRCh38
rs566119517 Chr.17: 1519329-1519329 on Build GRCh38
rs757671870 Chr.17: 1519291-1519291 on Build GRCh38
rs545382277 Chr.17: 1519067-1519067 on Build GRCh38
rs1027985234 Chr.17: 1518902-1518902 on Build GRCh38
rs902358659 Chr.17: 1519222-1519222 on Build GRCh38
rs954834283 Chr.17: 1519390-1519390 on Build GRCh38
rs775111267 Chr.17: 1518953-1518953 on Build GRCh38
rs934482590 Chr.17: 1518989-1518989 on Build GRCh38
rs889565829 Chr.17: 1518906-1518906 on Build GRCh38
rs905180141 Chr.17: 1519038-1519038 on Build GRCh38
rs910236533 Chr.17: 1519146-1519146 on Build GRCh38
rs982610540 Chr.17: 1519315-1519315 on Build GRCh38
rs943683794 Chr.17: 1519401-1519401 on Build GRCh38
rs1050677150 Chr.17: 1519377-1519377 on Build GRCh38
rs878960740 Chr.17: 1519165-1519165 on Build GRCh38
rs1017359355 Chr.17: 1519412-1519412 on Build GRCh38
rs551801295 Chr.17: 1519429-1519429 on Build GRCh38
rs889441984 Chr.17: 1519180-1519180 on Build GRCh38
rs932227575 Chr.17: 1519132-1519132 on Build GRCh38
rs993706951 Chr.17: 1519359-1519359 on Build GRCh38
rs879505905 Chr.17: 1519411-1519411 on Build GRCh38
rs958029652 Chr.17: 1519032-1519032 on Build GRCh38
rs970722276 Chr.17: 1519312-1519312 on Build GRCh38
rs1801396 Chr.17: 1519356-1519356 on Build GRCh38
rs916570466 Chr.17: 1519046-1519046 on Build GRCh38
rs904365359 Chr.17: 1518988-1518991 on Build GRCh38
rs966510499 Chr.17: 1518961-1518961 on Build GRCh38
rs1053349347 Chr.17: 1519251-1519251 on Build GRCh38
rs892534628 Chr.17: 1519031-1519031 on Build GRCh38
rs964415214 Chr.17: 1519416-1519416 on Build GRCh38
rs1801635 Chr.17: 1519304-1519304 on Build GRCh38
rs541276303 Chr.17: 1519300-1519300 on Build GRCh38
rs765603411 Chr.17: 1519341-1519341 on Build GRCh38
rs917052153 Chr.17: 1519283-1519283 on Build GRCh38
rs116900293 Chr.17: 1519041-1519041 on Build GRCh38
rs925861906 Chr.17: 1519244-1519244 on Build GRCh38
rs1026085285 Chr.17: 1519107-1519107 on Build GRCh38
rs1032176584 Chr.17: 1519255-1519255 on Build GRCh38
rs1037995508 Chr.17: 1519345-1519345 on Build GRCh38
rs116565266 Chr.17: 1519249-1519249 on Build GRCh38
rs932260488 Chr.17: 1519362-1519362 on Build GRCh38
rs534920539 Chr.17: 1519001-1519001 on Build GRCh38
rs561284352 Chr.17: 1519078-1519078 on Build GRCh38
rs547807669 Chr.17: 1519297-1519297 on Build GRCh38
rs908047357 Chr.17: 1519062-1519062 on Build GRCh38
rs1024946527 Chr.17: 1519037-1519037 on Build GRCh38
rs142059189 Chr.17: 1519063-1519063 on Build GRCh38
rs1007965453 Chr.17: 1518931-1518931 on Build GRCh38
rs920749012 Chr.17: 1519360-1519360 on Build GRCh38
rs527462117 Chr.17: 1519250-1519250 on Build GRCh38
rs1032228989 Chr.17: 1519016-1519016 on Build GRCh38

View Full Product Details