Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 12293810-12294405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDN1
Entrez Gene ID
1906
Gene Name
endothelin 1
Gene Aliases
ARCND3, ET1, HDLCQ7, PPET1, QME
UniGene
Hs.511899
Gene Chromosome Location
Chr.6: 12256464-12297194 on Build GRCh38
Transcripts
NM_001168319.1 NM_001955.4 XM_011514330.2 XM_011514331.2 XM_011514332.2 XM_017010331.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3828955 c.446C>A p.S149Y Chr.6: 12294317-12294317 on Build GRCh38
5869221 c.253C>T p.L85F Chr.6: 12293960-12293960 on Build GRCh38
3620077 c.346G>A p.D116N Chr.6: 12294053-12294053 on Build GRCh38
3620076 c.250G>A p.G84R Chr.6: 12293957-12293957 on Build GRCh38
3620079 c.382G>A p.E128K Chr.6: 12294089-12294089 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753790282 Chr.6: 12294359-12294359 on Build GRCh38
rs779139823 Chr.6: 12294023-12294023 on Build GRCh38
rs370038651 Chr.6: 12294110-12294110 on Build GRCh38
rs373444338 Chr.6: 12294140-12294140 on Build GRCh38
rs879154023 Chr.6: 12294126-12294126 on Build GRCh38
rs869109750 Chr.6: 12293928-12293929 on Build GRCh38
rs773705696 Chr.6: 12294037-12294037 on Build GRCh38
rs533746234 Chr.6: 12293905-12293905 on Build GRCh38
rs748326404 Chr.6: 12294274-12294274 on Build GRCh38
rs777774061 Chr.6: 12293896-12293896 on Build GRCh38
rs756054900 Chr.6: 12294216-12294216 on Build GRCh38
rs777308039 Chr.6: 12294317-12294317 on Build GRCh38
rs1027641839 Chr.6: 12294156-12294156 on Build GRCh38
rs750362973 Chr.6: 12294212-12294212 on Build GRCh38
rs754988566 Chr.6: 12294375-12294375 on Build GRCh38
rs757031803 Chr.6: 12294000-12294000 on Build GRCh38
rs1005542743 Chr.6: 12294214-12294214 on Build GRCh38
rs1053860232 Chr.6: 12294159-12294159 on Build GRCh38
rs749579399 Chr.6: 12293928-12293928 on Build GRCh38
rs1007178617 Chr.6: 12293893-12293893 on Build GRCh38
rs35104761 Chr.6: 12293952-12293953 on Build GRCh38
rs764653980 Chr.6: 12293968-12293968 on Build GRCh38
rs769329570 Chr.6: 12294060-12294060 on Build GRCh38
rs957997210 Chr.6: 12293875-12293875 on Build GRCh38
rs138650134 Chr.6: 12294165-12294165 on Build GRCh38
rs752388910 Chr.6: 12293973-12293973 on Build GRCh38
rs750339407 Chr.6: 12294014-12294014 on Build GRCh38
rs761581288 Chr.6: 12294053-12294053 on Build GRCh38
rs770051193 Chr.6: 12294363-12294370 on Build GRCh38
rs565865651 Chr.6: 12294171-12294171 on Build GRCh38
rs756265142 Chr.6: 12294358-12294358 on Build GRCh38
rs537170013 Chr.6: 12294353-12294353 on Build GRCh38
rs912028745 Chr.6: 12293838-12293838 on Build GRCh38
rs529698751 Chr.6: 12294028-12294028 on Build GRCh38
rs140854546 Chr.6: 12294021-12294021 on Build GRCh38
rs766009921 Chr.6: 12294203-12294203 on Build GRCh38
rs5369 Chr.6: 12294025-12294025 on Build GRCh38
rs587777231 Chr.6: 12293978-12293978 on Build GRCh38
rs563358414 Chr.6: 12293848-12293848 on Build GRCh38
rs774749384 Chr.6: 12293942-12293942 on Build GRCh38
rs917166185 Chr.6: 12294029-12294029 on Build GRCh38
rs983858211 Chr.6: 12294368-12294368 on Build GRCh38
rs555033554 Chr.6: 12294219-12294219 on Build GRCh38
rs901425551 Chr.6: 12294313-12294313 on Build GRCh38
rs781099441 Chr.6: 12293901-12293901 on Build GRCh38
rs144271441 Chr.6: 12293943-12293943 on Build GRCh38
rs755076473 Chr.6: 12294259-12294259 on Build GRCh38
rs761610909 Chr.6: 12294128-12294128 on Build GRCh38
rs768907875 Chr.6: 12294022-12294022 on Build GRCh38
rs577094675 Chr.6: 12293899-12293899 on Build GRCh38
rs532399522 Chr.6: 12294146-12294146 on Build GRCh38
rs762501115 Chr.6: 12294328-12294328 on Build GRCh38
rs763046520 Chr.6: 12294372-12294375 on Build GRCh38
rs1032364349 Chr.6: 12293831-12293831 on Build GRCh38
rs763955920 Chr.6: 12293984-12293984 on Build GRCh38
rs374017499 Chr.6: 12294109-12294109 on Build GRCh38
rs201765566 Chr.6: 12294257-12294257 on Build GRCh38
rs182154246 Chr.6: 12293917-12293917 on Build GRCh38
rs764945093 Chr.6: 12294196-12294196 on Build GRCh38
rs976952215 Chr.6: 12293922-12293922 on Build GRCh38
rs762660018 Chr.6: 12294080-12294080 on Build GRCh38
rs951534547 Chr.6: 12294352-12294352 on Build GRCh38
rs759606950 Chr.6: 12294237-12294238 on Build GRCh38
rs778052405 Chr.6: 12294298-12294298 on Build GRCh38
rs762749838 Chr.6: 12293976-12293976 on Build GRCh38
rs369379949 Chr.6: 12294020-12294020 on Build GRCh38
rs1800543 Chr.6: 12293904-12293904 on Build GRCh38
rs774861385 Chr.6: 12294248-12294248 on Build GRCh38
rs150128166 Chr.6: 12294061-12294061 on Build GRCh38
rs568430735 Chr.6: 12294275-12294275 on Build GRCh38
rs886461028 Chr.6: 12294175-12294175 on Build GRCh38
rs145546137 Chr.6: 12294351-12294351 on Build GRCh38
rs771602064 Chr.6: 12294314-12294314 on Build GRCh38
rs551874067 Chr.6: 12294015-12294015 on Build GRCh38
rs748806941 Chr.6: 12294370-12294372 on Build GRCh38
rs994934646 Chr.6: 12294155-12294155 on Build GRCh38
rs376346685 Chr.6: 12294348-12294348 on Build GRCh38
rs139407538 Chr.6: 12293952-12293952 on Build GRCh38
rs1041556753 Chr.6: 12294089-12294089 on Build GRCh38
rs769786599 Chr.6: 12293912-12293912 on Build GRCh38
rs759763323 Chr.6: 12294278-12294278 on Build GRCh38
rs761149345 Chr.6: 12294055-12294055 on Build GRCh38
rs751438470 Chr.6: 12293990-12293990 on Build GRCh38
rs1019227124 Chr.6: 12294256-12294256 on Build GRCh38
rs767323529 Chr.6: 12294349-12294349 on Build GRCh38
rs781134655 Chr.6: 12294010-12294010 on Build GRCh38
rs746926884 Chr.6: 12293897-12293897 on Build GRCh38
rs587777234 Chr.6: 12293956-12293956 on Build GRCh38
rs376892399 Chr.6: 12293953-12293953 on Build GRCh38
rs150375573 Chr.6: 12294305-12294305 on Build GRCh38
rs766257520 Chr.6: 12294357-12294357 on Build GRCh38
rs149750515 Chr.6: 12294301-12294301 on Build GRCh38
rs935417700 Chr.6: 12294158-12294158 on Build GRCh38
rs1006883209 Chr.6: 12294229-12294229 on Build GRCh38
rs762278997 Chr.6: 12293944-12293944 on Build GRCh38
rs534545977 Chr.6: 12294208-12294208 on Build GRCh38
rs773985828 Chr.6: 12294343-12294343 on Build GRCh38
rs768294608 Chr.6: 12294335-12294335 on Build GRCh38
rs768995734 Chr.6: 12293930-12293930 on Build GRCh38
rs10478721 Chr.6: 12293923-12293924 on Build GRCh38
rs147381256 Chr.6: 12293945-12293945 on Build GRCh38
rs753876378 Chr.6: 12294237-12294237 on Build GRCh38
rs779053288 Chr.6: 12294272-12294272 on Build GRCh38

View Full Product Details