Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 132552764-132553290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAAR8
Entrez Gene ID
83551
Gene Name
trace amine associated receptor 8
Gene Aliases
GPR102, TA5, TAR5, TRAR5, TaR-5, TaR-8
UniGene
Hs.350571
Gene Chromosome Location
Chr.6: 132552693-132553721 on Build GRCh38
Transcripts
NM_053278.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1440534 c.592G>T p.V198L Chr.6: 132553284-132553284 on Build GRCh38
6105136 c.594G>T p.V198V Chr.6: 132553286-132553286 on Build GRCh38
7265741 c.559G>T p.G187C Chr.6: 132553251-132553251 on Build GRCh38
5983504 c.490G>A p.G164S Chr.6: 132553182-132553182 on Build GRCh38
1440532 c.141T>C p.F47F Chr.6: 132552833-132552833 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768492555 Chr.6: 132553161-132553161 on Build GRCh38
rs371119002 Chr.6: 132553144-132553144 on Build GRCh38
rs750548264 Chr.6: 132553208-132553208 on Build GRCh38
rs749782015 Chr.6: 132553064-132553064 on Build GRCh38
rs753281485 Chr.6: 132552860-132552860 on Build GRCh38
rs759249124 Chr.6: 132553189-132553189 on Build GRCh38
rs755363720 Chr.6: 132553008-132553008 on Build GRCh38
rs565162806 Chr.6: 132553214-132553214 on Build GRCh38
rs780089641 Chr.6: 132553213-132553213 on Build GRCh38
rs746165024 Chr.6: 132553076-132553076 on Build GRCh38
rs1008287233 Chr.6: 132553187-132553187 on Build GRCh38
rs772798598 Chr.6: 132553181-132553181 on Build GRCh38
rs772121875 Chr.6: 132553081-132553081 on Build GRCh38
rs759336088 Chr.6: 132552851-132552851 on Build GRCh38
rs764618279 Chr.6: 132553197-132553197 on Build GRCh38
rs140382740 Chr.6: 132553174-132553174 on Build GRCh38
rs147701770 Chr.6: 132553235-132553235 on Build GRCh38
rs201849771 Chr.6: 132552961-132552961 on Build GRCh38
rs752176570 Chr.6: 132552784-132552784 on Build GRCh38
rs774037269 Chr.6: 132552846-132552846 on Build GRCh38
rs767181287 Chr.6: 132552852-132552852 on Build GRCh38
rs1028774616 Chr.6: 132552938-132552938 on Build GRCh38
rs749371410 Chr.6: 132553248-132553248 on Build GRCh38
rs755191333 Chr.6: 132553217-132553217 on Build GRCh38
rs762393215 Chr.6: 132553203-132553203 on Build GRCh38
rs755411375 Chr.6: 132552881-132552881 on Build GRCh38
rs771219437 Chr.6: 132552845-132552845 on Build GRCh38
rs565721758 Chr.6: 132552911-132552911 on Build GRCh38
rs747344572 Chr.6: 132552801-132552801 on Build GRCh38
rs746760369 Chr.6: 132553101-132553101 on Build GRCh38
rs573336419 Chr.6: 132553151-132553151 on Build GRCh38
rs905570965 Chr.6: 132553058-132553058 on Build GRCh38
rs771189275 Chr.6: 132553254-132553254 on Build GRCh38
rs140749468 Chr.6: 132552967-132552967 on Build GRCh38
rs780209885 Chr.6: 132553156-132553156 on Build GRCh38
rs200511412 Chr.6: 132553184-132553184 on Build GRCh38
rs777239652 Chr.6: 132552786-132552786 on Build GRCh38
rs148940676 Chr.6: 132553125-132553125 on Build GRCh38
rs776144316 Chr.6: 132552997-132552997 on Build GRCh38
rs988974184 Chr.6: 132553183-132553183 on Build GRCh38
rs753051178 Chr.6: 132553046-132553046 on Build GRCh38
rs201637607 Chr.6: 132553262-132553262 on Build GRCh38
rs934850360 Chr.6: 132552897-132552897 on Build GRCh38
rs748544948 Chr.6: 132553074-132553074 on Build GRCh38
rs201581058 Chr.6: 132553182-132553182 on Build GRCh38
rs747201166 Chr.6: 132553159-132553159 on Build GRCh38
rs557972980 Chr.6: 132552968-132552968 on Build GRCh38
rs753140767 Chr.6: 132553131-132553131 on Build GRCh38
rs200975199 Chr.6: 132552965-132552965 on Build GRCh38
rs61744654 Chr.6: 132553138-132553138 on Build GRCh38
rs142407090 Chr.6: 132553247-132553247 on Build GRCh38
rs200443098 Chr.6: 132553059-132553059 on Build GRCh38
rs749331001 Chr.6: 132552909-132552909 on Build GRCh38
rs199609338 Chr.6: 132553145-132553145 on Build GRCh38
rs974057238 Chr.6: 132552847-132552847 on Build GRCh38
rs145710596 Chr.6: 132552888-132552888 on Build GRCh38
rs370810430 Chr.6: 132552945-132552945 on Build GRCh38
rs908340565 Chr.6: 132552959-132552959 on Build GRCh38
rs140210221 Chr.6: 132552842-132552842 on Build GRCh38
rs764622922 Chr.6: 132553011-132553011 on Build GRCh38
rs377412353 Chr.6: 132553049-132553049 on Build GRCh38
rs376100244 Chr.6: 132553237-132553237 on Build GRCh38
rs747845684 Chr.6: 132553236-132553236 on Build GRCh38
rs757975539 Chr.6: 132553040-132553040 on Build GRCh38
rs779840708 Chr.6: 132553083-132553083 on Build GRCh38
rs774670270 Chr.6: 132552928-132552928 on Build GRCh38
rs754187443 Chr.6: 132553201-132553201 on Build GRCh38
rs537371286 Chr.6: 132553124-132553124 on Build GRCh38
rs528884799 Chr.6: 132553255-132553255 on Build GRCh38
rs763965008 Chr.6: 132552980-132552980 on Build GRCh38
rs774192446 Chr.6: 132553188-132553188 on Build GRCh38
rs753543784 Chr.6: 132552790-132552790 on Build GRCh38
rs955436418 Chr.6: 132552948-132552948 on Build GRCh38
rs748725205 Chr.6: 132552891-132552891 on Build GRCh38
rs568468609 Chr.6: 132552887-132552887 on Build GRCh38
rs376422022 Chr.6: 132552859-132552859 on Build GRCh38
rs146818293 Chr.6: 132553267-132553267 on Build GRCh38
rs8192626 Chr.6: 132553150-132553150 on Build GRCh38
rs200069565 Chr.6: 132553186-132553186 on Build GRCh38
rs758386854 Chr.6: 132553054-132553054 on Build GRCh38
rs201703072 Chr.6: 132552799-132552799 on Build GRCh38
rs769405120 Chr.6: 132553112-132553112 on Build GRCh38
rs200972633 Chr.6: 132553142-132553142 on Build GRCh38
rs767994607 Chr.6: 132553042-132553042 on Build GRCh38
rs201563249 Chr.6: 132553003-132553003 on Build GRCh38
rs577896707 Chr.6: 132553104-132553104 on Build GRCh38
rs987421696 Chr.6: 132553175-132553175 on Build GRCh38
rs756919927 Chr.6: 132552865-132552865 on Build GRCh38
rs369666436 Chr.6: 132552889-132552889 on Build GRCh38
rs147240285 Chr.6: 132552899-132552899 on Build GRCh38
rs781770068 Chr.6: 132553035-132553035 on Build GRCh38
rs760612944 Chr.6: 132552971-132552971 on Build GRCh38
rs770043553 Chr.6: 132552817-132552817 on Build GRCh38
rs776591102 Chr.6: 132553108-132553108 on Build GRCh38
rs199551401 Chr.6: 132553135-132553135 on Build GRCh38
rs905767744 Chr.6: 132552788-132552788 on Build GRCh38
rs921290047 Chr.6: 132552849-132552849 on Build GRCh38
rs780955366 Chr.6: 132553169-132553169 on Build GRCh38
rs746051465 Chr.6: 132552932-132552932 on Build GRCh38
rs772595157 Chr.6: 132553116-132553116 on Build GRCh38
rs550876037 Chr.6: 132553246-132553246 on Build GRCh38
rs145344024 Chr.6: 132553136-132553136 on Build GRCh38
rs776290451 Chr.6: 132553230-132553231 on Build GRCh38
rs61741482 Chr.6: 132552800-132552800 on Build GRCh38
rs750414571 Chr.6: 132553152-132553152 on Build GRCh38
rs6912620 Chr.6: 132552803-132552803 on Build GRCh38
rs201369607 Chr.6: 132553139-132553139 on Build GRCh38
rs150893347 Chr.6: 132552821-132552821 on Build GRCh38
rs764899008 Chr.6: 132552872-132552872 on Build GRCh38
rs148349370 Chr.6: 132552802-132552802 on Build GRCh38
rs151214183 Chr.6: 132553153-132553153 on Build GRCh38
rs769553462 Chr.6: 132553179-132553179 on Build GRCh38
rs761634033 Chr.6: 132553111-132553111 on Build GRCh38
rs755658505 Chr.6: 132552785-132552785 on Build GRCh38
rs751900849 Chr.6: 132552874-132552874 on Build GRCh38
rs368641020 Chr.6: 132553264-132553264 on Build GRCh38
rs974372843 Chr.6: 132553026-132553026 on Build GRCh38
rs986883329 Chr.6: 132552949-132552949 on Build GRCh38
rs757403255 Chr.6: 132553073-132553073 on Build GRCh38
rs1002831945 Chr.6: 132552789-132552789 on Build GRCh38
rs201936385 Chr.6: 132553119-132553119 on Build GRCh38
rs999481257 Chr.6: 132552823-132552823 on Build GRCh38
rs1053251006 Chr.6: 132552934-132552934 on Build GRCh38
rs778384278 Chr.6: 132553057-132553057 on Build GRCh38
rs778011066 Chr.6: 132552907-132552907 on Build GRCh38
rs758448277 Chr.6: 132553211-132553211 on Build GRCh38
rs775543633 Chr.6: 132552857-132552857 on Build GRCh38
rs77605736 Chr.6: 132552834-132552834 on Build GRCh38
rs765614134 Chr.6: 132553207-132553207 on Build GRCh38

View Full Product Details