Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151702218-151702796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF3
Entrez Gene ID
11189
Gene Name
CUGBP, Elav-like family member 3
Gene Aliases
BRUNOL1, CAGH4, ERDA4, ETR-1, TNRC4
UniGene
Hs.26047
Gene Chromosome Location
Chr.1: 151700058-151716821 on Build GRCh38
Transcripts
NM_001172648.3 NM_001172649.3 NM_001291106.1 NM_001291107.1 NM_007185.6 XM_006711139.2 XM_006711141.2 XM_011509111.2 XM_011509112.2 XM_011509114.2 XM_017000145.1 XM_017000146.1

SNP Detail

SNP ID Chromosome Location
rs1049824390 Chr.1: 151702317-151702317 on Build GRCh38
rs998496002 Chr.1: 151702351-151702351 on Build GRCh38
rs903067581 Chr.1: 151702736-151702736 on Build GRCh38
rs932725678 Chr.1: 151702299-151702299 on Build GRCh38
rs1045779906 Chr.1: 151702661-151702662 on Build GRCh38
rs1004785581 Chr.1: 151702432-151702432 on Build GRCh38
rs946318727 Chr.1: 151702250-151702250 on Build GRCh38
rs911582042 Chr.1: 151702517-151702517 on Build GRCh38
rs1055943159 Chr.1: 151702292-151702292 on Build GRCh38
rs921754780 Chr.1: 151702492-151702492 on Build GRCh38
rs887164520 Chr.1: 151702668-151702669 on Build GRCh38
rs1035365995 Chr.1: 151702360-151702360 on Build GRCh38
rs967096342 Chr.1: 151702757-151702757 on Build GRCh38
rs998757486 Chr.1: 151702738-151702738 on Build GRCh38
rs1056374498 Chr.1: 151702641-151702641 on Build GRCh38
rs894553355 Chr.1: 151702293-151702293 on Build GRCh38
rs945679568 Chr.1: 151702524-151702529 on Build GRCh38
rs554817400 Chr.1: 151702745-151702745 on Build GRCh38
rs1308149 Chr.1: 151702447-151702447 on Build GRCh38
rs888943890 Chr.1: 151702318-151702318 on Build GRCh38
rs772652948 Chr.1: 151702751-151702751 on Build GRCh38
rs767421455 Chr.1: 151702739-151702739 on Build GRCh38
rs568845684 Chr.1: 151702705-151702705 on Build GRCh38
rs961663608 Chr.1: 151702650-151702650 on Build GRCh38
rs1012598044 Chr.1: 151702754-151702754 on Build GRCh38
rs952513334 Chr.1: 151702763-151702763 on Build GRCh38
rs574654608 Chr.1: 151702766-151702766 on Build GRCh38
rs984308940 Chr.1: 151702635-151702635 on Build GRCh38
rs926216842 Chr.1: 151702596-151702596 on Build GRCh38
rs886228867 Chr.1: 151702363-151702363 on Build GRCh38
rs1011441842 Chr.1: 151702335-151702335 on Build GRCh38
rs186131351 Chr.1: 151702734-151702734 on Build GRCh38
rs565960054 Chr.1: 151702703-151702703 on Build GRCh38
rs1037338660 Chr.1: 151702532-151702532 on Build GRCh38
rs1041375758 Chr.1: 151702537-151702537 on Build GRCh38
rs549565418 Chr.1: 151702291-151702291 on Build GRCh38
rs952958988 Chr.1: 151702633-151702633 on Build GRCh38
rs369604333 Chr.1: 151702657-151702657 on Build GRCh38
rs746896258 Chr.1: 151702369-151702370 on Build GRCh38
rs867985858 Chr.1: 151702636-151702636 on Build GRCh38
rs922659240 Chr.1: 151702297-151702297 on Build GRCh38
rs932694509 Chr.1: 151702702-151702702 on Build GRCh38
rs1041971352 Chr.1: 151702258-151702258 on Build GRCh38
rs922586905 Chr.1: 151702680-151702680 on Build GRCh38
rs905510455 Chr.1: 151702543-151702543 on Build GRCh38
rs1012670967 Chr.1: 151702495-151702495 on Build GRCh38
rs947166918 Chr.1: 151702724-151702724 on Build GRCh38
rs973648331 Chr.1: 151702492-151702493 on Build GRCh38
rs1004190648 Chr.1: 151702675-151702675 on Build GRCh38
rs987214921 Chr.1: 151702511-151702511 on Build GRCh38
rs1042911782 Chr.1: 151702725-151702725 on Build GRCh38
rs1022757135 Chr.1: 151702512-151702512 on Build GRCh38
rs538249474 Chr.1: 151702609-151702609 on Build GRCh38
rs373095851 Chr.1: 151702345-151702345 on Build GRCh38
rs1016898913 Chr.1: 151702679-151702679 on Build GRCh38
rs143213542 Chr.1: 151702474-151702478 on Build GRCh38
rs555417459 Chr.1: 151702344-151702344 on Build GRCh38
rs1041042191 Chr.1: 151702266-151702266 on Build GRCh38
rs113724761 Chr.1: 151702290-151702290 on Build GRCh38
rs1023098137 Chr.1: 151702755-151702755 on Build GRCh38

View Full Product Details