Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 56637604-56638166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TFAP2C
Entrez Gene ID
7022
Gene Name
transcription factor AP-2 gamma
Gene Aliases
AP2-GAMMA, ERF1, TFAP2G, hAP-2g
UniGene
Hs.473152
Gene Chromosome Location
Chr.20: 56629302-56639283 on Build GRCh38
Transcripts
NM_003222.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6305970 c.1105G>T p.D369Y Chr.20: 56637765-56637765 on Build GRCh38
1681785 c.1151C>T p.T384M Chr.20: 56637811-56637811 on Build GRCh38
4926825 c.1166G>A p.C389Y Chr.20: 56637826-56637826 on Build GRCh38
3548072 c.1080A>G p.K360K Chr.20: 56637740-56637740 on Build GRCh38
7393607 c.1083A>T p.E361D Chr.20: 56637743-56637743 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760198839 Chr.20: 56637814-56637814 on Build GRCh38
rs575758117 Chr.20: 56638010-56638010 on Build GRCh38
rs377580128 Chr.20: 56637935-56637935 on Build GRCh38
rs989800069 Chr.20: 56637684-56637684 on Build GRCh38
rs200833204 Chr.20: 56637759-56637759 on Build GRCh38
rs376628585 Chr.20: 56637794-56637794 on Build GRCh38
rs754990815 Chr.20: 56637872-56637872 on Build GRCh38
rs779975084 Chr.20: 56637768-56637768 on Build GRCh38
rs567468432 Chr.20: 56637747-56637747 on Build GRCh38
rs779359639 Chr.20: 56637725-56637725 on Build GRCh38
rs777617571 Chr.20: 56637787-56637787 on Build GRCh38
rs1029846845 Chr.20: 56637683-56637683 on Build GRCh38
rs1016751097 Chr.20: 56638102-56638102 on Build GRCh38
rs774229244 Chr.20: 56637796-56637796 on Build GRCh38
rs981340231 Chr.20: 56637667-56637667 on Build GRCh38
rs773813715 Chr.20: 56637688-56637688 on Build GRCh38
rs991026912 Chr.20: 56637977-56637977 on Build GRCh38
rs780029040 Chr.20: 56638025-56638025 on Build GRCh38
rs79298586 Chr.20: 56637625-56637625 on Build GRCh38
rs765488659 Chr.20: 56637830-56637830 on Build GRCh38
rs758686490 Chr.20: 56637731-56637731 on Build GRCh38
rs141841777 Chr.20: 56637795-56637795 on Build GRCh38
rs944877315 Chr.20: 56637803-56637803 on Build GRCh38
rs761254160 Chr.20: 56637701-56637701 on Build GRCh38
rs1006678421 Chr.20: 56637720-56637720 on Build GRCh38
rs146275217 Chr.20: 56637812-56637812 on Build GRCh38
rs1720 Chr.20: 56638026-56638026 on Build GRCh38
rs188914795 Chr.20: 56637994-56637994 on Build GRCh38
rs1004704661 Chr.20: 56637639-56637639 on Build GRCh38
rs192275203 Chr.20: 56638021-56638021 on Build GRCh38
rs1055266092 Chr.20: 56637635-56637635 on Build GRCh38
rs746683639 Chr.20: 56637923-56637923 on Build GRCh38
rs762204012 Chr.20: 56637955-56637955 on Build GRCh38
rs754404050 Chr.20: 56637705-56637705 on Build GRCh38
rs752884297 Chr.20: 56638002-56638002 on Build GRCh38
rs560333842 Chr.20: 56638048-56638048 on Build GRCh38
rs771772325 Chr.20: 56637904-56637904 on Build GRCh38
rs201529764 Chr.20: 56637704-56637704 on Build GRCh38
rs373593640 Chr.20: 56637922-56637922 on Build GRCh38
rs750848287 Chr.20: 56637833-56637833 on Build GRCh38
rs751727764 Chr.20: 56637981-56637981 on Build GRCh38
rs111436406 Chr.20: 56637884-56637884 on Build GRCh38
rs771818189 Chr.20: 56637811-56637811 on Build GRCh38
rs752308981 Chr.20: 56637699-56637699 on Build GRCh38
rs112229750 Chr.20: 56637726-56637726 on Build GRCh38
rs139254561 Chr.20: 56637925-56637925 on Build GRCh38
rs764891958 Chr.20: 56637881-56637881 on Build GRCh38
rs763308431 Chr.20: 56637957-56637957 on Build GRCh38
rs757508821 Chr.20: 56637707-56637707 on Build GRCh38
rs1026000875 Chr.20: 56638144-56638144 on Build GRCh38
rs369895748 Chr.20: 56637875-56637875 on Build GRCh38
rs749713518 Chr.20: 56638124-56638124 on Build GRCh38
rs952130053 Chr.20: 56637630-56637630 on Build GRCh38
rs112950894 Chr.20: 56637727-56637727 on Build GRCh38
rs775989958 Chr.20: 56638041-56638041 on Build GRCh38
rs1021223946 Chr.20: 56638105-56638105 on Build GRCh38
rs751782811 Chr.20: 56637854-56637854 on Build GRCh38
rs532632092 Chr.20: 56638070-56638070 on Build GRCh38
rs375002279 Chr.20: 56637678-56637678 on Build GRCh38
rs906999374 Chr.20: 56637937-56637937 on Build GRCh38
rs1006585551 Chr.20: 56638049-56638049 on Build GRCh38
rs6025024 Chr.20: 56637655-56637655 on Build GRCh38
rs368851871 Chr.20: 56637679-56637679 on Build GRCh38
rs769712375 Chr.20: 56637782-56637782 on Build GRCh38
rs552402336 Chr.20: 56638127-56638127 on Build GRCh38
rs1048520 Chr.20: 56638131-56638131 on Build GRCh38
rs758535882 Chr.20: 56637834-56637834 on Build GRCh38
rs555033312 Chr.20: 56638040-56638040 on Build GRCh38
rs142582019 Chr.20: 56637974-56637974 on Build GRCh38
rs763712130 Chr.20: 56637825-56637825 on Build GRCh38
rs138750457 Chr.20: 56637769-56637769 on Build GRCh38
rs369684214 Chr.20: 56637891-56637891 on Build GRCh38
rs538786666 Chr.20: 56637640-56637640 on Build GRCh38
rs759092983 Chr.20: 56637806-56637806 on Build GRCh38
rs182584272 Chr.20: 56638022-56638022 on Build GRCh38
rs778563366 Chr.20: 56637885-56637885 on Build GRCh38
rs773443436 Chr.20: 56637956-56637956 on Build GRCh38
rs758220246 Chr.20: 56638023-56638023 on Build GRCh38
rs371445486 Chr.20: 56637698-56637698 on Build GRCh38
rs993933526 Chr.20: 56638143-56638143 on Build GRCh38
rs755219913 Chr.20: 56637985-56637985 on Build GRCh38
rs780319718 Chr.20: 56637851-56637851 on Build GRCh38
rs898322603 Chr.20: 56638103-56638103 on Build GRCh38
rs759244596 Chr.20: 56637694-56637694 on Build GRCh38
rs748311088 Chr.20: 56637711-56637714 on Build GRCh38
rs762413823 Chr.20: 56637826-56637826 on Build GRCh38

View Full Product Details