Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 55154399-55154955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLGAP5
Entrez Gene ID
9787
Gene Name
DLG associated protein 5
Gene Aliases
DLG7, HURP
UniGene
Hs.77695
Gene Chromosome Location
Chr.14: 55148116-55191678 on Build GRCh38
Transcripts
NM_001146015.1 NM_014750.4 XM_017021840.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
319909 c.1999G>T p.G667C Chr.14: 55154681-55154681 on Build GRCh38
1370155 c.1998C>T p.A666A Chr.14: 55154682-55154682 on Build GRCh38
3744380 c.2062A>G p.R688G Chr.14: 55154618-55154618 on Build GRCh38
6274494 c.1975C>A p.Q659K Chr.14: 55154705-55154705 on Build GRCh38
1370156 c.1884C>T p.V628V Chr.14: 55154796-55154796 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs890839965 Chr.14: 55154578-55154578 on Build GRCh38
rs767363794 Chr.14: 55154706-55154706 on Build GRCh38
rs539939610 Chr.14: 55154489-55154489 on Build GRCh38
rs748670376 Chr.14: 55154663-55154663 on Build GRCh38
rs113515725 Chr.14: 55154734-55154734 on Build GRCh38
rs957101513 Chr.14: 55154602-55154602 on Build GRCh38
rs750039780 Chr.14: 55154840-55154840 on Build GRCh38
rs77977490 Chr.14: 55154832-55154832 on Build GRCh38
rs1011158350 Chr.14: 55154909-55154909 on Build GRCh38
rs374575158 Chr.14: 55154621-55154621 on Build GRCh38
rs767955554 Chr.14: 55154850-55154850 on Build GRCh38
rs1044355126 Chr.14: 55154428-55154428 on Build GRCh38
rs909237980 Chr.14: 55154477-55154477 on Build GRCh38
rs997200881 Chr.14: 55154560-55154560 on Build GRCh38
rs767418572 Chr.14: 55154627-55154627 on Build GRCh38
rs771030717 Chr.14: 55154692-55154692 on Build GRCh38
rs760552022 Chr.14: 55154577-55154577 on Build GRCh38
rs777328639 Chr.14: 55154579-55154579 on Build GRCh38
rs752584629 Chr.14: 55154507-55154507 on Build GRCh38
rs780425303 Chr.14: 55154655-55154655 on Build GRCh38
rs775283854 Chr.14: 55154820-55154820 on Build GRCh38
rs148718414 Chr.14: 55154649-55154649 on Build GRCh38
rs763905386 Chr.14: 55154830-55154830 on Build GRCh38
rs1052909638 Chr.14: 55154874-55154874 on Build GRCh38
rs758148959 Chr.14: 55154848-55154848 on Build GRCh38
rs370832129 Chr.14: 55154662-55154662 on Build GRCh38
rs113097879 Chr.14: 55154774-55154774 on Build GRCh38
rs753319263 Chr.14: 55154737-55154737 on Build GRCh38
rs761224954 Chr.14: 55154833-55154833 on Build GRCh38
rs573867710 Chr.14: 55154668-55154668 on Build GRCh38
rs759308831 Chr.14: 55154699-55154699 on Build GRCh38
rs151036101 Chr.14: 55154682-55154682 on Build GRCh38
rs1022802814 Chr.14: 55154664-55154664 on Build GRCh38
rs375996766 Chr.14: 55154783-55154783 on Build GRCh38
rs900761132 Chr.14: 55154538-55154538 on Build GRCh38
rs751345710 Chr.14: 55154617-55154617 on Build GRCh38
rs774549425 Chr.14: 55154815-55154815 on Build GRCh38
rs35357011 Chr.14: 55154836-55154837 on Build GRCh38
rs76406065 Chr.14: 55154426-55154426 on Build GRCh38
rs778388069 Chr.14: 55154803-55154803 on Build GRCh38
rs750168705 Chr.14: 55154760-55154760 on Build GRCh38
rs997643124 Chr.14: 55154558-55154558 on Build GRCh38
rs1007461255 Chr.14: 55154503-55154503 on Build GRCh38
rs763610880 Chr.14: 55154718-55154718 on Build GRCh38
rs557482778 Chr.14: 55154868-55154868 on Build GRCh38
rs753083958 Chr.14: 55154856-55154856 on Build GRCh38
rs555127027 Chr.14: 55154681-55154681 on Build GRCh38
rs928671892 Chr.14: 55154458-55154458 on Build GRCh38
rs1051949853 Chr.14: 55154567-55154567 on Build GRCh38
rs573517258 Chr.14: 55154764-55154764 on Build GRCh38
rs942906463 Chr.14: 55154782-55154782 on Build GRCh38
rs760621299 Chr.14: 55154825-55154825 on Build GRCh38
rs773276286 Chr.14: 55154424-55154424 on Build GRCh38
rs769201607 Chr.14: 55154658-55154658 on Build GRCh38
rs61738578 Chr.14: 55154661-55154661 on Build GRCh38
rs1035542349 Chr.14: 55154684-55154684 on Build GRCh38
rs762955352 Chr.14: 55154469-55154469 on Build GRCh38
rs755329111 Chr.14: 55154761-55154761 on Build GRCh38
rs745603650 Chr.14: 55154767-55154768 on Build GRCh38
rs142950079 Chr.14: 55154600-55154600 on Build GRCh38
rs376290753 Chr.14: 55154557-55154557 on Build GRCh38
rs142873440 Chr.14: 55154639-55154639 on Build GRCh38
rs778417884 Chr.14: 55154749-55154749 on Build GRCh38
rs192457260 Chr.14: 55154646-55154646 on Build GRCh38
rs938710577 Chr.14: 55154452-55154452 on Build GRCh38
rs748619222 Chr.14: 55154771-55154771 on Build GRCh38
rs980327622 Chr.14: 55154644-55154644 on Build GRCh38
rs774600670 Chr.14: 55154697-55154697 on Build GRCh38
rs950454914 Chr.14: 55154615-55154615 on Build GRCh38
rs901436348 Chr.14: 55154801-55154801 on Build GRCh38
rs1057036574 Chr.14: 55154457-55154457 on Build GRCh38
rs34350722 Chr.14: 55154625-55154626 on Build GRCh38
rs771096570 Chr.14: 55154810-55154810 on Build GRCh38
rs761971472 Chr.14: 55154582-55154582 on Build GRCh38
rs749770971 Chr.14: 55154552-55154554 on Build GRCh38
rs199676521 Chr.14: 55154636-55154636 on Build GRCh38
rs367727288 Chr.14: 55154818-55154818 on Build GRCh38
rs144312189 Chr.14: 55154859-55154859 on Build GRCh38
rs746831155 Chr.14: 55154798-55154798 on Build GRCh38
rs752684753 Chr.14: 55154634-55154634 on Build GRCh38
rs1034113053 Chr.14: 55154888-55154888 on Build GRCh38
rs764846714 Chr.14: 55154837-55154837 on Build GRCh38
rs891894906 Chr.14: 55154460-55154460 on Build GRCh38
rs778629422 Chr.14: 55154739-55154739 on Build GRCh38
rs927201571 Chr.14: 55154439-55154439 on Build GRCh38
rs922153487 Chr.14: 55154842-55154842 on Build GRCh38
rs767457584 Chr.14: 55154502-55154502 on Build GRCh38
rs773419752 Chr.14: 55154597-55154597 on Build GRCh38
rs766151895 Chr.14: 55154604-55154604 on Build GRCh38
rs904728100 Chr.14: 55154501-55154501 on Build GRCh38
rs78313577 Chr.14: 55154843-55154843 on Build GRCh38
rs913197464 Chr.14: 55154679-55154679 on Build GRCh38
rs201652858 Chr.14: 55154667-55154667 on Build GRCh38
rs141260005 Chr.14: 55154714-55154714 on Build GRCh38
rs749303143 Chr.14: 55154806-55154806 on Build GRCh38
rs139782311 Chr.14: 55154467-55154467 on Build GRCh38
rs771857722 Chr.14: 55154844-55154845 on Build GRCh38
rs376208826 Chr.14: 55154585-55154585 on Build GRCh38

View Full Product Details