Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 2880953-2881530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GYG2
Entrez Gene ID
8908
Gene Name
glycogenin 2
Gene Aliases
GN-2, GN2
UniGene
Hs.567381
Gene Chromosome Location
Chr.X: 2828712-2882820 on Build GRCh38
Transcripts
NM_001079855.1 NM_001184702.1 NM_001184703.1 NM_001184704.1 NM_003918.2 XM_011545600.2 XM_017029927.1 XM_017029928.1 XM_017029929.1 XM_017029930.1 XM_017029931.1

SNP Detail

SNP ID Chromosome Location
rs890187867 Chr.X: 2881274-2881274 on Build GRCh38
rs780452237 Chr.X: 2881003-2881003 on Build GRCh38
rs772010665 Chr.X: 2881193-2881193 on Build GRCh38
rs767936495 Chr.X: 2881247-2881247 on Build GRCh38
rs766537885 Chr.X: 2881217-2881217 on Build GRCh38
rs185729955 Chr.X: 2881273-2881273 on Build GRCh38
rs752264511 Chr.X: 2881334-2881334 on Build GRCh38
rs750993574 Chr.X: 2881251-2881263 on Build GRCh38
rs368784786 Chr.X: 2881238-2881238 on Build GRCh38
rs988884276 Chr.X: 2881138-2881138 on Build GRCh38
rs957442583 Chr.X: 2881106-2881106 on Build GRCh38
rs886396799 Chr.X: 2881149-2881149 on Build GRCh38
rs781620723 Chr.X: 2881127-2881127 on Build GRCh38
rs774182208 Chr.X: 2881168-2881168 on Build GRCh38
rs761898966 Chr.X: 2881084-2881084 on Build GRCh38
rs755356862 Chr.X: 2881125-2881125 on Build GRCh38
rs757804471 Chr.X: 2881250-2881251 on Build GRCh38
rs760583657 Chr.X: 2881200-2881200 on Build GRCh38
rs948219456 Chr.X: 2880998-2880998 on Build GRCh38
rs211659 Chr.X: 2881438-2881438 on Build GRCh38
rs773092854 Chr.X: 2880972-2880980 on Build GRCh38
rs141116662 Chr.X: 2881073-2881073 on Build GRCh38
rs982212253 Chr.X: 2881011-2881011 on Build GRCh38
rs754165582 Chr.X: 2881460-2881460 on Build GRCh38
rs1030059369 Chr.X: 2881269-2881269 on Build GRCh38
rs765334857 Chr.X: 2881207-2881207 on Build GRCh38
rs981479643 Chr.X: 2881178-2881178 on Build GRCh38
rs373273783 Chr.X: 2881201-2881201 on Build GRCh38
rs1031089704 Chr.X: 2881360-2881360 on Build GRCh38
rs144947653 Chr.X: 2881077-2881077 on Build GRCh38
rs769189858 Chr.X: 2881005-2881005 on Build GRCh38
rs767123474 Chr.X: 2881313-2881313 on Build GRCh38
rs374613941 Chr.X: 2881249-2881249 on Build GRCh38
rs760427381 Chr.X: 2881394-2881394 on Build GRCh38
rs758594897 Chr.X: 2881099-2881099 on Build GRCh38
rs148296798 Chr.X: 2881150-2881150 on Build GRCh38
rs755268697 Chr.X: 2881240-2881240 on Build GRCh38
rs767525546 Chr.X: 2881075-2881075 on Build GRCh38
rs777341177 Chr.X: 2881065-2881066 on Build GRCh38
rs772854040 Chr.X: 2881039-2881039 on Build GRCh38
rs999179734 Chr.X: 2881286-2881286 on Build GRCh38
rs770749717 Chr.X: 2881151-2881151 on Build GRCh38
rs745613491 Chr.X: 2881170-2881170 on Build GRCh38
rs765297484 Chr.X: 2881087-2881087 on Build GRCh38
rs916744464 Chr.X: 2880981-2880981 on Build GRCh38
rs747644628 Chr.X: 2881016-2881016 on Build GRCh38
rs769201100 Chr.X: 2881038-2881038 on Build GRCh38
rs766697178 Chr.X: 2881101-2881101 on Build GRCh38
rs990154275 Chr.X: 2881443-2881443 on Build GRCh38
rs770927137 Chr.X: 2881139-2881139 on Build GRCh38
rs377320038 Chr.X: 2881215-2881215 on Build GRCh38
rs781686308 Chr.X: 2881007-2881007 on Build GRCh38
rs141440542 Chr.X: 2881199-2881199 on Build GRCh38
rs777207663 Chr.X: 2881146-2881146 on Build GRCh38
rs750641717 Chr.X: 2881094-2881094 on Build GRCh38
rs868078341 Chr.X: 2881357-2881357 on Build GRCh38
rs1029331425 Chr.X: 2881357-2881365 on Build GRCh38
rs751880695 Chr.X: 2881109-2881109 on Build GRCh38
rs755646840 Chr.X: 2881144-2881144 on Build GRCh38
rs747489863 Chr.X: 2881004-2881004 on Build GRCh38
rs762579802 Chr.X: 2881052-2881052 on Build GRCh38
rs757627175 Chr.X: 2881487-2881487 on Build GRCh38
rs774338348 Chr.X: 2881055-2881055 on Build GRCh38
rs1006842374 Chr.X: 2881493-2881493 on Build GRCh38
rs770837433 Chr.X: 2881054-2881054 on Build GRCh38

View Full Product Details