Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 154562636-154563127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGB
Entrez Gene ID
2244
Gene Name
fibrinogen beta chain
Gene Aliases
HEL-S-78p
UniGene
Hs.300774
Gene Chromosome Location
Chr.4: 154562980-154572763 on Build GRCh38
Transcripts
NM_001184741.1 NM_005141.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3129192 c.52C>A p.L18I Chr.4: 154563070-154563070 on Build GRCh38
1207017 c.17C>A p.S6Y Chr.4: 154563035-154563035 on Build GRCh38
1618508 c.89C>A p.S30Y Chr.4: 154563107-154563107 on Build GRCh38
5672861 c.7A>C p.R3R Chr.4: 154563025-154563025 on Build GRCh38
4520036 c.103G>A p.D35N Chr.4: 154563121-154563121 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543846564 Chr.4: 154563018-154563018 on Build GRCh38
rs757727864 Chr.4: 154563013-154563014 on Build GRCh38
rs375340782 Chr.4: 154563102-154563102 on Build GRCh38
rs925687739 Chr.4: 154562705-154562705 on Build GRCh38
rs113961753 Chr.4: 154563010-154563010 on Build GRCh38
rs1050760505 Chr.4: 154562697-154562697 on Build GRCh38
rs560092277 Chr.4: 154562764-154562764 on Build GRCh38
rs756304323 Chr.4: 154563049-154563049 on Build GRCh38
rs376146726 Chr.4: 154563054-154563054 on Build GRCh38
rs1012372802 Chr.4: 154562698-154562698 on Build GRCh38
rs985002706 Chr.4: 154562963-154562963 on Build GRCh38
rs761398967 Chr.4: 154562994-154562994 on Build GRCh38
rs569306440 Chr.4: 154562663-154562663 on Build GRCh38
rs1006793912 Chr.4: 154562733-154562733 on Build GRCh38
rs1023575391 Chr.4: 154562881-154562881 on Build GRCh38
rs969402791 Chr.4: 154562913-154562913 on Build GRCh38
rs566860745 Chr.4: 154562979-154562979 on Build GRCh38
rs945677478 Chr.4: 154562956-154562956 on Build GRCh38
rs11099960 Chr.4: 154563097-154563097 on Build GRCh38
rs959424809 Chr.4: 154562691-154562691 on Build GRCh38
rs921809068 Chr.4: 154562827-154562827 on Build GRCh38
rs999370131 Chr.4: 154562790-154562790 on Build GRCh38
rs758377428 Chr.4: 154563084-154563084 on Build GRCh38
rs933625761 Chr.4: 154562685-154562685 on Build GRCh38
rs375151084 Chr.4: 154563007-154563007 on Build GRCh38
rs777895153 Chr.4: 154563052-154563052 on Build GRCh38
rs1012425059 Chr.4: 154563047-154563047 on Build GRCh38
rs1028131034 Chr.4: 154563089-154563089 on Build GRCh38
rs6053 Chr.4: 154563022-154563022 on Build GRCh38
rs1800787 Chr.4: 154562863-154562863 on Build GRCh38
rs776370184 Chr.4: 154562991-154562991 on Build GRCh38
rs190571140 Chr.4: 154562966-154562966 on Build GRCh38
rs36203394 Chr.4: 154563013-154563013 on Build GRCh38
rs576348167 Chr.4: 154563004-154563004 on Build GRCh38
rs962481266 Chr.4: 154562986-154562986 on Build GRCh38
rs1022260106 Chr.4: 154562737-154562737 on Build GRCh38
rs779988633 Chr.4: 154562975-154562975 on Build GRCh38
rs746963310 Chr.4: 154563104-154563104 on Build GRCh38
rs768398827 Chr.4: 154562989-154562989 on Build GRCh38
rs752925190 Chr.4: 154563035-154563035 on Build GRCh38
rs767891657 Chr.4: 154563028-154563028 on Build GRCh38
rs966019301 Chr.4: 154562802-154562802 on Build GRCh38
rs529951990 Chr.4: 154562726-154562726 on Build GRCh38
rs866418301 Chr.4: 154562999-154562999 on Build GRCh38
rs564463049 Chr.4: 154562984-154562984 on Build GRCh38
rs867253600 Chr.4: 154563030-154563030 on Build GRCh38
rs907758209 Chr.4: 154562787-154562787 on Build GRCh38
rs36203396 Chr.4: 154562812-154562812 on Build GRCh38
rs771942417 Chr.4: 154562869-154562869 on Build GRCh38
rs931723663 Chr.4: 154562851-154562851 on Build GRCh38
rs1800788 Chr.4: 154562762-154562762 on Build GRCh38
rs751892977 Chr.4: 154563019-154563019 on Build GRCh38
rs754297667 Chr.4: 154562975-154562976 on Build GRCh38

View Full Product Details