Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 6015040-6015437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIMP2
Entrez Gene ID
7965
Gene Name
aminoacyl tRNA synthetase complex interacting multifunctional protein 2
Gene Aliases
JTV-1, JTV1, P38
UniGene
Hs.301613
Gene Chromosome Location
Chr.7: 6009241-6023834 on Build GRCh38
Transcripts
NM_001326606.1 NM_001326607.1 NM_001326609.1 NM_001326611.1 NM_006303.3 XM_005249847.3 XM_005249848.2 XM_005249850.3 XM_006715772.2 XM_011515519.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6992976 c.319G>A p.D107N Chr.7: 6015329-6015329 on Build GRCh38
1313217 c.324G>C p.L108F Chr.7: 6015334-6015334 on Build GRCh38
3639954 c.230C>T p.S77F Chr.7: 6015240-6015240 on Build GRCh38
318613 c.173G>T p.R58L Chr.7: 6015183-6015183 on Build GRCh38
7226080 c.342+4G>A p.? Chr.7: 6015356-6015356 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553403052 Chr.7: 6015089-6015089 on Build GRCh38
rs201989680 Chr.7: 6015113-6015113 on Build GRCh38
rs775575419 Chr.7: 6015243-6015243 on Build GRCh38
rs768287678 Chr.7: 6015376-6015376 on Build GRCh38
rs761360918 Chr.7: 6015120-6015120 on Build GRCh38
rs774134623 Chr.7: 6015324-6015324 on Build GRCh38
rs751974218 Chr.7: 6015353-6015353 on Build GRCh38
rs765302683 Chr.7: 6015160-6015160 on Build GRCh38
rs1000124086 Chr.7: 6015191-6015191 on Build GRCh38
rs765088234 Chr.7: 6015236-6015236 on Build GRCh38
rs186004008 Chr.7: 6015406-6015406 on Build GRCh38
rs764132138 Chr.7: 6015322-6015322 on Build GRCh38
rs368275022 Chr.7: 6015164-6015164 on Build GRCh38
rs762568088 Chr.7: 6015348-6015348 on Build GRCh38
rs754061405 Chr.7: 6015329-6015329 on Build GRCh38
rs868784274 Chr.7: 6015229-6015231 on Build GRCh38
rs564323960 Chr.7: 6015257-6015257 on Build GRCh38
rs375652114 Chr.7: 6015085-6015085 on Build GRCh38
rs6979676 Chr.7: 6015356-6015356 on Build GRCh38
rs190724823 Chr.7: 6015183-6015183 on Build GRCh38
rs772717015 Chr.7: 6015208-6015208 on Build GRCh38
rs771560702 Chr.7: 6015124-6015124 on Build GRCh38
rs776388344 Chr.7: 6015223-6015223 on Build GRCh38
rs771011761 Chr.7: 6015378-6015378 on Build GRCh38
rs766404549 Chr.7: 6015325-6015325 on Build GRCh38
rs746565414 Chr.7: 6015295-6015295 on Build GRCh38
rs368845750 Chr.7: 6015294-6015294 on Build GRCh38
rs775979886 Chr.7: 6015281-6015281 on Build GRCh38
rs780796147 Chr.7: 6015357-6015357 on Build GRCh38
rs935481403 Chr.7: 6015390-6015390 on Build GRCh38
rs374710868 Chr.7: 6015292-6015292 on Build GRCh38
rs752636335 Chr.7: 6015162-6015162 on Build GRCh38
rs200084577 Chr.7: 6015193-6015193 on Build GRCh38
rs961881056 Chr.7: 6015283-6015283 on Build GRCh38
rs746484855 Chr.7: 6015214-6015214 on Build GRCh38
rs780028792 Chr.7: 6015150-6015150 on Build GRCh38
rs563423266 Chr.7: 6015377-6015377 on Build GRCh38
rs771057770 Chr.7: 6015077-6015077 on Build GRCh38
rs762651435 Chr.7: 6015321-6015321 on Build GRCh38
rs759397085 Chr.7: 6015154-6015154 on Build GRCh38
rs990903521 Chr.7: 6015336-6015336 on Build GRCh38
rs766098337 Chr.7: 6015151-6015151 on Build GRCh38
rs375295993 Chr.7: 6015341-6015341 on Build GRCh38
rs776458004 Chr.7: 6015304-6015304 on Build GRCh38
rs935062671 Chr.7: 6015291-6015291 on Build GRCh38
rs759696898 Chr.7: 6015340-6015340 on Build GRCh38
rs1052534772 Chr.7: 6015296-6015296 on Build GRCh38
rs529247469 Chr.7: 6015361-6015361 on Build GRCh38
rs374046815 Chr.7: 6015263-6015263 on Build GRCh38
rs778920772 Chr.7: 6015165-6015165 on Build GRCh38
rs374687277 Chr.7: 6015300-6015300 on Build GRCh38
rs774809039 Chr.7: 6015097-6015097 on Build GRCh38
rs769728324 Chr.7: 6015312-6015312 on Build GRCh38
rs751107887 Chr.7: 6015382-6015382 on Build GRCh38
rs372295850 Chr.7: 6015318-6015318 on Build GRCh38
rs762892628 Chr.7: 6015249-6015249 on Build GRCh38
rs1010788517 Chr.7: 6015064-6015064 on Build GRCh38
rs1040948360 Chr.7: 6015405-6015405 on Build GRCh38
rs577935179 Chr.7: 6015182-6015182 on Build GRCh38
rs1003655363 Chr.7: 6015099-6015099 on Build GRCh38
rs61751884 Chr.7: 6015280-6015280 on Build GRCh38
rs770539126 Chr.7: 6015302-6015302 on Build GRCh38
rs748648312 Chr.7: 6015196-6015196 on Build GRCh38
rs756418478 Chr.7: 6015178-6015178 on Build GRCh38
rs745450651 Chr.7: 6015306-6015306 on Build GRCh38
rs779417356 Chr.7: 6015192-6015192 on Build GRCh38
rs138662841 Chr.7: 6015255-6015255 on Build GRCh38
rs749240715 Chr.7: 6015389-6015391 on Build GRCh38
rs934141232 Chr.7: 6015260-6015260 on Build GRCh38
rs181812380 Chr.7: 6015360-6015360 on Build GRCh38
rs538013036 Chr.7: 6015307-6015307 on Build GRCh38
rs1023486596 Chr.7: 6015075-6015075 on Build GRCh38
rs749158394 Chr.7: 6015366-6015366 on Build GRCh38
rs750626949 Chr.7: 6015169-6015169 on Build GRCh38
rs373626338 Chr.7: 6015347-6015347 on Build GRCh38
rs983198652 Chr.7: 6015170-6015170 on Build GRCh38
rs765479530 Chr.7: 6015387-6015387 on Build GRCh38
rs770679503 Chr.7: 6015222-6015222 on Build GRCh38
rs946967833 Chr.7: 6015127-6015127 on Build GRCh38
rs755313952 Chr.7: 6015187-6015187 on Build GRCh38
rs753742279 Chr.7: 6015152-6015152 on Build GRCh38
rs764302112 Chr.7: 6015404-6015404 on Build GRCh38
rs150139402 Chr.7: 6015228-6015228 on Build GRCh38
rs772768946 Chr.7: 6015128-6015128 on Build GRCh38
rs752838723 Chr.7: 6015403-6015403 on Build GRCh38
rs376793759 Chr.7: 6015136-6015136 on Build GRCh38
rs140850823 Chr.7: 6015203-6015203 on Build GRCh38
rs368008892 Chr.7: 6015279-6015279 on Build GRCh38
rs758656381 Chr.7: 6015350-6015350 on Build GRCh38

View Full Product Details