Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67876365-67876964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDC4
Entrez Gene ID
23644
Gene Name
enhancer of mRNA decapping 4
Gene Aliases
GE1, Ge-1, HEDL5, HEDLS, RCD-8, RCD8
UniGene
Hs.75682
Gene Chromosome Location
Chr.16: 67873023-67884514 on Build GRCh38
Transcripts
NM_014329.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
122768 c.442G>T p.A148S Chr.16: 67876963-67876963 on Build GRCh38
5740698 c.416C>T p.A139V Chr.16: 67876937-67876937 on Build GRCh38
6651560 c.436G>T p.A146S Chr.16: 67876957-67876957 on Build GRCh38
3988604 c.333G>A p.K111K Chr.16: 67876581-67876581 on Build GRCh38
6905134 c.292G>A p.V98M Chr.16: 67876540-67876540 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35859129 Chr.16: 67876834-67876835 on Build GRCh38
rs62059359 Chr.16: 67876719-67876719 on Build GRCh38
rs781433949 Chr.16: 67876871-67876871 on Build GRCh38
rs890075280 Chr.16: 67876819-67876819 on Build GRCh38
rs767005076 Chr.16: 67876462-67876462 on Build GRCh38
rs758843505 Chr.16: 67876839-67876839 on Build GRCh38
rs774085904 Chr.16: 67876796-67876796 on Build GRCh38
rs775563257 Chr.16: 67876598-67876598 on Build GRCh38
rs1017831429 Chr.16: 67876761-67876761 on Build GRCh38
rs972286307 Chr.16: 67876463-67876464 on Build GRCh38
rs550534829 Chr.16: 67876409-67876409 on Build GRCh38
rs751155152 Chr.16: 67876449-67876449 on Build GRCh38
rs771192856 Chr.16: 67876586-67876586 on Build GRCh38
rs768747829 Chr.16: 67876898-67876898 on Build GRCh38
rs577302689 Chr.16: 67876736-67876736 on Build GRCh38
rs779256251 Chr.16: 67876515-67876515 on Build GRCh38
rs538704143 Chr.16: 67876396-67876396 on Build GRCh38
rs774386928 Chr.16: 67876902-67876902 on Build GRCh38
rs755334363 Chr.16: 67876511-67876511 on Build GRCh38
rs966781119 Chr.16: 67876863-67876863 on Build GRCh38
rs929098782 Chr.16: 67876699-67876699 on Build GRCh38
rs370858216 Chr.16: 67876654-67876654 on Build GRCh38
rs554406609 Chr.16: 67876524-67876524 on Build GRCh38
rs771963310 Chr.16: 67876928-67876928 on Build GRCh38
rs1022359944 Chr.16: 67876438-67876438 on Build GRCh38
rs1022309002 Chr.16: 67876848-67876848 on Build GRCh38
rs978205285 Chr.16: 67876926-67876926 on Build GRCh38
rs1009547732 Chr.16: 67876755-67876755 on Build GRCh38
rs186605407 Chr.16: 67876770-67876770 on Build GRCh38
rs536161425 Chr.16: 67876502-67876502 on Build GRCh38
rs989905747 Chr.16: 67876428-67876428 on Build GRCh38
rs757338620 Chr.16: 67876857-67876857 on Build GRCh38
rs747874119 Chr.16: 67876917-67876917 on Build GRCh38
rs749240790 Chr.16: 67876604-67876604 on Build GRCh38
rs1009188109 Chr.16: 67876401-67876401 on Build GRCh38
rs1008232002 Chr.16: 67876413-67876413 on Build GRCh38
rs924477174 Chr.16: 67876504-67876504 on Build GRCh38
rs752969704 Chr.16: 67876838-67876838 on Build GRCh38
rs1001072909 Chr.16: 67876814-67876814 on Build GRCh38
rs917632949 Chr.16: 67876701-67876701 on Build GRCh38
rs768473976 Chr.16: 67876683-67876683 on Build GRCh38
rs890660273 Chr.16: 67876737-67876737 on Build GRCh38
rs558507618 Chr.16: 67876649-67876649 on Build GRCh38
rs761388290 Chr.16: 67876450-67876450 on Build GRCh38
rs749125922 Chr.16: 67876573-67876573 on Build GRCh38
rs760169301 Chr.16: 67876634-67876634 on Build GRCh38
rs774066997 Chr.16: 67876520-67876520 on Build GRCh38
rs62059358 Chr.16: 67876437-67876437 on Build GRCh38
rs904014097 Chr.16: 67876774-67876774 on Build GRCh38
rs1047879591 Chr.16: 67876702-67876702 on Build GRCh38
rs879563275 Chr.16: 67876681-67876681 on Build GRCh38
rs182290690 Chr.16: 67876633-67876633 on Build GRCh38
rs377331935 Chr.16: 67876609-67876609 on Build GRCh38
rs761488214 Chr.16: 67876629-67876629 on Build GRCh38
rs765594590 Chr.16: 67876829-67876829 on Build GRCh38
rs778091766 Chr.16: 67876541-67876541 on Build GRCh38
rs745809796 Chr.16: 67876590-67876590 on Build GRCh38
rs376569739 Chr.16: 67876445-67876445 on Build GRCh38
rs747034405 Chr.16: 67876542-67876542 on Build GRCh38
rs749933134 Chr.16: 67876475-67876475 on Build GRCh38
rs368608238 Chr.16: 67876622-67876622 on Build GRCh38
rs938589277 Chr.16: 67876514-67876514 on Build GRCh38
rs777844523 Chr.16: 67876844-67876844 on Build GRCh38
rs866262400 Chr.16: 67876481-67876481 on Build GRCh38
rs780521664 Chr.16: 67876488-67876488 on Build GRCh38
rs747302556 Chr.16: 67876846-67876846 on Build GRCh38
rs1049966072 Chr.16: 67876817-67876817 on Build GRCh38
rs900765117 Chr.16: 67876786-67876786 on Build GRCh38
rs749190020 Chr.16: 67876890-67876890 on Build GRCh38
rs984667834 Chr.16: 67876509-67876509 on Build GRCh38
rs373238184 Chr.16: 67876480-67876480 on Build GRCh38
rs964767369 Chr.16: 67876410-67876410 on Build GRCh38
rs765943856 Chr.16: 67876640-67876640 on Build GRCh38
rs767046538 Chr.16: 67876630-67876630 on Build GRCh38
rs1037340396 Chr.16: 67876659-67876659 on Build GRCh38

View Full Product Details