Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 52712473-52712998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LECT1
Entrez Gene ID
11061
Gene Name
leukocyte cell derived chemotaxin 1
Gene Aliases
BRICD3, CHM-I, CHM1, MYETS1
UniGene
Hs.421391
Gene Chromosome Location
Chr.13: 52703264-52739812 on Build GRCh38
Transcripts
NM_001011705.1 NM_007015.2 XM_011534897.2 XM_011534898.2 XM_011534899.2 XM_011534900.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3885432 c.516G>A p.W172* Chr.13: 52712822-52712822 on Build GRCh38
1300254 c.499G>C p.E167Q Chr.13: 52712839-52712839 on Build GRCh38
6576547 c.480C>G p.I160M Chr.13: 52712858-52712858 on Build GRCh38
5362143 c.582T>C p.G194G Chr.13: 52712756-52712756 on Build GRCh38
234903 c.540G>A p.K180K Chr.13: 52712798-52712798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925169199 Chr.13: 52712853-52712853 on Build GRCh38
rs199719465 Chr.13: 52712712-52712712 on Build GRCh38
rs371805564 Chr.13: 52712667-52712667 on Build GRCh38
rs113671906 Chr.13: 52712646-52712646 on Build GRCh38
rs62637607 Chr.13: 52712687-52712687 on Build GRCh38
rs374124583 Chr.13: 52712884-52712884 on Build GRCh38
rs566296905 Chr.13: 52712647-52712647 on Build GRCh38
rs955061745 Chr.13: 52712751-52712751 on Build GRCh38
rs1017174333 Chr.13: 52712674-52712674 on Build GRCh38
rs779508037 Chr.13: 52712703-52712703 on Build GRCh38
rs1014753508 Chr.13: 52712567-52712567 on Build GRCh38
rs746121576 Chr.13: 52712856-52712856 on Build GRCh38
rs149442067 Chr.13: 52712792-52712792 on Build GRCh38
rs756209622 Chr.13: 52712717-52712717 on Build GRCh38
rs148027577 Chr.13: 52712557-52712557 on Build GRCh38
rs911846670 Chr.13: 52712774-52712774 on Build GRCh38
rs905965654 Chr.13: 52712979-52712979 on Build GRCh38
rs752869305 Chr.13: 52712685-52712686 on Build GRCh38
rs138871406 Chr.13: 52712783-52712783 on Build GRCh38
rs768104398 Chr.13: 52712678-52712679 on Build GRCh38
rs761416837 Chr.13: 52712898-52712898 on Build GRCh38
rs748993805 Chr.13: 52712599-52712599 on Build GRCh38
rs376449381 Chr.13: 52712832-52712832 on Build GRCh38
rs766642834 Chr.13: 52712665-52712665 on Build GRCh38
rs555196133 Chr.13: 52712720-52712720 on Build GRCh38
rs904704066 Chr.13: 52712508-52712508 on Build GRCh38
rs757378127 Chr.13: 52712847-52712847 on Build GRCh38
rs763592902 Chr.13: 52712768-52712768 on Build GRCh38
rs781495712 Chr.13: 52712852-52712852 on Build GRCh38
rs375863795 Chr.13: 52712670-52712670 on Build GRCh38
rs985970015 Chr.13: 52712755-52712755 on Build GRCh38
rs759030253 Chr.13: 52712819-52712819 on Build GRCh38
rs373372110 Chr.13: 52712864-52712864 on Build GRCh38
rs771196703 Chr.13: 52712727-52712727 on Build GRCh38
rs34222212 Chr.13: 52712617-52712618 on Build GRCh38
rs866620960 Chr.13: 52712839-52712839 on Build GRCh38
rs747220935 Chr.13: 52712726-52712726 on Build GRCh38
rs369730829 Chr.13: 52712721-52712721 on Build GRCh38
rs1014664685 Chr.13: 52712659-52712659 on Build GRCh38
rs1025305838 Chr.13: 52712724-52712724 on Build GRCh38
rs770308795 Chr.13: 52712628-52712628 on Build GRCh38
rs2147692 Chr.13: 52712888-52712888 on Build GRCh38
rs760591570 Chr.13: 52712978-52712978 on Build GRCh38
rs143574181 Chr.13: 52712543-52712543 on Build GRCh38
rs7330220 Chr.13: 52712815-52712815 on Build GRCh38
rs150210923 Chr.13: 52712759-52712759 on Build GRCh38
rs754113180 Chr.13: 52712799-52712799 on Build GRCh38
rs755394845 Chr.13: 52712812-52712812 on Build GRCh38
rs868055725 Chr.13: 52712745-52712745 on Build GRCh38
rs375786315 Chr.13: 52712883-52712883 on Build GRCh38
rs745702710 Chr.13: 52712758-52712758 on Build GRCh38
rs373490649 Chr.13: 52712818-52712818 on Build GRCh38
rs553523982 Chr.13: 52712795-52712796 on Build GRCh38
rs950109401 Chr.13: 52712964-52712964 on Build GRCh38
rs547508641 Chr.13: 52712596-52712596 on Build GRCh38
rs775381625 Chr.13: 52712762-52712762 on Build GRCh38
rs772198667 Chr.13: 52712872-52712872 on Build GRCh38
rs776219123 Chr.13: 52712748-52712748 on Build GRCh38
rs747115835 Chr.13: 52712843-52712843 on Build GRCh38
rs773912770 Chr.13: 52712777-52712777 on Build GRCh38
rs1045839367 Chr.13: 52712609-52712609 on Build GRCh38
rs183600958 Chr.13: 52712957-52712957 on Build GRCh38
rs755732483 Chr.13: 52712503-52712503 on Build GRCh38
rs748191727 Chr.13: 52712713-52712713 on Build GRCh38
rs762920734 Chr.13: 52712763-52712763 on Build GRCh38
rs993798213 Chr.13: 52712723-52712723 on Build GRCh38
rs200298078 Chr.13: 52712685-52712685 on Build GRCh38
rs571587840 Chr.13: 52712551-52712551 on Build GRCh38
rs906012883 Chr.13: 52712612-52712612 on Build GRCh38
rs1053058826 Chr.13: 52712534-52712534 on Build GRCh38
rs199566973 Chr.13: 52712830-52712830 on Build GRCh38

View Full Product Details