Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 49705428-49705721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KPNA3
Entrez Gene ID
3839
Gene Name
karyopherin subunit alpha 3
Gene Aliases
IPOA4, SRP1, SRP1gamma, SRP4, hSRP1
UniGene
Hs.527919
Gene Chromosome Location
Chr.13: 49699307-49792921 on Build GRCh38
Transcripts
NM_002267.3 XM_017020561.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6490096 c.1299T>G p.D433E Chr.13: 49705694-49705694 on Build GRCh38
5704736 c.1301G>T p.G434V Chr.13: 49705692-49705692 on Build GRCh38
4047983 c.1276C>G p.Q426E Chr.13: 49705717-49705717 on Build GRCh38
1367387 c.1326C>T p.A442A Chr.13: 49705667-49705667 on Build GRCh38
4047982 c.1357A>G p.I453V Chr.13: 49705636-49705636 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141253451 Chr.13: 49705666-49705666 on Build GRCh38
rs746333005 Chr.13: 49705463-49705463 on Build GRCh38
rs201836074 Chr.13: 49705580-49705580 on Build GRCh38
rs757597269 Chr.13: 49705590-49705593 on Build GRCh38
rs919917685 Chr.13: 49705558-49705558 on Build GRCh38
rs773285455 Chr.13: 49705653-49705653 on Build GRCh38
rs528772095 Chr.13: 49705470-49705470 on Build GRCh38
rs543075858 Chr.13: 49705501-49705501 on Build GRCh38
rs150519849 Chr.13: 49705459-49705459 on Build GRCh38
rs758813009 Chr.13: 49705503-49705503 on Build GRCh38
rs1047076003 Chr.13: 49705473-49705473 on Build GRCh38
rs200666130 Chr.13: 49705640-49705640 on Build GRCh38
rs1038991192 Chr.13: 49705566-49705566 on Build GRCh38
rs369254638 Chr.13: 49705648-49705648 on Build GRCh38
rs748910216 Chr.13: 49705575-49705591 on Build GRCh38
rs912792431 Chr.13: 49705631-49705631 on Build GRCh38
rs79884028 Chr.13: 49705636-49705636 on Build GRCh38
rs775705842 Chr.13: 49705606-49705606 on Build GRCh38
rs887397797 Chr.13: 49705523-49705523 on Build GRCh38
rs535292951 Chr.13: 49705667-49705667 on Build GRCh38
rs372027513 Chr.13: 49705646-49705646 on Build GRCh38
rs1002758758 Chr.13: 49705585-49705585 on Build GRCh38
rs934007663 Chr.13: 49705471-49705471 on Build GRCh38
rs747369206 Chr.13: 49705595-49705595 on Build GRCh38
rs781677183 Chr.13: 49705587-49705587 on Build GRCh38
rs866925883 Chr.13: 49705654-49705654 on Build GRCh38
rs368142442 Chr.13: 49705610-49705610 on Build GRCh38
rs553205303 Chr.13: 49705639-49705639 on Build GRCh38
rs941611092 Chr.13: 49705559-49705559 on Build GRCh38
rs961442553 Chr.13: 49705508-49705508 on Build GRCh38
rs780332620 Chr.13: 49705522-49705522 on Build GRCh38

View Full Product Details