Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 114906439-114907016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTR2C
Entrez Gene ID
3358
Gene Name
5-hydroxytryptamine receptor 2C
Gene Aliases
5-HT1C, 5-HT2C, 5-HTR2C, 5HTR2C, HTR1C
UniGene
Hs.149037
Gene Chromosome Location
Chr.X: 114584078-114910061 on Build GRCh38
Transcripts
NM_000868.3 NM_001256760.2 NM_001256761.2

SNP Detail

SNP ID Chromosome Location
rs782047384 Chr.X: 114906885-114906885 on Build GRCh38
rs782747846 Chr.X: 114906756-114906756 on Build GRCh38
rs942398034 Chr.X: 114906606-114906606 on Build GRCh38
rs202077519 Chr.X: 114906960-114906960 on Build GRCh38
rs781957274 Chr.X: 114906787-114906787 on Build GRCh38
rs201632841 Chr.X: 114906579-114906579 on Build GRCh38
rs782359396 Chr.X: 114906893-114906893 on Build GRCh38
rs77459121 Chr.X: 114906877-114906877 on Build GRCh38
rs199674890 Chr.X: 114906551-114906551 on Build GRCh38
rs781891174 Chr.X: 114906903-114906903 on Build GRCh38
rs782307054 Chr.X: 114906538-114906538 on Build GRCh38
rs782814171 Chr.X: 114906901-114906901 on Build GRCh38
rs903627201 Chr.X: 114906659-114906659 on Build GRCh38
rs200607550 Chr.X: 114906921-114906921 on Build GRCh38
rs782188199 Chr.X: 114906688-114906688 on Build GRCh38
rs781954366 Chr.X: 114906897-114906897 on Build GRCh38
rs782118303 Chr.X: 114906898-114906898 on Build GRCh38
rs191640720 Chr.X: 114906497-114906497 on Build GRCh38
rs781788896 Chr.X: 114906780-114906780 on Build GRCh38
rs782371325 Chr.X: 114906562-114906562 on Build GRCh38
rs144618891 Chr.X: 114906602-114906602 on Build GRCh38
rs201964979 Chr.X: 114906910-114906910 on Build GRCh38
rs374812243 Chr.X: 114906755-114906755 on Build GRCh38
rs144149179 Chr.X: 114906840-114906840 on Build GRCh38
rs78281154 Chr.X: 114906928-114906928 on Build GRCh38
rs781927832 Chr.X: 114906586-114906586 on Build GRCh38
rs782028790 Chr.X: 114906759-114906759 on Build GRCh38
rs782121542 Chr.X: 114906737-114906737 on Build GRCh38
rs781938075 Chr.X: 114906683-114906683 on Build GRCh38
rs200171832 Chr.X: 114906583-114906583 on Build GRCh38
rs782198364 Chr.X: 114906978-114906978 on Build GRCh38
rs781830737 Chr.X: 114906919-114906919 on Build GRCh38
rs1033195176 Chr.X: 114906665-114906665 on Build GRCh38
rs782421726 Chr.X: 114906681-114906681 on Build GRCh38
rs782140216 Chr.X: 114906772-114906772 on Build GRCh38
rs782662523 Chr.X: 114906951-114906951 on Build GRCh38
rs782711180 Chr.X: 114906917-114906917 on Build GRCh38
rs782566294 Chr.X: 114906805-114906805 on Build GRCh38
rs200776874 Chr.X: 114906577-114906577 on Build GRCh38
rs781854947 Chr.X: 114906797-114906797 on Build GRCh38
rs201171585 Chr.X: 114906956-114906956 on Build GRCh38
rs368641796 Chr.X: 114906680-114906680 on Build GRCh38
rs199953828 Chr.X: 114906609-114906609 on Build GRCh38
rs187900506 Chr.X: 114906632-114906632 on Build GRCh38
rs782809944 Chr.X: 114906636-114906636 on Build GRCh38
rs368256898 Chr.X: 114906900-114906900 on Build GRCh38
rs782383804 Chr.X: 114906850-114906850 on Build GRCh38
rs782558692 Chr.X: 114906949-114906949 on Build GRCh38
rs781957272 Chr.X: 114906698-114906698 on Build GRCh38
rs201461246 Chr.X: 114906542-114906542 on Build GRCh38
rs781863069 Chr.X: 114906596-114906596 on Build GRCh38
rs781884311 Chr.X: 114906642-114906642 on Build GRCh38
rs927404564 Chr.X: 114906505-114906505 on Build GRCh38
rs183232813 Chr.X: 114906524-114906524 on Build GRCh38
rs148470204 Chr.X: 114906816-114906816 on Build GRCh38
rs782066714 Chr.X: 114906904-114906904 on Build GRCh38
rs146559807 Chr.X: 114906871-114906871 on Build GRCh38
rs782628883 Chr.X: 114906855-114906855 on Build GRCh38
rs782644122 Chr.X: 114906668-114906668 on Build GRCh38
rs971437186 Chr.X: 114906464-114906464 on Build GRCh38
rs782260813 Chr.X: 114906558-114906558 on Build GRCh38
rs201174125 Chr.X: 114906895-114906895 on Build GRCh38
rs782626623 Chr.X: 114906845-114906845 on Build GRCh38
rs781931599 Chr.X: 114906884-114906884 on Build GRCh38
rs149710250 Chr.X: 114906591-114906591 on Build GRCh38
rs782569195 Chr.X: 114906652-114906652 on Build GRCh38
rs201583846 Chr.X: 114906689-114906689 on Build GRCh38
rs980208671 Chr.X: 114906501-114906501 on Build GRCh38
rs782231891 Chr.X: 114906656-114906656 on Build GRCh38
rs781866325 Chr.X: 114906938-114906938 on Build GRCh38
rs782442713 Chr.X: 114906844-114906844 on Build GRCh38
rs200743712 Chr.X: 114906647-114906647 on Build GRCh38
rs973906280 Chr.X: 114906924-114906924 on Build GRCh38
rs782783355 Chr.X: 114906660-114906660 on Build GRCh38

View Full Product Details