Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 197042270-197042869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLG1
Entrez Gene ID
1739
Gene Name
discs large MAGUK scaffold protein 1
Gene Aliases
DLGH1, SAP-97, SAP97, dJ1061C18.1.1, hdlg
UniGene
Hs.292549
Gene Chromosome Location
Chr.3: 197042560-197299272 on Build GRCh38
Transcripts
NM_001098424.1 NM_001204386.1 NM_001204387.1 NM_001204388.1 NM_001290983.1 NM_004087.2 XM_005269289.3 XM_011512502.2 XM_011512503.1 XM_011512505.1 XM_011512506.1 XM_017005800.1 XM_017005801.1 XM_017005802.1 XM_017005804.1 XM_017005805.1 XM_017005806.1 XM_017005807.1 XM_017005808.1 XM_017005809.1 XM_017005810.1 XM_017005811.1 XM_017005812.1 XM_017005813.1 XM_017005814.1 XM_017005815.1 XM_017005816.1 XM_017005817.1 XM_017005818.1 XM_017005820.1 XM_017005821.1 XM_017005822.1 XM_017005823.1 XM_017005824.1

SNP Detail

SNP ID Chromosome Location
rs577340320 Chr.3: 197042400-197042400 on Build GRCh38
rs567148808 Chr.3: 197042347-197042347 on Build GRCh38
rs979531572 Chr.3: 197042415-197042421 on Build GRCh38
rs998576616 Chr.3: 197042672-197042672 on Build GRCh38
rs780868561 Chr.3: 197042587-197042590 on Build GRCh38
rs552616180 Chr.3: 197042375-197042375 on Build GRCh38
rs968405763 Chr.3: 197042650-197042650 on Build GRCh38
rs773317721 Chr.3: 197042443-197042443 on Build GRCh38
rs925803708 Chr.3: 197042808-197042808 on Build GRCh38
rs746024598 Chr.3: 197042769-197042769 on Build GRCh38
rs561261767 Chr.3: 197042511-197042511 on Build GRCh38
rs797020192 Chr.3: 197042368-197042368 on Build GRCh38
rs895512186 Chr.3: 197042481-197042481 on Build GRCh38
rs753736697 Chr.3: 197042583-197042583 on Build GRCh38
rs762303201 Chr.3: 197042414-197042420 on Build GRCh38
rs895576839 Chr.3: 197042819-197042819 on Build GRCh38
rs934019698 Chr.3: 197042728-197042728 on Build GRCh38
rs542552769 Chr.3: 197042505-197042505 on Build GRCh38
rs781036385 Chr.3: 197042634-197042635 on Build GRCh38
rs185904397 Chr.3: 197042359-197042359 on Build GRCh38
rs1057034427 Chr.3: 197042545-197042545 on Build GRCh38
rs1002802369 Chr.3: 197042399-197042399 on Build GRCh38
rs1021712002 Chr.3: 197042640-197042640 on Build GRCh38
rs751460105 Chr.3: 197042550-197042552 on Build GRCh38
rs113925412 Chr.3: 197042408-197042408 on Build GRCh38
rs1022554559 Chr.3: 197042493-197042493 on Build GRCh38
rs144683931 Chr.3: 197042421-197042422 on Build GRCh38
rs911202654 Chr.3: 197042763-197042763 on Build GRCh38
rs879212149 Chr.3: 197042418-197042420 on Build GRCh38
rs923544108 Chr.3: 197042450-197042450 on Build GRCh38
rs143968303 Chr.3: 197042418-197042420 on Build GRCh38
rs758821402 Chr.3: 197042817-197042817 on Build GRCh38
rs559860270 Chr.3: 197042809-197042809 on Build GRCh38
rs770942396 Chr.3: 197042683-197042683 on Build GRCh38
rs1014384184 Chr.3: 197042833-197042833 on Build GRCh38
rs933988224 Chr.3: 197042316-197042316 on Build GRCh38
rs976687771 Chr.3: 197042698-197042698 on Build GRCh38
rs1053315555 Chr.3: 197042815-197042815 on Build GRCh38
rs1013025955 Chr.3: 197042408-197042409 on Build GRCh38
rs922706915 Chr.3: 197042687-197042687 on Build GRCh38
rs142887611 Chr.3: 197042409-197042412 on Build GRCh38
rs571628624 Chr.3: 197042320-197042321 on Build GRCh38
rs968479604 Chr.3: 197042413-197042417 on Build GRCh38
rs909257976 Chr.3: 197042501-197042501 on Build GRCh38
rs894416172 Chr.3: 197042579-197042579 on Build GRCh38
rs897678973 Chr.3: 197042522-197042522 on Build GRCh38
rs984311386 Chr.3: 197042524-197042524 on Build GRCh38
rs547594097 Chr.3: 197042723-197042723 on Build GRCh38
rs1050892959 Chr.3: 197042497-197042497 on Build GRCh38
rs931991047 Chr.3: 197042474-197042474 on Build GRCh38
rs953800359 Chr.3: 197042726-197042726 on Build GRCh38
rs765545741 Chr.3: 197042417-197042418 on Build GRCh38
rs774491912 Chr.3: 197042414-197042416 on Build GRCh38
rs986112955 Chr.3: 197042731-197042732 on Build GRCh38
rs1042982789 Chr.3: 197042794-197042794 on Build GRCh38
rs144235731 Chr.3: 197042628-197042628 on Build GRCh38
rs556745887 Chr.3: 197042409-197042409 on Build GRCh38
rs201301325 Chr.3: 197042418-197042418 on Build GRCh38
rs757250331 Chr.3: 197042572-197042572 on Build GRCh38
rs766678462 Chr.3: 197042416-197042420 on Build GRCh38
rs1017069688 Chr.3: 197042525-197042525 on Build GRCh38
rs943232893 Chr.3: 197042370-197042370 on Build GRCh38
rs923541420 Chr.3: 197042719-197042721 on Build GRCh38
rs1013916725 Chr.3: 197042488-197042488 on Build GRCh38
rs1031106550 Chr.3: 197042679-197042679 on Build GRCh38
rs537355740 Chr.3: 197042320-197042320 on Build GRCh38
rs1036167574 Chr.3: 197042506-197042506 on Build GRCh38
rs976381291 Chr.3: 197042423-197042423 on Build GRCh38
rs909224074 Chr.3: 197042739-197042739 on Build GRCh38
rs956925129 Chr.3: 197042419-197042421 on Build GRCh38
rs140434774 Chr.3: 197042564-197042564 on Build GRCh38
rs1031403149 Chr.3: 197042430-197042430 on Build GRCh38
rs549693245 Chr.3: 197042344-197042344 on Build GRCh38
rs973252152 Chr.3: 197042673-197042673 on Build GRCh38
rs922621307 Chr.3: 197042315-197042315 on Build GRCh38
rs767782806 Chr.3: 197042416-197042418 on Build GRCh38
rs985465657 Chr.3: 197042758-197042758 on Build GRCh38
rs7617835 Chr.3: 197042420-197042420 on Build GRCh38

View Full Product Details