Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 29396974-29397521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR12D2
Entrez Gene ID
26529
Gene Name
olfactory receptor family 12 subfamily D member 2 (gene/pseudogene)
Gene Aliases
DJ994E9.8, HS6M1-20
UniGene
Hs.247862
Gene Chromosome Location
Chr.6: 29396639-29397671 on Build GRCh38
Transcripts
NM_013936.3 XM_011514929.2 XM_011514930.2

Gene Symbol
OR5V1
Entrez Gene ID
81696
Gene Name
olfactory receptor family 5 subfamily V member 1
Gene Aliases
6M1-21, hs6M1-21
UniGene
Hs.666316
Gene Chromosome Location
Chr.6: 29355211-29457067 on Build GRCh38
Transcripts
NM_013936.3 XM_011514929.2 XM_011514930.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6431500 c.629T>C p.F210S Chr.6: 29397328-29397328 on Build GRCh38
3921270 c.792G>A p.E264E Chr.6: 29397491-29397491 on Build GRCh38
5545464 c.324G>A p.T108T Chr.6: 29397023-29397023 on Build GRCh38
5615653 c.608C>A p.T203K Chr.6: 29397307-29397307 on Build GRCh38
3250470 c.342C>T p.A114A Chr.6: 29397041-29397041 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2073151 Chr.6: 29397174-29397174 on Build GRCh38
rs753550222 Chr.6: 29397442-29397442 on Build GRCh38
rs766327077 Chr.6: 29397119-29397119 on Build GRCh38
rs1031473975 Chr.6: 29397115-29397115 on Build GRCh38
rs144667456 Chr.6: 29397421-29397421 on Build GRCh38
rs140378013 Chr.6: 29397145-29397145 on Build GRCh38
rs201460005 Chr.6: 29397302-29397302 on Build GRCh38
rs746643882 Chr.6: 29397365-29397365 on Build GRCh38
rs764326758 Chr.6: 29396997-29396997 on Build GRCh38
rs770255133 Chr.6: 29397382-29397382 on Build GRCh38
rs535902260 Chr.6: 29397261-29397261 on Build GRCh38
rs552924253 Chr.6: 29397295-29397295 on Build GRCh38
rs757746357 Chr.6: 29397009-29397009 on Build GRCh38
rs200560011 Chr.6: 29397287-29397287 on Build GRCh38
rs201917348 Chr.6: 29397225-29397225 on Build GRCh38
rs200807564 Chr.6: 29397224-29397224 on Build GRCh38
rs777127001 Chr.6: 29397305-29397305 on Build GRCh38
rs3128853 Chr.6: 29397010-29397010 on Build GRCh38
rs775339584 Chr.6: 29397187-29397187 on Build GRCh38
rs928122893 Chr.6: 29397303-29397303 on Build GRCh38
rs768195266 Chr.6: 29397186-29397186 on Build GRCh38
rs776880463 Chr.6: 29397374-29397374 on Build GRCh38
rs148576363 Chr.6: 29397485-29397485 on Build GRCh38
rs868419230 Chr.6: 29397477-29397477 on Build GRCh38
rs763241460 Chr.6: 29397317-29397317 on Build GRCh38
rs199879529 Chr.6: 29397221-29397221 on Build GRCh38
rs767949185 Chr.6: 29397207-29397207 on Build GRCh38
rs138029205 Chr.6: 29397194-29397194 on Build GRCh38
rs371147818 Chr.6: 29397256-29397256 on Build GRCh38
rs746747604 Chr.6: 29397149-29397149 on Build GRCh38
rs765400893 Chr.6: 29397159-29397159 on Build GRCh38
rs764288282 Chr.6: 29397320-29397320 on Build GRCh38
rs763069877 Chr.6: 29397315-29397315 on Build GRCh38
rs768158100 Chr.6: 29397268-29397268 on Build GRCh38
rs752856472 Chr.6: 29397232-29397232 on Build GRCh38
rs773043894 Chr.6: 29397288-29397288 on Build GRCh38
rs765252057 Chr.6: 29397307-29397307 on Build GRCh38
rs751041601 Chr.6: 29397017-29397017 on Build GRCh38
rs746000851 Chr.6: 29397220-29397221 on Build GRCh38
rs752805027 Chr.6: 29397311-29397311 on Build GRCh38
rs766751851 Chr.6: 29397381-29397381 on Build GRCh38
rs998622436 Chr.6: 29397103-29397103 on Build GRCh38
rs779038206 Chr.6: 29397348-29397348 on Build GRCh38
rs774082520 Chr.6: 29397204-29397204 on Build GRCh38
rs761573226 Chr.6: 29397206-29397206 on Build GRCh38
rs991601138 Chr.6: 29397074-29397074 on Build GRCh38
rs771268170 Chr.6: 29397372-29397372 on Build GRCh38
rs148199079 Chr.6: 29397095-29397095 on Build GRCh38
rs771761465 Chr.6: 29397363-29397363 on Build GRCh38
rs761158108 Chr.6: 29397211-29397211 on Build GRCh38
rs2073150 Chr.6: 29397464-29397464 on Build GRCh38
rs762410587 Chr.6: 29397059-29397059 on Build GRCh38
rs780200413 Chr.6: 29397028-29397028 on Build GRCh38
rs898016669 Chr.6: 29397024-29397024 on Build GRCh38
rs749425110 Chr.6: 29397030-29397030 on Build GRCh38
rs61742210 Chr.6: 29397423-29397423 on Build GRCh38
rs770876919 Chr.6: 29397181-29397182 on Build GRCh38
rs527311789 Chr.6: 29397420-29397420 on Build GRCh38
rs2073153 Chr.6: 29397058-29397058 on Build GRCh38
rs751848169 Chr.6: 29397322-29397322 on Build GRCh38
rs768119708 Chr.6: 29397195-29397195 on Build GRCh38
rs11752608 Chr.6: 29397094-29397094 on Build GRCh38
rs779179960 Chr.6: 29397146-29397146 on Build GRCh38
rs751922065 Chr.6: 29397007-29397007 on Build GRCh38
rs868614547 Chr.6: 29397449-29397449 on Build GRCh38
rs773747429 Chr.6: 29397209-29397209 on Build GRCh38
rs746782398 Chr.6: 29397042-29397042 on Build GRCh38
rs752746320 Chr.6: 29397453-29397453 on Build GRCh38
rs776544199 Chr.6: 29397048-29397048 on Build GRCh38
rs2073154 Chr.6: 29397038-29397038 on Build GRCh38
rs780950332 Chr.6: 29397209-29397209 on Build GRCh38
rs757911898 Chr.6: 29397454-29397454 on Build GRCh38
rs1040447011 Chr.6: 29397259-29397259 on Build GRCh38
rs372907278 Chr.6: 29397276-29397276 on Build GRCh38
rs977115582 Chr.6: 29397182-29397182 on Build GRCh38
rs866716023 Chr.6: 29397101-29397101 on Build GRCh38
rs113268717 Chr.6: 29397300-29397300 on Build GRCh38
rs200322739 Chr.6: 29397234-29397234 on Build GRCh38
rs376613164 Chr.6: 29397019-29397019 on Build GRCh38
rs762267766 Chr.6: 29397068-29397068 on Build GRCh38
rs113539956 Chr.6: 29397484-29397484 on Build GRCh38
rs748819910 Chr.6: 29397189-29397189 on Build GRCh38
rs36211074 Chr.6: 29397257-29397257 on Build GRCh38
rs549345993 Chr.6: 29397057-29397057 on Build GRCh38
rs745756832 Chr.6: 29397175-29397175 on Build GRCh38
rs747636006 Chr.6: 29397350-29397350 on Build GRCh38
rs764165559 Chr.6: 29397405-29397405 on Build GRCh38
rs886043176 Chr.6: 29397422-29397422 on Build GRCh38
rs756199165 Chr.6: 29397023-29397023 on Build GRCh38
rs372775261 Chr.6: 29397393-29397393 on Build GRCh38
rs753763219 Chr.6: 29397335-29397335 on Build GRCh38
rs201926899 Chr.6: 29397216-29397216 on Build GRCh38
rs770768233 Chr.6: 29397047-29397047 on Build GRCh38
rs2073152 Chr.6: 29397061-29397061 on Build GRCh38
rs753919686 Chr.6: 29397135-29397135 on Build GRCh38
rs755872859 Chr.6: 29397330-29397330 on Build GRCh38
rs745640610 Chr.6: 29397254-29397254 on Build GRCh38
rs775910954 Chr.6: 29397501-29397501 on Build GRCh38
rs768000545 Chr.6: 29397072-29397072 on Build GRCh38
rs778977218 Chr.6: 29397452-29397452 on Build GRCh38
rs112592732 Chr.6: 29397496-29397496 on Build GRCh38
rs77550261 Chr.6: 29397236-29397236 on Build GRCh38
rs746206468 Chr.6: 29397499-29397499 on Build GRCh38
rs756977247 Chr.6: 29397173-29397173 on Build GRCh38
rs779729879 Chr.6: 29397332-29397332 on Build GRCh38
rs552058342 Chr.6: 29397085-29397085 on Build GRCh38
rs754865920 Chr.6: 29397451-29397451 on Build GRCh38
rs751750360 Chr.6: 29397412-29397412 on Build GRCh38
rs1031453659 Chr.6: 29397446-29397446 on Build GRCh38
rs773674392 Chr.6: 29397364-29397364 on Build GRCh38
rs754906725 Chr.6: 29397346-29397346 on Build GRCh38
rs748434337 Chr.6: 29397153-29397153 on Build GRCh38
rs778592864 Chr.6: 29397269-29397269 on Build GRCh38

View Full Product Details