Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 106629377-106629804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
QRSL1
Entrez Gene ID
55278
Gene Name
glutaminyl-tRNA synthase (glutamine-hydrolyzing)-like 1
Gene Aliases
GatA
UniGene
Hs.406917
Gene Chromosome Location
Chr.6: 106629566-106668417 on Build GRCh38
Transcripts
NM_018292.4 XM_011535924.2 NM_001318746.1 NM_032730.5 XM_011536192.2 XM_017011375.1 XM_017011376.1

Gene Symbol
RTN4IP1
Entrez Gene ID
84816
Gene Name
reticulon 4 interacting protein 1
Gene Aliases
NIMP, OPA10
UniGene
Hs.155839
Gene Chromosome Location
Chr.6: 106559237-106630921 on Build GRCh38
Transcripts
NM_018292.4 XM_011535924.2 NM_001318746.1 NM_032730.5 XM_011536192.2 XM_017011375.1 XM_017011376.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6298654 c.3G>T p.M1I Chr.6: 106629684-106629684 on Build GRCh38
3154538 c.7G>A p.G3S Chr.6: 106629688-106629688 on Build GRCh38
6298653 c.3G>T p.M1I Chr.6: 106629684-106629684 on Build GRCh38
4625635 c.7G>A p.G3S Chr.6: 106629688-106629688 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs112247788 Chr.6: 106629443-106629443 on Build GRCh38
rs1003481015 Chr.6: 106629514-106629514 on Build GRCh38
rs892733163 Chr.6: 106629528-106629528 on Build GRCh38
rs930520973 Chr.6: 106629461-106629461 on Build GRCh38
rs377764340 Chr.6: 106629669-106629669 on Build GRCh38
rs536241304 Chr.6: 106629573-106629573 on Build GRCh38
rs755143563 Chr.6: 106629682-106629682 on Build GRCh38
rs1048898863 Chr.6: 106629462-106629462 on Build GRCh38
rs768562253 Chr.6: 106629690-106629690 on Build GRCh38
rs2054365 Chr.6: 106629627-106629627 on Build GRCh38
rs371067222 Chr.6: 106629484-106629484 on Build GRCh38
rs766072877 Chr.6: 106629748-106629748 on Build GRCh38
rs774254711 Chr.6: 106629427-106629427 on Build GRCh38
rs189221387 Chr.6: 106629770-106629770 on Build GRCh38
rs374070818 Chr.6: 106629723-106629723 on Build GRCh38
rs778418291 Chr.6: 106629658-106629658 on Build GRCh38
rs750820793 Chr.6: 106629585-106629585 on Build GRCh38
rs749321658 Chr.6: 106629695-106629695 on Build GRCh38
rs974663453 Chr.6: 106629430-106629430 on Build GRCh38
rs763511451 Chr.6: 106629530-106629530 on Build GRCh38
rs777255423 Chr.6: 106629721-106629721 on Build GRCh38
rs985092097 Chr.6: 106629614-106629614 on Build GRCh38
rs758001176 Chr.6: 106629740-106629740 on Build GRCh38
rs910351597 Chr.6: 106629469-106629469 on Build GRCh38
rs374426112 Chr.6: 106629645-106629645 on Build GRCh38
rs773176842 Chr.6: 106629637-106629637 on Build GRCh38
rs748326430 Chr.6: 106629632-106629633 on Build GRCh38
rs914257152 Chr.6: 106629426-106629426 on Build GRCh38
rs567357808 Chr.6: 106629554-106629554 on Build GRCh38
rs1053738294 Chr.6: 106629768-106629769 on Build GRCh38
rs969515418 Chr.6: 106629540-106629540 on Build GRCh38
rs528876493 Chr.6: 106629405-106629405 on Build GRCh38
rs774304169 Chr.6: 106629701-106629701 on Build GRCh38
rs749967147 Chr.6: 106629730-106629730 on Build GRCh38
rs563550716 Chr.6: 106629783-106629783 on Build GRCh38
rs3747792 Chr.6: 106629675-106629675 on Build GRCh38
rs779785453 Chr.6: 106629681-106629681 on Build GRCh38
rs142692484 Chr.6: 106629702-106629702 on Build GRCh38
rs940123231 Chr.6: 106629450-106629450 on Build GRCh38
rs570669053 Chr.6: 106629725-106629725 on Build GRCh38
rs369690951 Chr.6: 106629639-106629639 on Build GRCh38
rs764475806 Chr.6: 106629715-106629715 on Build GRCh38
rs1035115815 Chr.6: 106629720-106629720 on Build GRCh38
rs376227484 Chr.6: 106629703-106629703 on Build GRCh38
rs761223504 Chr.6: 106629641-106629641 on Build GRCh38
rs777888194 Chr.6: 106629651-106629652 on Build GRCh38
rs1039794794 Chr.6: 106629486-106629486 on Build GRCh38
rs907169023 Chr.6: 106629755-106629755 on Build GRCh38
rs919182604 Chr.6: 106629445-106629445 on Build GRCh38
rs150702171 Chr.6: 106629471-106629471 on Build GRCh38
rs980470698 Chr.6: 106629423-106629423 on Build GRCh38
rs775507799 Chr.6: 106629438-106629438 on Build GRCh38
rs551249260 Chr.6: 106629782-106629782 on Build GRCh38
rs542692517 Chr.6: 106629574-106629574 on Build GRCh38
rs767932701 Chr.6: 106629638-106629638 on Build GRCh38
rs549125657 Chr.6: 106629420-106629420 on Build GRCh38
rs901215638 Chr.6: 106629489-106629489 on Build GRCh38
rs781233024 Chr.6: 106629691-106629691 on Build GRCh38
rs1052831689 Chr.6: 106629525-106629525 on Build GRCh38
rs75903806 Chr.6: 106629482-106629482 on Build GRCh38
rs898504436 Chr.6: 106629452-106629452 on Build GRCh38
rs577143404 Chr.6: 106629502-106629502 on Build GRCh38
rs993628047 Chr.6: 106629732-106629732 on Build GRCh38
rs111609677 Chr.6: 106629722-106629722 on Build GRCh38
rs1038395400 Chr.6: 106629451-106629451 on Build GRCh38
rs761353548 Chr.6: 106629543-106629543 on Build GRCh38
rs758619755 Chr.6: 106629638-106629638 on Build GRCh38
rs960404207 Chr.6: 106629569-106629569 on Build GRCh38
rs1045081096 Chr.6: 106629465-106629465 on Build GRCh38
rs72945092 Chr.6: 106629632-106629632 on Build GRCh38
rs1030688397 Chr.6: 106629647-106629647 on Build GRCh38
rs144848117 Chr.6: 106629642-106629642 on Build GRCh38
rs113968187 Chr.6: 106629435-106629435 on Build GRCh38
rs998191611 Chr.6: 106629491-106629491 on Build GRCh38
rs750036064 Chr.6: 106629666-106629666 on Build GRCh38
rs112005113 Chr.6: 106629628-106629628 on Build GRCh38
rs952014078 Chr.6: 106629599-106629599 on Build GRCh38
rs1011741883 Chr.6: 106629778-106629778 on Build GRCh38
rs3747791 Chr.6: 106629533-106629533 on Build GRCh38
rs559181750 Chr.6: 106629421-106629421 on Build GRCh38
rs761168507 Chr.6: 106629704-106629704 on Build GRCh38
rs772155620 Chr.6: 106629437-106629437 on Build GRCh38
rs373031772 Chr.6: 106629697-106629697 on Build GRCh38
rs759963753 Chr.6: 106629633-106629633 on Build GRCh38
rs747070229 Chr.6: 106629655-106629656 on Build GRCh38
rs938962684 Chr.6: 106629761-106629761 on Build GRCh38

View Full Product Details