Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 66631482-66632038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VCPIP1
Entrez Gene ID
80124
Gene Name
valosin containing protein interacting protein 1
Gene Aliases
DUBA3, VCIP135
UniGene
Hs.632066
Gene Chromosome Location
Chr.8: 66630253-66667217 on Build GRCh38
Transcripts
NM_025054.4

SNP Detail

SNP ID Chromosome Location
rs897410715 Chr.8: 66631926-66631926 on Build GRCh38
rs770785318 Chr.8: 66631761-66631761 on Build GRCh38
rs1005188137 Chr.8: 66631897-66631897 on Build GRCh38
rs887548395 Chr.8: 66631869-66631869 on Build GRCh38
rs946759037 Chr.8: 66631618-66631618 on Build GRCh38
rs957199478 Chr.8: 66631577-66631577 on Build GRCh38
rs532975679 Chr.8: 66631704-66631704 on Build GRCh38
rs910449101 Chr.8: 66631607-66631607 on Build GRCh38
rs772401920 Chr.8: 66631948-66631948 on Build GRCh38
rs961068990 Chr.8: 66631559-66631559 on Build GRCh38
rs978648706 Chr.8: 66631725-66631725 on Build GRCh38
rs985950751 Chr.8: 66631592-66631592 on Build GRCh38
rs991290895 Chr.8: 66632014-66632014 on Build GRCh38
rs560882135 Chr.8: 66631935-66631935 on Build GRCh38
rs796966851 Chr.8: 66631759-66631760 on Build GRCh38
rs1035138168 Chr.8: 66631958-66631958 on Build GRCh38
rs944612459 Chr.8: 66631723-66631723 on Build GRCh38
rs1040714864 Chr.8: 66631758-66631759 on Build GRCh38
rs933720319 Chr.8: 66631829-66631829 on Build GRCh38
rs56682762 Chr.8: 66631760-66631760 on Build GRCh38
rs1014468293 Chr.8: 66632004-66632004 on Build GRCh38
rs972882523 Chr.8: 66631514-66631514 on Build GRCh38
rs566327276 Chr.8: 66631827-66631827 on Build GRCh38
rs550654248 Chr.8: 66631563-66631563 on Build GRCh38
rs916550013 Chr.8: 66631609-66631609 on Build GRCh38
rs960928492 Chr.8: 66631986-66631986 on Build GRCh38
rs560951684 Chr.8: 66631542-66631542 on Build GRCh38
rs941494473 Chr.8: 66631924-66631924 on Build GRCh38
rs1017105469 Chr.8: 66631898-66631898 on Build GRCh38
rs889426036 Chr.8: 66631841-66631841 on Build GRCh38
rs894653180 Chr.8: 66631999-66631999 on Build GRCh38
rs1049517791 Chr.8: 66631833-66631833 on Build GRCh38
rs778719301 Chr.8: 66631769-66631769 on Build GRCh38
rs570185987 Chr.8: 66631644-66631644 on Build GRCh38
rs1024284936 Chr.8: 66631537-66631537 on Build GRCh38
rs745681731 Chr.8: 66631784-66631784 on Build GRCh38

View Full Product Details