Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 13110022-13110618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OPTN
Entrez Gene ID
10133
Gene Name
optineurin
Gene Aliases
ALS12, FIP2, GLC1E, HIP7, HYPL, NRP, TFIIIA-INTP
UniGene
Hs.332706
Gene Chromosome Location
Chr.10: 13100082-13138276 on Build GRCh38
Transcripts
NM_001008211.1 NM_001008212.1 NM_001008213.1 NM_021980.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3997844 c.293T>A p.M98K Chr.10: 13110400-13110400 on Build GRCh38
3414821 c.315G>T p.E105D Chr.10: 13110422-13110422 on Build GRCh38
6561532 c.259G>C p.E87Q Chr.10: 13110366-13110366 on Build GRCh38
4444301 c.257T>A p.F86Y Chr.10: 13110364-13110364 on Build GRCh38
276446 c.248G>A p.R83H Chr.10: 13110355-13110355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1020576559 Chr.10: 13110524-13110524 on Build GRCh38
rs756622651 Chr.10: 13110354-13110354 on Build GRCh38
rs989063297 Chr.10: 13110222-13110222 on Build GRCh38
rs978988466 Chr.10: 13110547-13110547 on Build GRCh38
rs764979306 Chr.10: 13110417-13110417 on Build GRCh38
rs1007297279 Chr.10: 13110085-13110085 on Build GRCh38
rs181141625 Chr.10: 13110393-13110393 on Build GRCh38
rs753185069 Chr.10: 13110436-13110437 on Build GRCh38
rs758707459 Chr.10: 13110461-13110462 on Build GRCh38
rs200047182 Chr.10: 13110516-13110516 on Build GRCh38
rs60221241 Chr.10: 13110124-13110124 on Build GRCh38
rs10047381 Chr.10: 13110295-13110295 on Build GRCh38
rs779887367 Chr.10: 13110516-13110517 on Build GRCh38
rs184561087 Chr.10: 13110394-13110394 on Build GRCh38
rs767361319 Chr.10: 13110208-13110208 on Build GRCh38
rs765891109 Chr.10: 13110249-13110249 on Build GRCh38
rs761092487 Chr.10: 13110519-13110519 on Build GRCh38
rs951364762 Chr.10: 13110184-13110184 on Build GRCh38
rs750177059 Chr.10: 13110274-13110274 on Build GRCh38
rs113349726 Chr.10: 13110478-13110478 on Build GRCh38
rs550305453 Chr.10: 13110506-13110507 on Build GRCh38
rs969356738 Chr.10: 13110538-13110538 on Build GRCh38
rs780313745 Chr.10: 13110359-13110359 on Build GRCh38
rs752001857 Chr.10: 13110325-13110325 on Build GRCh38
rs79529484 Chr.10: 13110515-13110515 on Build GRCh38
rs947213211 Chr.10: 13110381-13110381 on Build GRCh38
rs902890138 Chr.10: 13110045-13110045 on Build GRCh38
rs398096802 Chr.10: 13110516-13110516 on Build GRCh38
rs781755765 Chr.10: 13110334-13110334 on Build GRCh38
rs928750477 Chr.10: 13110271-13110271 on Build GRCh38
rs754013605 Chr.10: 13110472-13110472 on Build GRCh38
rs1013464707 Chr.10: 13110094-13110094 on Build GRCh38
rs759022584 Chr.10: 13110414-13110414 on Build GRCh38
rs11819428 Chr.10: 13110533-13110533 on Build GRCh38
rs767061853 Chr.10: 13110520-13110520 on Build GRCh38
rs767786342 Chr.10: 13110459-13110459 on Build GRCh38
rs75617318 Chr.10: 13110514-13110514 on Build GRCh38
rs761806855 Chr.10: 13110134-13110134 on Build GRCh38
rs766971765 Chr.10: 13110467-13110467 on Build GRCh38
rs757528160 Chr.10: 13110333-13110333 on Build GRCh38
rs758583995 Chr.10: 13110482-13110482 on Build GRCh38
rs947592811 Chr.10: 13110344-13110344 on Build GRCh38
rs937486597 Chr.10: 13110589-13110589 on Build GRCh38
rs922923097 Chr.10: 13110141-13110141 on Build GRCh38
rs760240571 Chr.10: 13110247-13110247 on Build GRCh38
rs562730214 Chr.10: 13110217-13110217 on Build GRCh38
rs11258194 Chr.10: 13110400-13110400 on Build GRCh38
rs530316633 Chr.10: 13110506-13110506 on Build GRCh38
rs780736613 Chr.10: 13110504-13110505 on Build GRCh38
rs778173490 Chr.10: 13110502-13110505 on Build GRCh38
rs745448084 Chr.10: 13110503-13110503 on Build GRCh38
rs1016507326 Chr.10: 13110475-13110475 on Build GRCh38
rs201978271 Chr.10: 13110507-13110507 on Build GRCh38
rs374267986 Chr.10: 13110487-13110487 on Build GRCh38
rs538362037 Chr.10: 13110057-13110057 on Build GRCh38
rs898674543 Chr.10: 13110529-13110529 on Build GRCh38
rs759214569 Chr.10: 13110267-13110267 on Build GRCh38
rs558573187 Chr.10: 13110081-13110081 on Build GRCh38
rs895824243 Chr.10: 13110387-13110387 on Build GRCh38
rs549625644 Chr.10: 13110506-13110506 on Build GRCh38
rs753273532 Chr.10: 13110251-13110251 on Build GRCh38
rs775000021 Chr.10: 13110425-13110425 on Build GRCh38
rs554936728 Chr.10: 13110121-13110121 on Build GRCh38
rs766200278 Chr.10: 13110374-13110374 on Build GRCh38
rs950574109 Chr.10: 13110100-13110100 on Build GRCh38
rs745395582 Chr.10: 13110505-13110507 on Build GRCh38
rs1057059212 Chr.10: 13110075-13110075 on Build GRCh38
rs76776236 Chr.10: 13110574-13110574 on Build GRCh38
rs1026148878 Chr.10: 13110098-13110098 on Build GRCh38
rs147274458 Chr.10: 13110373-13110373 on Build GRCh38
rs572290743 Chr.10: 13110093-13110093 on Build GRCh38
rs760492259 Chr.10: 13110370-13110370 on Build GRCh38
rs371927112 Chr.10: 13110451-13110451 on Build GRCh38
rs961818154 Chr.10: 13110263-13110263 on Build GRCh38
rs754036757 Chr.10: 13110383-13110383 on Build GRCh38
rs913462685 Chr.10: 13110272-13110272 on Build GRCh38
rs531691395 Chr.10: 13110338-13110338 on Build GRCh38
rs756712179 Chr.10: 13110462-13110462 on Build GRCh38
rs545373466 Chr.10: 13110505-13110506 on Build GRCh38
rs954325019 Chr.10: 13110152-13110152 on Build GRCh38
rs1006078948 Chr.10: 13110420-13110420 on Build GRCh38
rs750777534 Chr.10: 13110460-13110460 on Build GRCh38
rs60399947 Chr.10: 13110117-13110117 on Build GRCh38
rs1006440917 Chr.10: 13110106-13110106 on Build GRCh38
rs762516858 Chr.10: 13110508-13110508 on Build GRCh38
rs997103923 Chr.10: 13110071-13110071 on Build GRCh38
rs34617995 Chr.10: 13110505-13110505 on Build GRCh38
rs202044898 Chr.10: 13110382-13110382 on Build GRCh38
rs765679498 Chr.10: 13110359-13110360 on Build GRCh38
rs543054978 Chr.10: 13110166-13110166 on Build GRCh38
rs776138037 Chr.10: 13110402-13110402 on Build GRCh38
rs749333165 Chr.10: 13110368-13110368 on Build GRCh38
rs959069574 Chr.10: 13110307-13110307 on Build GRCh38
rs764745062 Chr.10: 13110273-13110273 on Build GRCh38
rs750952641 Chr.10: 13110335-13110335 on Build GRCh38
rs567649467 Chr.10: 13110488-13110488 on Build GRCh38
rs188882035 Chr.10: 13110474-13110474 on Build GRCh38
rs773021961 Chr.10: 13110231-13110231 on Build GRCh38

View Full Product Details