Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 88914207-88914641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAMBPL1
Entrez Gene ID
57559
Gene Name
STAM binding protein like 1
Gene Aliases
ALMalpha, AMSH-FP, AMSH-LP, bA399O19.2
UniGene
Hs.16229
Gene Chromosome Location
Chr.10: 88879368-88923502 on Build GRCh38
Transcripts
NM_020799.3 XM_006717928.2 XM_006717930.3 XM_011539983.1 XM_011539985.1 XM_017016453.1 XM_017016454.1 XM_017016455.1 XM_017016456.1 XM_017016457.1 XM_017016458.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2153631 c.883G>T p.G295* Chr.10: 88914638-88914638 on Build GRCh38
5442259 c.886A>G p.I296V Chr.10: 88914641-88914641 on Build GRCh38
5630978 c.800G>A p.R267Q Chr.10: 88914555-88914555 on Build GRCh38
6243436 c.846G>T p.L282L Chr.10: 88914601-88914601 on Build GRCh38
5442258 c.886A>G p.I296V Chr.10: 88914641-88914641 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200651856 Chr.10: 88914555-88914555 on Build GRCh38
rs758305864 Chr.10: 88914576-88914576 on Build GRCh38
rs977591082 Chr.10: 88914346-88914346 on Build GRCh38
rs538518690 Chr.10: 88914554-88914554 on Build GRCh38
rs943413350 Chr.10: 88914470-88914470 on Build GRCh38
rs771385050 Chr.10: 88914598-88914598 on Build GRCh38
rs747782167 Chr.10: 88914593-88914593 on Build GRCh38
rs765045857 Chr.10: 88914571-88914571 on Build GRCh38
rs928972713 Chr.10: 88914334-88914334 on Build GRCh38
rs943302153 Chr.10: 88914274-88914274 on Build GRCh38
rs11202889 Chr.10: 88914494-88914494 on Build GRCh38
rs746285008 Chr.10: 88914614-88914614 on Build GRCh38
rs868537874 Chr.10: 88914583-88914583 on Build GRCh38
rs1024597347 Chr.10: 88914301-88914301 on Build GRCh38
rs750257823 Chr.10: 88914574-88914574 on Build GRCh38
rs76371936 Chr.10: 88914486-88914486 on Build GRCh38
rs1014379460 Chr.10: 88914373-88914373 on Build GRCh38
rs760164456 Chr.10: 88914235-88914235 on Build GRCh38
rs373894376 Chr.10: 88914588-88914588 on Build GRCh38
rs955491685 Chr.10: 88914545-88914545 on Build GRCh38
rs114521067 Chr.10: 88914493-88914493 on Build GRCh38
rs990203082 Chr.10: 88914443-88914443 on Build GRCh38
rs761546991 Chr.10: 88914557-88914557 on Build GRCh38
rs559832088 Chr.10: 88914266-88914266 on Build GRCh38
rs754446775 Chr.10: 88914584-88914584 on Build GRCh38
rs542215774 Chr.10: 88914342-88914342 on Build GRCh38
rs1047968702 Chr.10: 88914347-88914347 on Build GRCh38
rs766686835 Chr.10: 88914290-88914290 on Build GRCh38
rs749604212 Chr.10: 88914495-88914495 on Build GRCh38
rs140951805 Chr.10: 88914234-88914234 on Build GRCh38
rs760155767 Chr.10: 88914525-88914525 on Build GRCh38
rs774953336 Chr.10: 88914522-88914522 on Build GRCh38
rs779911647 Chr.10: 88914578-88914578 on Build GRCh38
rs779392226 Chr.10: 88914611-88914611 on Build GRCh38
rs957414596 Chr.10: 88914417-88914417 on Build GRCh38
rs906010124 Chr.10: 88914463-88914463 on Build GRCh38
rs541154469 Chr.10: 88914265-88914265 on Build GRCh38
rs563690477 Chr.10: 88914352-88914352 on Build GRCh38
rs976386575 Chr.10: 88914473-88914473 on Build GRCh38
rs941725126 Chr.10: 88914570-88914570 on Build GRCh38
rs751071561 Chr.10: 88914580-88914580 on Build GRCh38
rs113516829 Chr.10: 88914249-88914249 on Build GRCh38
rs747831350 Chr.10: 88914526-88914526 on Build GRCh38
rs964494874 Chr.10: 88914585-88914585 on Build GRCh38
rs772747163 Chr.10: 88914528-88914528 on Build GRCh38
rs199673105 Chr.10: 88914508-88914508 on Build GRCh38
rs1013165363 Chr.10: 88914297-88914297 on Build GRCh38
rs778766063 Chr.10: 88914518-88914519 on Build GRCh38
rs369751532 Chr.10: 88914536-88914536 on Build GRCh38
rs1032277307 Chr.10: 88914386-88914386 on Build GRCh38
rs771440392 Chr.10: 88914520-88914520 on Build GRCh38
rs139363999 Chr.10: 88914539-88914539 on Build GRCh38
rs866364221 Chr.10: 88914602-88914602 on Build GRCh38
rs754123978 Chr.10: 88914407-88914407 on Build GRCh38
rs910546803 Chr.10: 88914457-88914457 on Build GRCh38
rs1023154785 Chr.10: 88914436-88914436 on Build GRCh38
rs971952086 Chr.10: 88914302-88914302 on Build GRCh38
rs76962149 Chr.10: 88914272-88914272 on Build GRCh38

View Full Product Details