Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 96343637-96344131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OPALIN
Entrez Gene ID
93377
Gene Name
oligodendrocytic myelin paranodal and inner loop protein
Gene Aliases
HTMP10, TMEM10, TMP10
UniGene
Hs.12449
Gene Chromosome Location
Chr.10: 96343216-96359365 on Build GRCh38
Transcripts
NM_001040102.2 NM_001040103.2 NM_001284320.1 NM_001284321.1 NM_001284322.1 NM_001284323.1 NM_001284324.1 NM_001284326.1 NM_001284327.1 NM_033207.4 NR_104296.1 NR_104297.1

SNP Detail

SNP ID Chromosome Location
rs888642577 Chr.10: 96344059-96344059 on Build GRCh38
rs74153642 Chr.10: 96343868-96343868 on Build GRCh38
rs934295443 Chr.10: 96343901-96343901 on Build GRCh38
rs747087367 Chr.10: 96343859-96343860 on Build GRCh38
rs546177067 Chr.10: 96343680-96343680 on Build GRCh38
rs139967785 Chr.10: 96343807-96343807 on Build GRCh38
rs1052072990 Chr.10: 96344024-96344024 on Build GRCh38
rs903398092 Chr.10: 96343969-96343969 on Build GRCh38
rs3814225 Chr.10: 96343954-96343954 on Build GRCh38
rs935001641 Chr.10: 96343986-96343986 on Build GRCh38
rs115933470 Chr.10: 96344065-96344065 on Build GRCh38
rs566734367 Chr.10: 96344050-96344050 on Build GRCh38
rs975577475 Chr.10: 96343866-96343866 on Build GRCh38
rs769287898 Chr.10: 96343853-96343853 on Build GRCh38
rs559935093 Chr.10: 96343752-96343752 on Build GRCh38
rs956500908 Chr.10: 96343865-96343865 on Build GRCh38
rs944059037 Chr.10: 96344000-96344000 on Build GRCh38
rs530879621 Chr.10: 96343821-96343821 on Build GRCh38
rs755463624 Chr.10: 96344026-96344026 on Build GRCh38
rs372105180 Chr.10: 96343757-96343757 on Build GRCh38
rs947477178 Chr.10: 96343924-96343924 on Build GRCh38
rs180681737 Chr.10: 96343792-96343792 on Build GRCh38
rs981989275 Chr.10: 96343817-96343817 on Build GRCh38
rs191382229 Chr.10: 96344001-96344001 on Build GRCh38
rs909976790 Chr.10: 96343779-96343779 on Build GRCh38
rs144676884 Chr.10: 96344052-96344052 on Build GRCh38
rs10218881 Chr.10: 96343776-96343776 on Build GRCh38
rs149779132 Chr.10: 96344037-96344037 on Build GRCh38

View Full Product Details