Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64284328-64284900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAD
Entrez Gene ID
572
Gene Name
BCL2 associated agonist of cell death
Gene Aliases
BBC2, BCL2L8
UniGene
Hs.370254
Gene Chromosome Location
Chr.11: 64269828-64284704 on Build GRCh38
Transcripts
NM_004322.3 NM_032989.2 NM_001170726.1 NM_001170881.1 NM_001177358.1 XM_005274101.2 XM_011545168.2 XM_011545170.2 XM_011545171.2 XM_011545172.2 XM_017018014.1 XM_017018016.1

Gene Symbol
GPR137
Entrez Gene ID
56834
Gene Name
G protein-coupled receptor 137
Gene Aliases
C11orf4, GPR137A, TM7SF1L1
UniGene
Hs.523763
Gene Chromosome Location
Chr.11: 64270337-64289500 on Build GRCh38
Transcripts
NM_004322.3 NM_032989.2 NM_001170726.1 NM_001170881.1 NM_001177358.1 XM_005274101.2 XM_011545168.2 XM_011545170.2 XM_011545171.2 XM_011545172.2 XM_017018014.1 XM_017018016.1

SNP Detail

SNP ID Chromosome Location
rs771524361 Chr.11: 64284420-64284420 on Build GRCh38
rs568391428 Chr.11: 64284615-64284615 on Build GRCh38
rs1019152940 Chr.11: 64284579-64284579 on Build GRCh38
rs761233693 Chr.11: 64284414-64284414 on Build GRCh38
rs768868726 Chr.11: 64284415-64284415 on Build GRCh38
rs775022318 Chr.11: 64284450-64284450 on Build GRCh38
rs957783243 Chr.11: 64284625-64284626 on Build GRCh38
rs981928957 Chr.11: 64284652-64284652 on Build GRCh38
rs1009076116 Chr.11: 64284519-64284519 on Build GRCh38
rs372868356 Chr.11: 64284418-64284418 on Build GRCh38
rs903418016 Chr.11: 64284597-64284597 on Build GRCh38
rs995487552 Chr.11: 64284795-64284795 on Build GRCh38
rs920518344 Chr.11: 64284833-64284833 on Build GRCh38
rs900737074 Chr.11: 64284737-64284737 on Build GRCh38
rs773671122 Chr.11: 64284423-64284423 on Build GRCh38
rs930706458 Chr.11: 64284677-64284677 on Build GRCh38
rs987170142 Chr.11: 64284716-64284716 on Build GRCh38
rs540148087 Chr.11: 64284604-64284604 on Build GRCh38
rs751467516 Chr.11: 64284487-64284487 on Build GRCh38
rs5031004 Chr.11: 64284836-64284836 on Build GRCh38
rs963294098 Chr.11: 64284384-64284384 on Build GRCh38
rs777079956 Chr.11: 64284434-64284435 on Build GRCh38
rs991177813 Chr.11: 64284817-64284817 on Build GRCh38
rs886640711 Chr.11: 64284470-64284470 on Build GRCh38
rs1055115245 Chr.11: 64284660-64284660 on Build GRCh38
rs746057329 Chr.11: 64284394-64284394 on Build GRCh38
rs1037263072 Chr.11: 64284624-64284624 on Build GRCh38
rs1043346730 Chr.11: 64284549-64284549 on Build GRCh38
rs780368700 Chr.11: 64284525-64284525 on Build GRCh38
rs772530289 Chr.11: 64284422-64284422 on Build GRCh38
rs1027982025 Chr.11: 64284766-64284766 on Build GRCh38
rs987526821 Chr.11: 64284433-64284433 on Build GRCh38
rs752307890 Chr.11: 64284355-64284355 on Build GRCh38
rs201614479 Chr.11: 64284364-64284365 on Build GRCh38
rs1009043121 Chr.11: 64284729-64284729 on Build GRCh38
rs764878897 Chr.11: 64284431-64284431 on Build GRCh38
rs1028432318 Chr.11: 64284620-64284620 on Build GRCh38
rs966478908 Chr.11: 64284640-64284640 on Build GRCh38
rs652611 Chr.11: 64284834-64284834 on Build GRCh38
rs368562037 Chr.11: 64284398-64284398 on Build GRCh38
rs1057435606 Chr.11: 64284609-64284609 on Build GRCh38
rs28417526 Chr.11: 64284687-64284687 on Build GRCh38
rs947553266 Chr.11: 64284536-64284536 on Build GRCh38
rs556266858 Chr.11: 64284643-64284643 on Build GRCh38
rs932178667 Chr.11: 64284866-64284866 on Build GRCh38
rs755069330 Chr.11: 64284691-64284691 on Build GRCh38
rs748639671 Chr.11: 64284771-64284771 on Build GRCh38
rs536241063 Chr.11: 64284637-64284637 on Build GRCh38
rs975562595 Chr.11: 64284512-64284512 on Build GRCh38
rs75052600 Chr.11: 64284381-64284381 on Build GRCh38
rs371971125 Chr.11: 64284575-64284575 on Build GRCh38
rs752859657 Chr.11: 64284750-64284762 on Build GRCh38
rs866025616 Chr.11: 64284689-64284689 on Build GRCh38
rs767287994 Chr.11: 64284633-64284633 on Build GRCh38
rs927810128 Chr.11: 64284805-64284805 on Build GRCh38
rs762479156 Chr.11: 64284465-64284465 on Build GRCh38
rs757234728 Chr.11: 64284632-64284632 on Build GRCh38
rs937954688 Chr.11: 64284655-64284655 on Build GRCh38
rs1047921787 Chr.11: 64284443-64284443 on Build GRCh38
rs772761198 Chr.11: 64284772-64284772 on Build GRCh38
rs943095812 Chr.11: 64284475-64284475 on Build GRCh38
rs911666324 Chr.11: 64284462-64284462 on Build GRCh38
rs994945721 Chr.11: 64284437-64284437 on Build GRCh38
rs751524539 Chr.11: 64284386-64284386 on Build GRCh38
rs781006183 Chr.11: 64284392-64284392 on Build GRCh38
rs944798802 Chr.11: 64284846-64284846 on Build GRCh38
rs780777733 Chr.11: 64284701-64284701 on Build GRCh38
rs376064584 Chr.11: 64284395-64284395 on Build GRCh38
rs1039101376 Chr.11: 64284365-64284375 on Build GRCh38
rs1012971065 Chr.11: 64284621-64284621 on Build GRCh38
rs749321296 Chr.11: 64284397-64284397 on Build GRCh38
rs895916201 Chr.11: 64284612-64284612 on Build GRCh38
rs779169430 Chr.11: 64284761-64284761 on Build GRCh38
rs989225711 Chr.11: 64284634-64284634 on Build GRCh38
rs111793546 Chr.11: 64284800-64284800 on Build GRCh38
rs940184560 Chr.11: 64284448-64284448 on Build GRCh38
rs375058825 Chr.11: 64284769-64284769 on Build GRCh38
rs757853078 Chr.11: 64284373-64284373 on Build GRCh38
rs545065054 Chr.11: 64284669-64284669 on Build GRCh38
rs889834107 Chr.11: 64284808-64284808 on Build GRCh38
rs576112604 Chr.11: 64284654-64284654 on Build GRCh38
rs921391590 Chr.11: 64284529-64284529 on Build GRCh38
rs965347800 Chr.11: 64284589-64284589 on Build GRCh38
rs920630421 Chr.11: 64284671-64284671 on Build GRCh38
rs375119597 Chr.11: 64284417-64284417 on Build GRCh38
rs936449293 Chr.11: 64284366-64284366 on Build GRCh38
rs563409227 Chr.11: 64284869-64284869 on Build GRCh38
rs763629672 Chr.11: 64284476-64284476 on Build GRCh38
rs1040953380 Chr.11: 64284731-64284731 on Build GRCh38
rs534613882 Chr.11: 64284568-64284568 on Build GRCh38
rs34882006 Chr.11: 64284351-64284351 on Build GRCh38
rs763067502 Chr.11: 64284800-64284801 on Build GRCh38
rs543422577 Chr.11: 64284856-64284856 on Build GRCh38
rs558907072 Chr.11: 64284685-64284685 on Build GRCh38
rs371764981 Chr.11: 64284391-64284391 on Build GRCh38
rs541370892 Chr.11: 64284811-64284811 on Build GRCh38
rs979318645 Chr.11: 64284826-64284826 on Build GRCh38
rs773441698 Chr.11: 64284782-64284782 on Build GRCh38
rs375409072 Chr.11: 64284350-64284350 on Build GRCh38
rs981954375 Chr.11: 64284802-64284802 on Build GRCh38
rs543991860 Chr.11: 64284696-64284704 on Build GRCh38
rs957668799 Chr.11: 64284768-64284768 on Build GRCh38
rs572555375 Chr.11: 64284801-64284801 on Build GRCh38
rs1010531924 Chr.11: 64284770-64284770 on Build GRCh38
rs756252556 Chr.11: 64284692-64284692 on Build GRCh38
rs748056064 Chr.11: 64284713-64284713 on Build GRCh38
rs968884764 Chr.11: 64284820-64284820 on Build GRCh38
rs1022775163 Chr.11: 64284383-64284383 on Build GRCh38
rs917295477 Chr.11: 64284880-64284880 on Build GRCh38

View Full Product Details