Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 36017713-36018137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL23
Entrez Gene ID
6368
Gene Name
C-C motif chemokine ligand 23
Gene Aliases
CK-BETA-8, CKb8, Ckb-8, Ckb-8-1, MIP-3, MIP3, MPIF-1, SCYA23, hmrp-2a
UniGene
Hs.169191
Gene Chromosome Location
Chr.17: 36013056-36018179 on Build GRCh38
Transcripts
NM_005064.5 NM_145898.3

SNP Detail

SNP ID Chromosome Location
rs769009492 Chr.17: 36017834-36017834 on Build GRCh38
rs763176287 Chr.17: 36017829-36017829 on Build GRCh38
rs998377293 Chr.17: 36018078-36018078 on Build GRCh38
rs1006984545 Chr.17: 36018039-36018039 on Build GRCh38
rs534083505 Chr.17: 36018090-36018090 on Build GRCh38
rs566759893 Chr.17: 36018084-36018084 on Build GRCh38
rs774564379 Chr.17: 36017840-36017840 on Build GRCh38
rs1032013475 Chr.17: 36017755-36017755 on Build GRCh38
rs777411727 Chr.17: 36017910-36017910 on Build GRCh38
rs369023759 Chr.17: 36017904-36017904 on Build GRCh38
rs766146131 Chr.17: 36017868-36017868 on Build GRCh38
rs983536452 Chr.17: 36017861-36017861 on Build GRCh38
rs1051516 Chr.17: 36017865-36017865 on Build GRCh38
rs775705980 Chr.17: 36017814-36017814 on Build GRCh38
rs770031075 Chr.17: 36017808-36017808 on Build GRCh38
rs759487613 Chr.17: 36017885-36017885 on Build GRCh38
rs1800535 Chr.17: 36018015-36018015 on Build GRCh38
rs200802399 Chr.17: 36017867-36017867 on Build GRCh38
rs999240133 Chr.17: 36017748-36017748 on Build GRCh38
rs957783068 Chr.17: 36017764-36017764 on Build GRCh38
rs978875408 Chr.17: 36018110-36018110 on Build GRCh38
rs991096403 Chr.17: 36017774-36017774 on Build GRCh38
rs111879665 Chr.17: 36017924-36017924 on Build GRCh38
rs929256670 Chr.17: 36017847-36017847 on Build GRCh38
rs568579695 Chr.17: 36017942-36017942 on Build GRCh38
rs111373433 Chr.17: 36017959-36017959 on Build GRCh38
rs201639291 Chr.17: 36017895-36017895 on Build GRCh38
rs766654541 Chr.17: 36017831-36017831 on Build GRCh38
rs201394186 Chr.17: 36017783-36017783 on Build GRCh38
rs1023456825 Chr.17: 36017788-36017788 on Build GRCh38
rs537571761 Chr.17: 36017957-36017957 on Build GRCh38
rs185517547 Chr.17: 36017784-36017784 on Build GRCh38
rs937107573 Chr.17: 36017845-36017845 on Build GRCh38
rs888491228 Chr.17: 36018026-36018027 on Build GRCh38
rs893577290 Chr.17: 36018081-36018081 on Build GRCh38
rs753766586 Chr.17: 36017877-36017877 on Build GRCh38
rs138184965 Chr.17: 36017832-36017832 on Build GRCh38
rs925729072 Chr.17: 36017786-36017786 on Build GRCh38
rs757847589 Chr.17: 36017903-36017903 on Build GRCh38
rs971073889 Chr.17: 36017815-36017815 on Build GRCh38
rs375886133 Chr.17: 36017886-36017886 on Build GRCh38
rs371466808 Chr.17: 36017923-36017923 on Build GRCh38
rs933581414 Chr.17: 36017929-36017929 on Build GRCh38
rs548121653 Chr.17: 36018057-36018057 on Build GRCh38
rs2072667 Chr.17: 36017925-36017925 on Build GRCh38
rs753281119 Chr.17: 36018005-36018005 on Build GRCh38
rs140430245 Chr.17: 36017833-36017833 on Build GRCh38
rs999188030 Chr.17: 36018052-36018052 on Build GRCh38
rs372628580 Chr.17: 36017772-36017772 on Build GRCh38

View Full Product Details